Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 42164902-42165420 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP3M2
Entrez Gene ID
10947
Gene Name
adaptor related protein complex 3 mu 2 subunit
Gene Aliases
AP47B, CLA20, P47B
UniGene
Hs.654529
Gene Chromosome Location
Chr.8: 42152946-42171183 on Build GRCh38
Transcripts
NM_001134296.1 NM_006803.3 XM_017012977.1 XM_017012978.1 XM_017012979.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1099970 c.650A>C p.D217A Chr.8: 42165137-42165137 on Build GRCh38
1727282 c.584-2A>G p.? Chr.8: 42165069-42165069 on Build GRCh38
1099971 c.663C>A p.S221S Chr.8: 42165150-42165150 on Build GRCh38
5874707 c.645G>A p.M215I Chr.8: 42165132-42165132 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1050698301 Chr.8: 42165227-42165227 on Build GRCh38
rs994178051 Chr.8: 42165270-42165270 on Build GRCh38
rs751298834 Chr.8: 42165037-42165037 on Build GRCh38
rs558099497 Chr.8: 42165243-42165243 on Build GRCh38
rs527556409 Chr.8: 42164985-42164985 on Build GRCh38
rs759277553 Chr.8: 42165164-42165164 on Build GRCh38
rs771396560 Chr.8: 42165149-42165149 on Build GRCh38
rs753233042 Chr.8: 42165100-42165100 on Build GRCh38
rs778072806 Chr.8: 42165022-42165023 on Build GRCh38
rs374989996 Chr.8: 42165384-42165384 on Build GRCh38
rs34420240 Chr.8: 42165059-42165059 on Build GRCh38
rs770752372 Chr.8: 42165148-42165148 on Build GRCh38
rs767042375 Chr.8: 42165045-42165045 on Build GRCh38
rs762934642 Chr.8: 42165129-42165129 on Build GRCh38
rs76570663 Chr.8: 42165228-42165228 on Build GRCh38
rs768676669 Chr.8: 42165020-42165020 on Build GRCh38
rs202115677 Chr.8: 42165087-42165087 on Build GRCh38
rs755705157 Chr.8: 42165060-42165060 on Build GRCh38
rs958896907 Chr.8: 42165373-42165373 on Build GRCh38
rs1019656116 Chr.8: 42164952-42164952 on Build GRCh38
rs989847927 Chr.8: 42165349-42165349 on Build GRCh38
rs747546329 Chr.8: 42165078-42165078 on Build GRCh38
rs371601370 Chr.8: 42165149-42165149 on Build GRCh38
rs773147874 Chr.8: 42165023-42165023 on Build GRCh38
rs777357730 Chr.8: 42165061-42165061 on Build GRCh38
rs755177037 Chr.8: 42165387-42165387 on Build GRCh38
rs781297965 Chr.8: 42165391-42165391 on Build GRCh38
rs754872795 Chr.8: 42165006-42165006 on Build GRCh38
rs750893273 Chr.8: 42165379-42165379 on Build GRCh38
rs747178641 Chr.8: 42165109-42165109 on Build GRCh38
rs562428423 Chr.8: 42165313-42165314 on Build GRCh38
rs904439530 Chr.8: 42165215-42165215 on Build GRCh38
rs200936959 Chr.8: 42165169-42165169 on Build GRCh38
rs776687190 Chr.8: 42165113-42165113 on Build GRCh38
rs758705138 Chr.8: 42165380-42165380 on Build GRCh38
rs201236735 Chr.8: 42165103-42165103 on Build GRCh38
rs753381401 Chr.8: 42165184-42165184 on Build GRCh38
rs762809479 Chr.8: 42165024-42165024 on Build GRCh38
rs748194852 Chr.8: 42165396-42165396 on Build GRCh38
rs937102155 Chr.8: 42165144-42165144 on Build GRCh38
rs921168912 Chr.8: 42165355-42165355 on Build GRCh38
rs746190523 Chr.8: 42165099-42165099 on Build GRCh38
rs571582369 Chr.8: 42164970-42164970 on Build GRCh38
rs148764977 Chr.8: 42165083-42165083 on Build GRCh38
rs767291186 Chr.8: 42165165-42165165 on Build GRCh38
rs1025606096 Chr.8: 42165295-42165295 on Build GRCh38
rs754465510 Chr.8: 42165079-42165079 on Build GRCh38
rs555414398 Chr.8: 42165267-42165267 on Build GRCh38
rs765695596 Chr.8: 42165200-42165200 on Build GRCh38
rs1000094092 Chr.8: 42165220-42165220 on Build GRCh38
rs932465202 Chr.8: 42165146-42165146 on Build GRCh38
rs781213705 Chr.8: 42165010-42165010 on Build GRCh38
rs368506487 Chr.8: 42165105-42165105 on Build GRCh38
rs554780824 Chr.8: 42165299-42165299 on Build GRCh38
rs760436898 Chr.8: 42164938-42164939 on Build GRCh38
rs150897190 Chr.8: 42165314-42165315 on Build GRCh38
rs898505439 Chr.8: 42165190-42165190 on Build GRCh38
rs543951450 Chr.8: 42165329-42165329 on Build GRCh38
rs144255322 Chr.8: 42165166-42165166 on Build GRCh38
rs139629942 Chr.8: 42164963-42164963 on Build GRCh38
rs770474750 Chr.8: 42165250-42165250 on Build GRCh38
rs572431574 Chr.8: 42165261-42165261 on Build GRCh38
rs768728193 Chr.8: 42165110-42165110 on Build GRCh38
rs148686139 Chr.8: 42165174-42165174 on Build GRCh38
rs1019708977 Chr.8: 42165266-42165266 on Build GRCh38
rs574933353 Chr.8: 42165328-42165328 on Build GRCh38
rs759281307 Chr.8: 42165044-42165044 on Build GRCh38
rs780400687 Chr.8: 42165381-42165381 on Build GRCh38
rs745407754 Chr.8: 42165396-42165398 on Build GRCh38
rs780822823 Chr.8: 42165385-42165385 on Build GRCh38
rs1054374844 Chr.8: 42164931-42164931 on Build GRCh38
rs766226828 Chr.8: 42165028-42165028 on Build GRCh38
rs757883026 Chr.8: 42165197-42165197 on Build GRCh38

View Full Product Details