Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 1857820-1858362 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF10
Entrez Gene ID
9639
Gene Name
Rho guanine nucleotide exchange factor 10
Gene Aliases
GEF10, SNCV
UniGene
Hs.98594
Gene Chromosome Location
Chr.8: 1823976-1958641 on Build GRCh38
Transcripts
NM_001308152.1 NM_001308153.1 NM_014629.3 XM_005266041.3 XM_011534767.2 XM_011534768.1 XM_011534770.2 XM_017014003.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3675092 c.216C>A p.S72S Chr.8: 1858066-1858066 on Build GRCh38
4454620 c.53A>G p.Y18C Chr.8: 1857975-1857975 on Build GRCh38
6890874 c.43G>T p.E15* Chr.8: 1857965-1857965 on Build GRCh38
5685483 c.127G>A p.D43N Chr.8: 1857977-1857977 on Build GRCh38
4950575 c.94G>A p.D32N Chr.8: 1858016-1858016 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776745780 Chr.8: 1858029-1858029 on Build GRCh38
rs988355139 Chr.8: 1858238-1858261 on Build GRCh38
rs760611109 Chr.8: 1857931-1857931 on Build GRCh38
rs764859351 Chr.8: 1858043-1858043 on Build GRCh38
rs1042239314 Chr.8: 1858003-1858003 on Build GRCh38
rs542129880 Chr.8: 1858199-1858199 on Build GRCh38
rs753288918 Chr.8: 1857911-1857911 on Build GRCh38
rs200417501 Chr.8: 1858025-1858025 on Build GRCh38
rs775208226 Chr.8: 1858215-1858215 on Build GRCh38
rs796297716 Chr.8: 1858196-1858207 on Build GRCh38
rs1016873539 Chr.8: 1858263-1858263 on Build GRCh38
rs780194085 Chr.8: 1858163-1858164 on Build GRCh38
rs528155960 Chr.8: 1858266-1858266 on Build GRCh38
rs118016818 Chr.8: 1858215-1858215 on Build GRCh38
rs564034014 Chr.8: 1858261-1858261 on Build GRCh38
rs778341447 Chr.8: 1857930-1857931 on Build GRCh38
rs753438592 Chr.8: 1858105-1858105 on Build GRCh38
rs777671041 Chr.8: 1858121-1858121 on Build GRCh38
rs765036876 Chr.8: 1857936-1857936 on Build GRCh38
rs754136956 Chr.8: 1858103-1858103 on Build GRCh38
rs559084209 Chr.8: 1858067-1858067 on Build GRCh38
rs550183399 Chr.8: 1858282-1858282 on Build GRCh38
rs1055879778 Chr.8: 1857888-1857888 on Build GRCh38
rs548405211 Chr.8: 1858056-1858056 on Build GRCh38
rs774578667 Chr.8: 1858028-1858028 on Build GRCh38
rs986075562 Chr.8: 1858172-1858172 on Build GRCh38
rs781440599 Chr.8: 1857952-1857952 on Build GRCh38
rs772027392 Chr.8: 1858165-1858166 on Build GRCh38
rs773000055 Chr.8: 1857967-1857967 on Build GRCh38
rs373921275 Chr.8: 1858059-1858059 on Build GRCh38
rs66620071 Chr.8: 1857871-1857874 on Build GRCh38
rs369775414 Chr.8: 1857946-1857946 on Build GRCh38
rs35698984 Chr.8: 1857916-1857919 on Build GRCh38
rs754530974 Chr.8: 1857935-1857935 on Build GRCh38
rs55930580 Chr.8: 1857927-1857928 on Build GRCh38
rs550021765 Chr.8: 1857879-1857880 on Build GRCh38
rs976960709 Chr.8: 1857926-1857926 on Build GRCh38
rs771776264 Chr.8: 1858144-1858144 on Build GRCh38
rs754365949 Chr.8: 1858058-1858058 on Build GRCh38
rs9801821 Chr.8: 1858206-1858206 on Build GRCh38
rs769692992 Chr.8: 1858132-1858132 on Build GRCh38
rs1055136892 Chr.8: 1858223-1858223 on Build GRCh38
rs965205260 Chr.8: 1857925-1857925 on Build GRCh38
rs561245022 Chr.8: 1858252-1858252 on Build GRCh38
rs759503562 Chr.8: 1858032-1858032 on Build GRCh38
rs772951132 Chr.8: 1857875-1857878 on Build GRCh38
rs138043149 Chr.8: 1858085-1858085 on Build GRCh38
rs35312941 Chr.8: 1857877-1857877 on Build GRCh38
rs910236210 Chr.8: 1858236-1858236 on Build GRCh38
rs757733769 Chr.8: 1857948-1857948 on Build GRCh38
rs781376216 Chr.8: 1858155-1858156 on Build GRCh38
rs761312396 Chr.8: 1858048-1858048 on Build GRCh38
rs775518807 Chr.8: 1858068-1858068 on Build GRCh38
rs769952055 Chr.8: 1857978-1857979 on Build GRCh38
rs541605308 Chr.8: 1858273-1858273 on Build GRCh38
rs376516046 Chr.8: 1858203-1858203 on Build GRCh38
rs55846495 Chr.8: 1858187-1858187 on Build GRCh38
rs778967190 Chr.8: 1858060-1858060 on Build GRCh38
rs763092598 Chr.8: 1857918-1857918 on Build GRCh38
rs930004334 Chr.8: 1857853-1857853 on Build GRCh38
rs761646911 Chr.8: 1857924-1857924 on Build GRCh38
rs761533179 Chr.8: 1858140-1858140 on Build GRCh38
rs202242115 Chr.8: 1857932-1857932 on Build GRCh38
rs375269445 Chr.8: 1857912-1857912 on Build GRCh38
rs56179568 Chr.8: 1858251-1858251 on Build GRCh38
rs746164823 Chr.8: 1858156-1858157 on Build GRCh38
rs752425670 Chr.8: 1857983-1857983 on Build GRCh38
rs759162890 Chr.8: 1857981-1857981 on Build GRCh38
rs75038583 Chr.8: 1858123-1858123 on Build GRCh38
rs200387156 Chr.8: 1857955-1857955 on Build GRCh38
rs1023385736 Chr.8: 1857892-1857892 on Build GRCh38
rs769751354 Chr.8: 1857874-1857874 on Build GRCh38
rs35711467 Chr.8: 1857878-1857879 on Build GRCh38
rs11331147 Chr.8: 1858227-1858227 on Build GRCh38
rs147384188 Chr.8: 1858195-1858195 on Build GRCh38
rs56203106 Chr.8: 1858167-1858167 on Build GRCh38
rs776418700 Chr.8: 1857976-1857976 on Build GRCh38
rs749140805 Chr.8: 1858134-1858134 on Build GRCh38
rs917315009 Chr.8: 1857977-1857977 on Build GRCh38
rs910777689 Chr.8: 1858234-1858234 on Build GRCh38
rs749307761 Chr.8: 1857947-1857948 on Build GRCh38
rs201837497 Chr.8: 1857952-1857952 on Build GRCh38
rs4875949 Chr.8: 1858280-1858280 on Build GRCh38
rs893467067 Chr.8: 1857916-1857916 on Build GRCh38
rs34286586 Chr.8: 1857876-1857876 on Build GRCh38
rs752372411 Chr.8: 1857941-1857941 on Build GRCh38
rs763893829 Chr.8: 1857880-1857883 on Build GRCh38
rs71528657 Chr.8: 1858143-1858144 on Build GRCh38
rs1002905393 Chr.8: 1858324-1858324 on Build GRCh38
rs71211518 Chr.8: 1858166-1858167 on Build GRCh38
rs768938335 Chr.8: 1857920-1857920 on Build GRCh38
rs76916066 Chr.8: 1858129-1858129 on Build GRCh38
rs201522949 Chr.8: 1857951-1857951 on Build GRCh38
rs776286696 Chr.8: 1858167-1858168 on Build GRCh38
rs200372897 Chr.8: 1857891-1857891 on Build GRCh38
rs774547207 Chr.8: 1857922-1857922 on Build GRCh38
rs753179497 Chr.8: 1858113-1858113 on Build GRCh38
rs550467895 Chr.8: 1858049-1858049 on Build GRCh38
rs563247783 Chr.8: 1857900-1857900 on Build GRCh38
rs200470716 Chr.8: 1857948-1857952 on Build GRCh38
rs984491821 Chr.8: 1857855-1857855 on Build GRCh38
rs147897258 Chr.8: 1858154-1858154 on Build GRCh38
rs758540863 Chr.8: 1858117-1858117 on Build GRCh38
rs981816665 Chr.8: 1858143-1858143 on Build GRCh38
rs376424164 Chr.8: 1858066-1858066 on Build GRCh38
rs777177605 Chr.8: 1857871-1857878 on Build GRCh38
rs758989601 Chr.8: 1858144-1858145 on Build GRCh38
rs369568036 Chr.8: 1857917-1857917 on Build GRCh38
rs757651864 Chr.8: 1858156-1858167 on Build GRCh38
rs375251991 Chr.8: 1858160-1858160 on Build GRCh38
rs527779906 Chr.8: 1857995-1857995 on Build GRCh38
rs796927633 Chr.8: 1857878-1857879 on Build GRCh38
rs1044683627 Chr.8: 1858311-1858311 on Build GRCh38
rs765533595 Chr.8: 1858318-1858318 on Build GRCh38
rs796568199 Chr.8: 1857895-1857895 on Build GRCh38
rs764632181 Chr.8: 1858146-1858146 on Build GRCh38
rs772994240 Chr.8: 1858142-1858143 on Build GRCh38
rs559487739 Chr.8: 1858222-1858222 on Build GRCh38
rs76522101 Chr.8: 1858122-1858122 on Build GRCh38
rs756273909 Chr.8: 1858160-1858161 on Build GRCh38
rs767780155 Chr.8: 1858000-1858000 on Build GRCh38
rs558780378 Chr.8: 1858153-1858153 on Build GRCh38
rs768235943 Chr.8: 1858135-1858135 on Build GRCh38
rs761339369 Chr.8: 1858107-1858107 on Build GRCh38
rs77453867 Chr.8: 1858131-1858131 on Build GRCh38
rs936167304 Chr.8: 1858219-1858219 on Build GRCh38
rs746051457 Chr.8: 1857975-1857977 on Build GRCh38
rs942376044 Chr.8: 1858237-1858237 on Build GRCh38
rs775425610 Chr.8: 1858129-1858129 on Build GRCh38
rs975708213 Chr.8: 1858255-1858255 on Build GRCh38
rs941961621 Chr.8: 1858295-1858295 on Build GRCh38
rs560936971 Chr.8: 1857883-1857883 on Build GRCh38
rs752006734 Chr.8: 1858146-1858147 on Build GRCh38
rs748385032 Chr.8: 1858016-1858016 on Build GRCh38
rs56122964 Chr.8: 1858239-1858239 on Build GRCh38
rs398067492 Chr.8: 1857879-1857882 on Build GRCh38
rs567607534 Chr.8: 1858015-1858015 on Build GRCh38
rs756425124 Chr.8: 1858010-1858010 on Build GRCh38
rs55847035 Chr.8: 1858168-1858168 on Build GRCh38
rs386720694 Chr.8: 1858187-1858207 on Build GRCh38
rs796313718 Chr.8: 1857875-1857876 on Build GRCh38
rs771937600 Chr.8: 1858129-1858130 on Build GRCh38
rs766593781 Chr.8: 1858034-1858034 on Build GRCh38
rs1022200702 Chr.8: 1858115-1858115 on Build GRCh38
rs574390641 Chr.8: 1857889-1857889 on Build GRCh38
rs765012369 Chr.8: 1858327-1858327 on Build GRCh38
rs746925927 Chr.8: 1858167-1858167 on Build GRCh38
rs770800066 Chr.8: 1858167-1858174 on Build GRCh38
rs534718210 Chr.8: 1858183-1858183 on Build GRCh38
rs758971170 Chr.8: 1857924-1857925 on Build GRCh38
rs28512172 Chr.8: 1858163-1858163 on Build GRCh38
rs547819707 Chr.8: 1858242-1858242 on Build GRCh38
rs747323174 Chr.8: 1857947-1857947 on Build GRCh38
rs760532587 Chr.8: 1857973-1857973 on Build GRCh38
rs1022974188 Chr.8: 1858069-1858069 on Build GRCh38
rs762577053 Chr.8: 1858082-1858082 on Build GRCh38
rs138713415 Chr.8: 1857950-1857950 on Build GRCh38
rs148285858 Chr.8: 1858061-1858061 on Build GRCh38
rs773678489 Chr.8: 1858136-1858136 on Build GRCh38
rs775558900 Chr.8: 1858073-1858073 on Build GRCh38
rs887678038 Chr.8: 1857891-1857907 on Build GRCh38
rs763500882 Chr.8: 1858100-1858100 on Build GRCh38
rs923047870 Chr.8: 1858256-1858256 on Build GRCh38
rs746880011 Chr.8: 1858026-1858026 on Build GRCh38
rs750359766 Chr.8: 1857928-1857928 on Build GRCh38
rs950205850 Chr.8: 1857998-1857998 on Build GRCh38
rs545934959 Chr.8: 1858258-1858258 on Build GRCh38
rs562574210 Chr.8: 1857875-1857876 on Build GRCh38
rs141582464 Chr.8: 1857964-1857964 on Build GRCh38
rs68161747 Chr.8: 1858179-1858180 on Build GRCh38
rs918478544 Chr.8: 1857848-1857848 on Build GRCh38
rs200832839 Chr.8: 1857875-1857875 on Build GRCh38
rs749763498 Chr.8: 1858011-1858011 on Build GRCh38
rs146139864 Chr.8: 1857975-1857975 on Build GRCh38
rs188978848 Chr.8: 1858240-1858240 on Build GRCh38
rs11268265 Chr.8: 1857879-1857879 on Build GRCh38
rs774596559 Chr.8: 1857958-1857958 on Build GRCh38
rs772836051 Chr.8: 1858146-1858146 on Build GRCh38
rs765555139 Chr.8: 1858156-1858156 on Build GRCh38
rs1041987114 Chr.8: 1857894-1857894 on Build GRCh38
rs775076236 Chr.8: 1858047-1858047 on Build GRCh38
rs896014470 Chr.8: 1857890-1857890 on Build GRCh38
rs768024049 Chr.8: 1858084-1858084 on Build GRCh38
rs1039303466 Chr.8: 1858238-1858238 on Build GRCh38
rs547614439 Chr.8: 1858005-1858005 on Build GRCh38
rs111294316 Chr.8: 1858102-1858102 on Build GRCh38
rs879204348 Chr.8: 1857876-1857883 on Build GRCh38
rs62477517 Chr.8: 1858293-1858293 on Build GRCh38
rs537190053 Chr.8: 1858104-1858104 on Build GRCh38
rs367859280 Chr.8: 1858321-1858321 on Build GRCh38
rs748137940 Chr.8: 1858130-1858153 on Build GRCh38

View Full Product Details