Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 1860098-1860605 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF10
Entrez Gene ID
9639
Gene Name
Rho guanine nucleotide exchange factor 10
Gene Aliases
GEF10, SNCV
UniGene
Hs.98594
Gene Chromosome Location
Chr.8: 1823976-1958641 on Build GRCh38
Transcripts
NM_001308152.1 NM_001308153.1 NM_014629.3 XM_005266041.3 XM_011534767.2 XM_011534768.1 XM_011534770.2 XM_017014003.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5979487 c.433G>A p.A145T Chr.8: 1860136-1860136 on Build GRCh38
5979488 c.505G>A p.A169T Chr.8: 1860136-1860136 on Build GRCh38
5615972 c.530G>C p.C177S Chr.8: 1860161-1860161 on Build GRCh38
6603803 c.401C>T p.A134V Chr.8: 1860104-1860104 on Build GRCh38
3924961 c.493C>T p.L165F Chr.8: 1860124-1860124 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373352124 Chr.8: 1860233-1860233 on Build GRCh38
rs796311922 Chr.8: 1860384-1860384 on Build GRCh38
rs781746444 Chr.8: 1860211-1860211 on Build GRCh38
rs747476397 Chr.8: 1860123-1860123 on Build GRCh38
rs911546612 Chr.8: 1860367-1860367 on Build GRCh38
rs80343271 Chr.8: 1860400-1860400 on Build GRCh38
rs547545973 Chr.8: 1860413-1860413 on Build GRCh38
rs189890537 Chr.8: 1860425-1860425 on Build GRCh38
rs766186644 Chr.8: 1860153-1860153 on Build GRCh38
rs749464811 Chr.8: 1860180-1860180 on Build GRCh38
rs766944309 Chr.8: 1860149-1860149 on Build GRCh38
rs530470887 Chr.8: 1860150-1860150 on Build GRCh38
rs186418700 Chr.8: 1860505-1860505 on Build GRCh38
rs201433824 Chr.8: 1860163-1860163 on Build GRCh38
rs918974876 Chr.8: 1860528-1860528 on Build GRCh38
rs1020968862 Chr.8: 1860471-1860471 on Build GRCh38
rs187100964 Chr.8: 1860309-1860309 on Build GRCh38
rs774067194 Chr.8: 1860188-1860188 on Build GRCh38
rs71499049 Chr.8: 1860250-1860250 on Build GRCh38
rs140947400 Chr.8: 1860213-1860213 on Build GRCh38
rs895270113 Chr.8: 1860488-1860488 on Build GRCh38
rs558134381 Chr.8: 1860531-1860531 on Build GRCh38
rs966081249 Chr.8: 1860521-1860521 on Build GRCh38
rs147914724 Chr.8: 1860177-1860177 on Build GRCh38
rs1027692628 Chr.8: 1860148-1860148 on Build GRCh38
rs765943695 Chr.8: 1860202-1860202 on Build GRCh38
rs771735902 Chr.8: 1860226-1860226 on Build GRCh38
rs967249167 Chr.8: 1860569-1860569 on Build GRCh38
rs527621877 Chr.8: 1860406-1860406 on Build GRCh38
rs200556384 Chr.8: 1860181-1860181 on Build GRCh38
rs898394211 Chr.8: 1860445-1860446 on Build GRCh38
rs373946063 Chr.8: 1860435-1860435 on Build GRCh38
rs189893267 Chr.8: 1860555-1860555 on Build GRCh38
rs140941264 Chr.8: 1860118-1860118 on Build GRCh38
rs13277388 Chr.8: 1860349-1860349 on Build GRCh38
rs995940879 Chr.8: 1860461-1860461 on Build GRCh38
rs761513006 Chr.8: 1860192-1860192 on Build GRCh38
rs369004222 Chr.8: 1860480-1860480 on Build GRCh38
rs770934547 Chr.8: 1860124-1860124 on Build GRCh38
rs760275902 Chr.8: 1860199-1860199 on Build GRCh38
rs201352567 Chr.8: 1860212-1860212 on Build GRCh38
rs1028536099 Chr.8: 1860499-1860499 on Build GRCh38
rs533580747 Chr.8: 1860136-1860136 on Build GRCh38
rs769840503 Chr.8: 1860178-1860178 on Build GRCh38
rs560558586 Chr.8: 1860332-1860332 on Build GRCh38
rs74513766 Chr.8: 1860247-1860247 on Build GRCh38
rs4999711 Chr.8: 1860383-1860383 on Build GRCh38
rs146894758 Chr.8: 1860214-1860214 on Build GRCh38
rs80149445 Chr.8: 1860357-1860357 on Build GRCh38
rs758217784 Chr.8: 1860135-1860135 on Build GRCh38
rs368908745 Chr.8: 1860168-1860168 on Build GRCh38
rs773003509 Chr.8: 1860196-1860196 on Build GRCh38
rs754129635 Chr.8: 1860507-1860507 on Build GRCh38
rs4322045 Chr.8: 1860416-1860416 on Build GRCh38
rs13275000 Chr.8: 1860273-1860273 on Build GRCh38
rs939966687 Chr.8: 1860345-1860345 on Build GRCh38
rs1027555215 Chr.8: 1860219-1860219 on Build GRCh38
rs376008740 Chr.8: 1860172-1860172 on Build GRCh38
rs548984736 Chr.8: 1860194-1860194 on Build GRCh38
rs558430028 Chr.8: 1860310-1860310 on Build GRCh38
rs879286429 Chr.8: 1860239-1860273 on Build GRCh38
rs578089918 Chr.8: 1860319-1860319 on Build GRCh38
rs531345786 Chr.8: 1860377-1860377 on Build GRCh38
rs770573903 Chr.8: 1860230-1860230 on Build GRCh38
rs13277414 Chr.8: 1860376-1860376 on Build GRCh38
rs966988431 Chr.8: 1860223-1860223 on Build GRCh38
rs781545843 Chr.8: 1860173-1860173 on Build GRCh38
rs7460669 Chr.8: 1860316-1860316 on Build GRCh38
rs921489828 Chr.8: 1860348-1860348 on Build GRCh38
rs372102236 Chr.8: 1860350-1860350 on Build GRCh38
rs753268156 Chr.8: 1860574-1860574 on Build GRCh38
rs979670675 Chr.8: 1860311-1860311 on Build GRCh38
rs572986879 Chr.8: 1860463-1860463 on Build GRCh38
rs1055122093 Chr.8: 1860467-1860467 on Build GRCh38
rs1035060496 Chr.8: 1860434-1860434 on Build GRCh38
rs761752023 Chr.8: 1860146-1860146 on Build GRCh38
rs747480171 Chr.8: 1860167-1860167 on Build GRCh38
rs753508179 Chr.8: 1860154-1860154 on Build GRCh38
rs911591251 Chr.8: 1860508-1860508 on Build GRCh38
rs1021106185 Chr.8: 1860301-1860301 on Build GRCh38
rs1033038906 Chr.8: 1860580-1860580 on Build GRCh38
rs763838160 Chr.8: 1860145-1860145 on Build GRCh38
rs71499047 Chr.8: 1860241-1860241 on Build GRCh38
rs112712259 Chr.8: 1860473-1860473 on Build GRCh38
rs760477581 Chr.8: 1860229-1860229 on Build GRCh38
rs546332163 Chr.8: 1860141-1860141 on Build GRCh38
rs4431631 Chr.8: 1860443-1860443 on Build GRCh38
rs60543999 Chr.8: 1860368-1860368 on Build GRCh38
rs775674072 Chr.8: 1860142-1860142 on Build GRCh38
rs144697825 Chr.8: 1860129-1860129 on Build GRCh38
rs758861758 Chr.8: 1860162-1860162 on Build GRCh38
rs926434211 Chr.8: 1860561-1860561 on Build GRCh38
rs551355759 Chr.8: 1860385-1860385 on Build GRCh38
rs957289548 Chr.8: 1860248-1860248 on Build GRCh38
rs753563787 Chr.8: 1860204-1860204 on Build GRCh38
rs865982324 Chr.8: 1860489-1860489 on Build GRCh38
rs754614918 Chr.8: 1860206-1860206 on Build GRCh38
rs971826443 Chr.8: 1860524-1860524 on Build GRCh38
rs531284960 Chr.8: 1860455-1860456 on Build GRCh38
rs932402543 Chr.8: 1860285-1860285 on Build GRCh38
rs1037332920 Chr.8: 1860399-1860399 on Build GRCh38
rs553154533 Chr.8: 1860224-1860224 on Build GRCh38
rs575275326 Chr.8: 1860258-1860258 on Build GRCh38
rs905207040 Chr.8: 1860245-1860245 on Build GRCh38
rs778462020 Chr.8: 1860121-1860121 on Build GRCh38
rs780179687 Chr.8: 1860151-1860151 on Build GRCh38
rs763905252 Chr.8: 1860389-1860391 on Build GRCh38
rs890180191 Chr.8: 1860208-1860208 on Build GRCh38
rs1002527993 Chr.8: 1860430-1860430 on Build GRCh38
rs768936663 Chr.8: 1860217-1860217 on Build GRCh38
rs540837350 Chr.8: 1860329-1860329 on Build GRCh38
rs775723006 Chr.8: 1860179-1860179 on Build GRCh38
rs13258095 Chr.8: 1860459-1860459 on Build GRCh38
rs985776805 Chr.8: 1860360-1860360 on Build GRCh38
rs979538337 Chr.8: 1860498-1860498 on Build GRCh38
rs762308160 Chr.8: 1860473-1860473 on Build GRCh38
rs778994451 Chr.8: 1860221-1860221 on Build GRCh38
rs200516301 Chr.8: 1860207-1860207 on Build GRCh38
rs912362726 Chr.8: 1860306-1860306 on Build GRCh38
rs751472043 Chr.8: 1860326-1860328 on Build GRCh38
rs188430170 Chr.8: 1860209-1860209 on Build GRCh38
rs566938735 Chr.8: 1860228-1860228 on Build GRCh38
rs988621804 Chr.8: 1860296-1860296 on Build GRCh38
rs921049103 Chr.8: 1860262-1860262 on Build GRCh38
rs182601908 Chr.8: 1860453-1860453 on Build GRCh38
rs13277372 Chr.8: 1860340-1860340 on Build GRCh38
rs58567079 Chr.8: 1860282-1860282 on Build GRCh38
rs762778852 Chr.8: 1860144-1860144 on Build GRCh38

View Full Product Details