Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 58148094-58148639 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM110B
Entrez Gene ID
90362
Gene Name
family with sequence similarity 110 member B
Gene Aliases
C8orf72
UniGene
Hs.154652
Gene Chromosome Location
Chr.8: 57994444-58149718 on Build GRCh38
Transcripts
NM_147189.2 XM_005251324.2 XM_005251325.3 XM_005251326.2 XM_017013948.1

SNP Detail

SNP ID Chromosome Location
rs150522357 Chr.8: 58148177-58148178 on Build GRCh38
rs769950339 Chr.8: 58148291-58148291 on Build GRCh38
rs775702556 Chr.8: 58148330-58148330 on Build GRCh38
rs190313936 Chr.8: 58148422-58148422 on Build GRCh38
rs770930593 Chr.8: 58148273-58148273 on Build GRCh38
rs752156469 Chr.8: 58148167-58148177 on Build GRCh38
rs796431380 Chr.8: 58148152-58148152 on Build GRCh38
rs886812000 Chr.8: 58148309-58148309 on Build GRCh38
rs141893293 Chr.8: 58148166-58148166 on Build GRCh38
rs549296290 Chr.8: 58148174-58148174 on Build GRCh38
rs1022703375 Chr.8: 58148322-58148322 on Build GRCh38
rs955945802 Chr.8: 58148393-58148393 on Build GRCh38
rs11554813 Chr.8: 58148478-58148478 on Build GRCh38
rs901089985 Chr.8: 58148560-58148560 on Build GRCh38
rs931028593 Chr.8: 58148486-58148486 on Build GRCh38
rs138581719 Chr.8: 58148542-58148542 on Build GRCh38
rs567022372 Chr.8: 58148147-58148147 on Build GRCh38
rs185738097 Chr.8: 58148385-58148385 on Build GRCh38
rs58388044 Chr.8: 58148140-58148140 on Build GRCh38
rs200430917 Chr.8: 58148171-58148172 on Build GRCh38
rs867948434 Chr.8: 58148172-58148172 on Build GRCh38
rs558186897 Chr.8: 58148191-58148191 on Build GRCh38
rs895464704 Chr.8: 58148247-58148247 on Build GRCh38
rs920784461 Chr.8: 58148173-58148173 on Build GRCh38
rs536477603 Chr.8: 58148313-58148313 on Build GRCh38
rs1019459266 Chr.8: 58148293-58148293 on Build GRCh38
rs534980952 Chr.8: 58148178-58148178 on Build GRCh38
rs796737138 Chr.8: 58148173-58148176 on Build GRCh38
rs953993472 Chr.8: 58148269-58148269 on Build GRCh38
rs574160323 Chr.8: 58148248-58148248 on Build GRCh38
rs529721286 Chr.8: 58148462-58148462 on Build GRCh38
rs948366023 Chr.8: 58148565-58148565 on Build GRCh38
rs1047979546 Chr.8: 58148214-58148214 on Build GRCh38
rs774634334 Chr.8: 58148444-58148444 on Build GRCh38
rs925235806 Chr.8: 58148394-58148394 on Build GRCh38
rs1056026001 Chr.8: 58148304-58148304 on Build GRCh38
rs552854177 Chr.8: 58148314-58148314 on Build GRCh38
rs769059636 Chr.8: 58148379-58148379 on Build GRCh38
rs896891199 Chr.8: 58148222-58148222 on Build GRCh38
rs191671706 Chr.8: 58148231-58148231 on Build GRCh38
rs1036770100 Chr.8: 58148181-58148181 on Build GRCh38
rs147111880 Chr.8: 58148449-58148449 on Build GRCh38
rs1010109114 Chr.8: 58148575-58148575 on Build GRCh38
rs1037158844 Chr.8: 58148470-58148470 on Build GRCh38
rs536585046 Chr.8: 58148474-58148474 on Build GRCh38
rs776725483 Chr.8: 58148345-58148345 on Build GRCh38
rs939654882 Chr.8: 58148534-58148535 on Build GRCh38
rs114979600 Chr.8: 58148421-58148421 on Build GRCh38
rs1045369305 Chr.8: 58148157-58148157 on Build GRCh38
rs979436220 Chr.8: 58148382-58148382 on Build GRCh38
rs1056838406 Chr.8: 58148247-58148249 on Build GRCh38
rs559645492 Chr.8: 58148524-58148524 on Build GRCh38
rs995171902 Chr.8: 58148229-58148229 on Build GRCh38
rs77096367 Chr.8: 58148177-58148177 on Build GRCh38
rs566021677 Chr.8: 58148509-58148509 on Build GRCh38
rs949607056 Chr.8: 58148461-58148461 on Build GRCh38
rs965051576 Chr.8: 58148429-58148429 on Build GRCh38
rs377258015 Chr.8: 58148186-58148187 on Build GRCh38
rs56862248 Chr.8: 58148513-58148513 on Build GRCh38
rs183828533 Chr.8: 58148242-58148242 on Build GRCh38
rs551041655 Chr.8: 58148558-58148558 on Build GRCh38
rs191168824 Chr.8: 58148368-58148368 on Build GRCh38
rs770746969 Chr.8: 58148581-58148581 on Build GRCh38
rs776121362 Chr.8: 58148477-58148477 on Build GRCh38
rs117860611 Chr.8: 58148378-58148378 on Build GRCh38
rs1020097464 Chr.8: 58148590-58148590 on Build GRCh38
rs186663957 Chr.8: 58148245-58148245 on Build GRCh38

View Full Product Details