Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 58148528-58149022 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM110B
Entrez Gene ID
90362
Gene Name
family with sequence similarity 110 member B
Gene Aliases
C8orf72
UniGene
Hs.154652
Gene Chromosome Location
Chr.8: 57994444-58149718 on Build GRCh38
Transcripts
NM_147189.2 XM_005251324.2 XM_005251325.3 XM_005251326.2 XM_017013948.1

SNP Detail

SNP ID Chromosome Location
rs764552398 Chr.8: 58148725-58148725 on Build GRCh38
rs1019252437 Chr.8: 58148742-58148742 on Build GRCh38
rs550976371 Chr.8: 58148758-58148758 on Build GRCh38
rs916790272 Chr.8: 58148928-58148928 on Build GRCh38
rs772800301 Chr.8: 58148681-58148681 on Build GRCh38
rs763471597 Chr.8: 58148724-58148724 on Build GRCh38
rs942401581 Chr.8: 58148831-58148831 on Build GRCh38
rs948423511 Chr.8: 58148956-58148956 on Build GRCh38
rs931080925 Chr.8: 58148761-58148761 on Build GRCh38
rs918714932 Chr.8: 58148759-58148759 on Build GRCh38
rs979489836 Chr.8: 58148645-58148645 on Build GRCh38
rs182081274 Chr.8: 58148789-58148789 on Build GRCh38
rs1010109114 Chr.8: 58148575-58148575 on Build GRCh38
rs904241136 Chr.8: 58148640-58148640 on Build GRCh38
rs901089985 Chr.8: 58148560-58148560 on Build GRCh38
rs760276437 Chr.8: 58148911-58148911 on Build GRCh38
rs961141782 Chr.8: 58148910-58148910 on Build GRCh38
rs908064720 Chr.8: 58148841-58148841 on Build GRCh38
rs1000035111 Chr.8: 58148673-58148673 on Build GRCh38
rs935772427 Chr.8: 58148999-58148999 on Build GRCh38
rs766067211 Chr.8: 58148964-58148964 on Build GRCh38
rs966789163 Chr.8: 58148632-58148632 on Build GRCh38
rs538445329 Chr.8: 58148872-58148872 on Build GRCh38
rs1010118929 Chr.8: 58148975-58148975 on Build GRCh38
rs770910224 Chr.8: 58148675-58148675 on Build GRCh38
rs149132940 Chr.8: 58148772-58148772 on Build GRCh38
rs918202597 Chr.8: 58148684-58148684 on Build GRCh38
rs148303573 Chr.8: 58148927-58148927 on Build GRCh38
rs753540897 Chr.8: 58148981-58148981 on Build GRCh38
rs952169203 Chr.8: 58148793-58148793 on Build GRCh38
rs143224302 Chr.8: 58148828-58148828 on Build GRCh38
rs374422511 Chr.8: 58148977-58148977 on Build GRCh38
rs551041655 Chr.8: 58148558-58148558 on Build GRCh38
rs983491620 Chr.8: 58148819-58148819 on Build GRCh38
rs3750193 Chr.8: 58148932-58148932 on Build GRCh38
rs750055380 Chr.8: 58148726-58148726 on Build GRCh38
rs554835940 Chr.8: 58148671-58148671 on Build GRCh38
rs188844571 Chr.8: 58148979-58148979 on Build GRCh38
rs770746969 Chr.8: 58148581-58148581 on Build GRCh38
rs766343333 Chr.8: 58148983-58148984 on Build GRCh38
rs185151068 Chr.8: 58148863-58148863 on Build GRCh38
rs948366023 Chr.8: 58148565-58148565 on Build GRCh38
rs879160472 Chr.8: 58148778-58148778 on Build GRCh38
rs537470217 Chr.8: 58148712-58148712 on Build GRCh38
rs1020097464 Chr.8: 58148590-58148590 on Build GRCh38

View Full Product Details