Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144510900-144511394 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MFSD3
Entrez Gene ID
113655
Gene Name
major facilitator superfamily domain containing 3
Gene Aliases
-
UniGene
Hs.7678
Gene Chromosome Location
Chr.8: 144508081-144511228 on Build GRCh38
Transcripts
NM_138431.2 NR_130120.1 XM_011516806.2 XM_017013005.1 NM_004260.3 XM_011517384.2 XM_017013991.1 XM_017013992.1 XM_017013993.1 XM_017013994.1 XM_017013995.1 XM_017013996.1 XM_017013997.1 XM_017013998.1 XM_017013999.1 XM_017014000.1 XM_017014001.1

Gene Symbol
RECQL4
Entrez Gene ID
9401
Gene Name
RecQ like helicase 4
Gene Aliases
RECQ4
UniGene
Hs.31442
Gene Chromosome Location
Chr.8: 144511284-144517833 on Build GRCh38
Transcripts
NM_138431.2 NR_130120.1 XM_011516806.2 XM_017013005.1 NM_004260.3 XM_011517384.2 XM_017013991.1 XM_017013992.1 XM_017013993.1 XM_017013994.1 XM_017013995.1 XM_017013996.1 XM_017013997.1 XM_017013998.1 XM_017013999.1 XM_017014000.1 XM_017014001.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6198783 c.1200C>T p.P400P Chr.8: 144511125-144511125 on Build GRCh38
3768751 c.1065+9A>G p.? Chr.8: 144510912-144510912 on Build GRCh38
4138646 c.1125T>C p.T375T Chr.8: 144511050-144511050 on Build GRCh38
3779096 c.1126C>T p.L376L Chr.8: 144511051-144511051 on Build GRCh38
5027011 c.1182C>T p.L394L Chr.8: 144511107-144511107 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs549818678 Chr.8: 144510974-144510975 on Build GRCh38
rs747652501 Chr.8: 144511114-144511114 on Build GRCh38
rs766611800 Chr.8: 144511105-144511105 on Build GRCh38
rs200235235 Chr.8: 144510959-144510959 on Build GRCh38
rs1007688755 Chr.8: 144511161-144511161 on Build GRCh38
rs866356147 Chr.8: 144511204-144511204 on Build GRCh38
rs1022008379 Chr.8: 144511282-144511282 on Build GRCh38
rs756363188 Chr.8: 144511101-144511101 on Build GRCh38
rs768988175 Chr.8: 144510966-144510971 on Build GRCh38
rs150812852 Chr.8: 144511024-144511024 on Build GRCh38
rs368438583 Chr.8: 144511201-144511201 on Build GRCh38
rs373704217 Chr.8: 144511205-144511205 on Build GRCh38
rs780966733 Chr.8: 144511182-144511182 on Build GRCh38
rs914780285 Chr.8: 144511102-144511102 on Build GRCh38
rs542293319 Chr.8: 144511126-144511126 on Build GRCh38
rs776248905 Chr.8: 144511217-144511217 on Build GRCh38
rs899269136 Chr.8: 144511362-144511362 on Build GRCh38
rs998813713 Chr.8: 144511197-144511197 on Build GRCh38
rs779511959 Chr.8: 144511113-144511113 on Build GRCh38
rs779071346 Chr.8: 144510931-144510931 on Build GRCh38
rs777431156 Chr.8: 144511119-144511119 on Build GRCh38
rs35440532 Chr.8: 144510941-144510941 on Build GRCh38
rs745637170 Chr.8: 144511184-144511184 on Build GRCh38
rs756154228 Chr.8: 144510927-144510927 on Build GRCh38
rs748193518 Chr.8: 144510933-144510933 on Build GRCh38
rs368010480 Chr.8: 144511179-144511179 on Build GRCh38
rs771867504 Chr.8: 144511014-144511014 on Build GRCh38
rs371239453 Chr.8: 144510942-144510942 on Build GRCh38
rs771051109 Chr.8: 144511210-144511210 on Build GRCh38
rs1016139803 Chr.8: 144511021-144511021 on Build GRCh38
rs570033509 Chr.8: 144511077-144511077 on Build GRCh38
rs575083921 Chr.8: 144510979-144510979 on Build GRCh38
rs764044040 Chr.8: 144511029-144511029 on Build GRCh38
rs1037793012 Chr.8: 144511163-144511163 on Build GRCh38
rs776270377 Chr.8: 144511132-144511132 on Build GRCh38
rs545906114 Chr.8: 144510981-144510981 on Build GRCh38
rs773062208 Chr.8: 144511062-144511062 on Build GRCh38
rs538936253 Chr.8: 144511107-144511107 on Build GRCh38
rs1033339194 Chr.8: 144511224-144511224 on Build GRCh38
rs781434796 Chr.8: 144511038-144511038 on Build GRCh38
rs749690601 Chr.8: 144511048-144511048 on Build GRCh38
rs151312082 Chr.8: 144510927-144510928 on Build GRCh38
rs866822564 Chr.8: 144511286-144511286 on Build GRCh38
rs946356595 Chr.8: 144511326-144511326 on Build GRCh38
rs760634410 Chr.8: 144510964-144510964 on Build GRCh38
rs959143297 Chr.8: 144511060-144511060 on Build GRCh38
rs143762437 Chr.8: 144511156-144511156 on Build GRCh38
rs750586320 Chr.8: 144511099-144511099 on Build GRCh38
rs755414603 Chr.8: 144511112-144511112 on Build GRCh38
rs757450679 Chr.8: 144511032-144511032 on Build GRCh38
rs773882707 Chr.8: 144511026-144511034 on Build GRCh38
rs762891711 Chr.8: 144511137-144511137 on Build GRCh38
rs769574636 Chr.8: 144511187-144511187 on Build GRCh38
rs563306335 Chr.8: 144510963-144510963 on Build GRCh38
rs762437055 Chr.8: 144510945-144510945 on Build GRCh38
rs753192223 Chr.8: 144511174-144511174 on Build GRCh38
rs201260493 Chr.8: 144510936-144510936 on Build GRCh38
rs753469049 Chr.8: 144511004-144511004 on Build GRCh38
rs771816346 Chr.8: 144511061-144511061 on Build GRCh38
rs977635246 Chr.8: 144511287-144511287 on Build GRCh38
rs766975589 Chr.8: 144511196-144511197 on Build GRCh38
rs535744209 Chr.8: 144511241-144511241 on Build GRCh38
rs753664348 Chr.8: 144511020-144511020 on Build GRCh38
rs759345204 Chr.8: 144511082-144511082 on Build GRCh38
rs372265598 Chr.8: 144511215-144511215 on Build GRCh38
rs552879888 Chr.8: 144510939-144510939 on Build GRCh38
rs781475931 Chr.8: 144510984-144510984 on Build GRCh38
rs768601470 Chr.8: 144511192-144511192 on Build GRCh38
rs760758309 Chr.8: 144511141-144511141 on Build GRCh38
rs772495570 Chr.8: 144510980-144510981 on Build GRCh38
rs910516641 Chr.8: 144511333-144511333 on Build GRCh38
rs984206661 Chr.8: 144511238-144511238 on Build GRCh38
rs373242211 Chr.8: 144510938-144510938 on Build GRCh38
rs942889609 Chr.8: 144511345-144511345 on Build GRCh38
rs774420773 Chr.8: 144510966-144510967 on Build GRCh38
rs879151131 Chr.8: 144510980-144510980 on Build GRCh38
rs756831197 Chr.8: 144511180-144511180 on Build GRCh38
rs777372134 Chr.8: 144511178-144511178 on Build GRCh38
rs374406640 Chr.8: 144511173-144511173 on Build GRCh38
rs758036287 Chr.8: 144511115-144511115 on Build GRCh38
rs752821323 Chr.8: 144510971-144510971 on Build GRCh38
rs780230278 Chr.8: 144510938-144510939 on Build GRCh38
rs199720124 Chr.8: 144511057-144511057 on Build GRCh38
rs773309344 Chr.8: 144511140-144511140 on Build GRCh38
rs1016971780 Chr.8: 144511269-144511269 on Build GRCh38
rs556947725 Chr.8: 144511283-144511283 on Build GRCh38
rs557914343 Chr.8: 144511350-144511350 on Build GRCh38
rs147414329 Chr.8: 144510990-144510990 on Build GRCh38
rs759007988 Chr.8: 144511177-144511177 on Build GRCh38
rs141800219 Chr.8: 144511006-144511006 on Build GRCh38
rs770642610 Chr.8: 144511130-144511130 on Build GRCh38
rs937488922 Chr.8: 144510926-144510926 on Build GRCh38
rs763220003 Chr.8: 144511191-144511191 on Build GRCh38
rs988016143 Chr.8: 144511295-144511296 on Build GRCh38
rs745717348 Chr.8: 144511134-144511134 on Build GRCh38
rs754249842 Chr.8: 144511111-144511111 on Build GRCh38
rs201427026 Chr.8: 144511017-144511017 on Build GRCh38
rs139004523 Chr.8: 144511138-144511138 on Build GRCh38
rs35482656 Chr.8: 144510985-144510985 on Build GRCh38
rs149994942 Chr.8: 144511042-144511042 on Build GRCh38
rs769616407 Chr.8: 144511135-144511135 on Build GRCh38
rs17850833 Chr.8: 144511162-144511162 on Build GRCh38
rs375403219 Chr.8: 144510970-144510970 on Build GRCh38
rs1043462052 Chr.8: 144511145-144511145 on Build GRCh38
rs758452424 Chr.8: 144510973-144510973 on Build GRCh38
rs375311882 Chr.8: 144511074-144511074 on Build GRCh38
rs776524641 Chr.8: 144511081-144511081 on Build GRCh38
rs751685090 Chr.8: 144511030-144511030 on Build GRCh38
rs751671277 Chr.8: 144511223-144511223 on Build GRCh38
rs760847213 Chr.8: 144511209-144511209 on Build GRCh38
rs760247336 Chr.8: 144510946-144510946 on Build GRCh38
rs749295420 Chr.8: 144510945-144510945 on Build GRCh38
rs761949393 Chr.8: 144511092-144511092 on Build GRCh38
rs959111004 Chr.8: 144511262-144511262 on Build GRCh38
rs144273718 Chr.8: 144511089-144511089 on Build GRCh38
rs770568697 Chr.8: 144511080-144511080 on Build GRCh38
rs775465367 Chr.8: 144511136-144511136 on Build GRCh38
rs370663265 Chr.8: 144511144-144511144 on Build GRCh38
rs113964706 Chr.8: 144511281-144511281 on Build GRCh38
rs879018093 Chr.8: 144511360-144511360 on Build GRCh38
rs758637224 Chr.8: 144510935-144510935 on Build GRCh38
rs903143208 Chr.8: 144510978-144510978 on Build GRCh38
rs575210383 Chr.8: 144511294-144511294 on Build GRCh38
rs33988375 Chr.8: 144511039-144511039 on Build GRCh38
rs556521203 Chr.8: 144510951-144510951 on Build GRCh38
rs1052252003 Chr.8: 144510986-144510986 on Build GRCh38
rs1037763038 Chr.8: 144511341-144511341 on Build GRCh38
rs769235369 Chr.8: 144511052-144511052 on Build GRCh38
rs751118468 Chr.8: 144511220-144511221 on Build GRCh38
rs767713219 Chr.8: 144511095-144511095 on Build GRCh38
rs138176001 Chr.8: 144510993-144510993 on Build GRCh38
rs748809740 Chr.8: 144510988-144510988 on Build GRCh38
rs987876045 Chr.8: 144511328-144511328 on Build GRCh38
rs773925703 Chr.8: 144510954-144510954 on Build GRCh38
rs773131643 Chr.8: 144511016-144511016 on Build GRCh38
rs943359122 Chr.8: 144511334-144511334 on Build GRCh38
rs377594353 Chr.8: 144511295-144511295 on Build GRCh38
rs754117686 Chr.8: 144511148-144511148 on Build GRCh38
rs993359779 Chr.8: 144511193-144511193 on Build GRCh38
rs545465885 Chr.8: 144511310-144511310 on Build GRCh38
rs746624115 Chr.8: 144511125-144511125 on Build GRCh38
rs771712122 Chr.8: 144511158-144511159 on Build GRCh38
rs112193719 Chr.8: 144511234-144511234 on Build GRCh38
rs761581173 Chr.8: 144510955-144510955 on Build GRCh38
rs750338309 Chr.8: 144510923-144510923 on Build GRCh38
rs552740699 Chr.8: 144511212-144511212 on Build GRCh38
rs201216756 Chr.8: 144511056-144511056 on Build GRCh38
rs757473077 Chr.8: 144510983-144510983 on Build GRCh38
rs368092848 Chr.8: 144511116-144511116 on Build GRCh38
rs202038518 Chr.8: 144511091-144511091 on Build GRCh38

View Full Product Details