Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 93673507-93674000 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC107987098
Entrez Gene ID
107987098
Gene Name
fibroin heavy chain-like
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.9: 93667249-93673578 on Build GRCh38
Transcripts
XM_017015401.1 NM_005392.3 XM_005252051.2 XM_006717143.2

Gene Symbol
PHF2
Entrez Gene ID
5253
Gene Name
PHD finger protein 2
Gene Aliases
CENP-35, GRC5, JHDM1E
UniGene
Hs.211441
Gene Chromosome Location
Chr.9: 93576627-93679587 on Build GRCh38
Transcripts
XM_017015401.1 NM_005392.3 XM_005252051.2 XM_006717143.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6745025 c.2439C>A p.T813T Chr.9: 93673675-93673675 on Build GRCh38
246564 c.2620G>A p.D874N Chr.9: 93673856-93673856 on Build GRCh38
7181789 c.2506G>T p.G836C Chr.9: 93673742-93673742 on Build GRCh38
3909081 c.2402T>C p.M801T Chr.9: 93673638-93673638 on Build GRCh38
1187769 c.2436G>T p.Q812H Chr.9: 93673672-93673672 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762805652 Chr.9: 93673542-93673542 on Build GRCh38
rs960052723 Chr.9: 93673965-93673965 on Build GRCh38
rs373563214 Chr.9: 93673563-93673563 on Build GRCh38
rs768861189 Chr.9: 93673631-93673631 on Build GRCh38
rs769947880 Chr.9: 93673742-93673742 on Build GRCh38
rs368595483 Chr.9: 93673807-93673807 on Build GRCh38
rs200988002 Chr.9: 93673539-93673539 on Build GRCh38
rs201272131 Chr.9: 93673798-93673798 on Build GRCh38
rs748425400 Chr.9: 93673861-93673861 on Build GRCh38
rs777572332 Chr.9: 93673809-93673811 on Build GRCh38
rs760504806 Chr.9: 93673777-93673777 on Build GRCh38
rs776891812 Chr.9: 93673728-93673728 on Build GRCh38
rs746280885 Chr.9: 93673591-93673591 on Build GRCh38
rs371961365 Chr.9: 93673894-93673894 on Build GRCh38
rs751469501 Chr.9: 93673669-93673669 on Build GRCh38
rs763700284 Chr.9: 93673794-93673794 on Build GRCh38
rs369307356 Chr.9: 93673893-93673893 on Build GRCh38
rs753343889 Chr.9: 93673689-93673689 on Build GRCh38
rs372293881 Chr.9: 93673725-93673725 on Build GRCh38
rs375602363 Chr.9: 93673708-93673708 on Build GRCh38
rs755339922 Chr.9: 93673814-93673814 on Build GRCh38
rs969628318 Chr.9: 93673931-93673931 on Build GRCh38
rs566977630 Chr.9: 93673957-93673957 on Build GRCh38
rs113951079 Chr.9: 93673979-93673979 on Build GRCh38
rs548206015 Chr.9: 93673552-93673552 on Build GRCh38
rs868155741 Chr.9: 93673822-93673822 on Build GRCh38
rs41276200 Chr.9: 93673755-93673755 on Build GRCh38
rs767034947 Chr.9: 93673905-93673905 on Build GRCh38
rs758136005 Chr.9: 93673765-93673805 on Build GRCh38
rs569488851 Chr.9: 93673567-93673567 on Build GRCh38
rs779458980 Chr.9: 93673590-93673590 on Build GRCh38
rs749890391 Chr.9: 93673802-93673802 on Build GRCh38
rs148170207 Chr.9: 93673678-93673678 on Build GRCh38
rs763975881 Chr.9: 93673887-93673887 on Build GRCh38
rs760036413 Chr.9: 93673899-93673899 on Build GRCh38
rs991932991 Chr.9: 93673978-93673978 on Build GRCh38
rs893582216 Chr.9: 93673885-93673885 on Build GRCh38
rs779599558 Chr.9: 93673696-93673696 on Build GRCh38
rs752140557 Chr.9: 93673907-93673907 on Build GRCh38
rs534466352 Chr.9: 93673727-93673727 on Build GRCh38
rs916006866 Chr.9: 93673788-93673788 on Build GRCh38
rs145764244 Chr.9: 93673658-93673658 on Build GRCh38
rs781374042 Chr.9: 93673831-93673831 on Build GRCh38
rs777477117 Chr.9: 93673683-93673683 on Build GRCh38
rs773284481 Chr.9: 93673875-93673875 on Build GRCh38
rs766338671 Chr.9: 93673549-93673549 on Build GRCh38
rs746405383 Chr.9: 93673702-93673702 on Build GRCh38
rs901951676 Chr.9: 93673700-93673700 on Build GRCh38
rs193921057 Chr.9: 93673799-93673799 on Build GRCh38
rs1016888973 Chr.9: 93673862-93673862 on Build GRCh38
rs748762955 Chr.9: 93673565-93673565 on Build GRCh38
rs769849954 Chr.9: 93673874-93673874 on Build GRCh38
rs960329731 Chr.9: 93673743-93673743 on Build GRCh38
rs752378246 Chr.9: 93673555-93673555 on Build GRCh38
rs200992138 Chr.9: 93673550-93673550 on Build GRCh38
rs369709048 Chr.9: 93673880-93673880 on Build GRCh38
rs748218124 Chr.9: 93673738-93673738 on Build GRCh38
rs141975474 Chr.9: 93673748-93673748 on Build GRCh38
rs759429518 Chr.9: 93673677-93673677 on Build GRCh38
rs780863219 Chr.9: 93673553-93673553 on Build GRCh38
rs140677384 Chr.9: 93673594-93673594 on Build GRCh38
rs890105167 Chr.9: 93673819-93673819 on Build GRCh38
rs772653023 Chr.9: 93673707-93673707 on Build GRCh38
rs929190311 Chr.9: 93673808-93673808 on Build GRCh38
rs199537702 Chr.9: 93673657-93673657 on Build GRCh38
rs752641809 Chr.9: 93673704-93673707 on Build GRCh38
rs377434686 Chr.9: 93673609-93673609 on Build GRCh38
rs548256223 Chr.9: 93673804-93673804 on Build GRCh38
rs756766990 Chr.9: 93673690-93673690 on Build GRCh38
rs372056066 Chr.9: 93673813-93673813 on Build GRCh38
rs190945254 Chr.9: 93673903-93673903 on Build GRCh38
rs903855514 Chr.9: 93673662-93673662 on Build GRCh38
rs760424385 Chr.9: 93673886-93673886 on Build GRCh38
rs767375895 Chr.9: 93673766-93673766 on Build GRCh38
rs749364456 Chr.9: 93673876-93673876 on Build GRCh38
rs775328716 Chr.9: 93673767-93673767 on Build GRCh38
rs78337831 Chr.9: 93673879-93673879 on Build GRCh38
rs760994623 Chr.9: 93673610-93673610 on Build GRCh38
rs764859067 Chr.9: 93673801-93673801 on Build GRCh38
rs776644776 Chr.9: 93673636-93673636 on Build GRCh38
rs777160754 Chr.9: 93673564-93673564 on Build GRCh38

View Full Product Details