Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 98056543-98057083 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NANS
Entrez Gene ID
54187
Gene Name
N-acetylneuraminate synthase
Gene Aliases
HEL-S-100, SAS, SEMDG
UniGene
Hs.522310
Gene Chromosome Location
Chr.9: 98056677-98083083 on Build GRCh38
Transcripts
NM_018946.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7378877 c.74T>A p.I25N Chr.9: 98056882-98056882 on Build GRCh38
6315158 c.35G>T p.W12L Chr.9: 98056843-98056843 on Build GRCh38
1187782 c.103G>A p.V35I Chr.9: 98056911-98056911 on Build GRCh38
3763481 c.102C>T p.D34D Chr.9: 98056910-98056910 on Build GRCh38
5600008 c.75C>T p.I25I Chr.9: 98056883-98056883 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764226833 Chr.9: 98056957-98056957 on Build GRCh38
rs976184179 Chr.9: 98056621-98056622 on Build GRCh38
rs896049240 Chr.9: 98056592-98056592 on Build GRCh38
rs996792985 Chr.9: 98056793-98056793 on Build GRCh38
rs925337787 Chr.9: 98056802-98056802 on Build GRCh38
rs766439136 Chr.9: 98056851-98056851 on Build GRCh38
rs549193701 Chr.9: 98056995-98056995 on Build GRCh38
rs371399380 Chr.9: 98056778-98056778 on Build GRCh38
rs952035625 Chr.9: 98056563-98056563 on Build GRCh38
rs569435914 Chr.9: 98057034-98057034 on Build GRCh38
rs536951859 Chr.9: 98056746-98056747 on Build GRCh38
rs763988137 Chr.9: 98056835-98056835 on Build GRCh38
rs746720794 Chr.9: 98056920-98056920 on Build GRCh38
rs938313095 Chr.9: 98056624-98056624 on Build GRCh38
rs1000982954 Chr.9: 98057061-98057061 on Build GRCh38
rs754197886 Chr.9: 98056958-98056958 on Build GRCh38
rs780848926 Chr.9: 98056991-98056991 on Build GRCh38
rs768283157 Chr.9: 98056988-98056988 on Build GRCh38
rs752851244 Chr.9: 98056939-98056939 on Build GRCh38
rs891861018 Chr.9: 98056767-98056767 on Build GRCh38
rs761597906 Chr.9: 98056929-98056929 on Build GRCh38
rs542085176 Chr.9: 98056779-98056779 on Build GRCh38
rs139706489 Chr.9: 98056861-98056861 on Build GRCh38
rs979418327 Chr.9: 98056622-98056623 on Build GRCh38
rs752366951 Chr.9: 98056773-98056773 on Build GRCh38
rs760068312 Chr.9: 98056954-98056955 on Build GRCh38
rs878970319 Chr.9: 98056716-98056716 on Build GRCh38
rs760344751 Chr.9: 98056833-98056833 on Build GRCh38
rs150529610 Chr.9: 98057057-98057057 on Build GRCh38
rs765874154 Chr.9: 98056982-98056983 on Build GRCh38
rs367632845 Chr.9: 98056759-98056759 on Build GRCh38
rs773169954 Chr.9: 98056930-98056930 on Build GRCh38
rs78604260 Chr.9: 98057060-98057060 on Build GRCh38
rs756535085 Chr.9: 98056798-98056798 on Build GRCh38
rs866659432 Chr.9: 98056885-98056885 on Build GRCh38
rs368286579 Chr.9: 98056860-98056860 on Build GRCh38
rs536620558 Chr.9: 98056697-98056697 on Build GRCh38
rs11545355 Chr.9: 98056847-98056847 on Build GRCh38
rs910356007 Chr.9: 98056696-98056696 on Build GRCh38
rs965848771 Chr.9: 98056621-98056621 on Build GRCh38
rs753296138 Chr.9: 98056993-98056993 on Build GRCh38
rs544262885 Chr.9: 98056828-98056828 on Build GRCh38
rs920340992 Chr.9: 98056709-98056709 on Build GRCh38
rs751005513 Chr.9: 98056976-98056976 on Build GRCh38
rs1054132494 Chr.9: 98056621-98056621 on Build GRCh38
rs766492808 Chr.9: 98056784-98056784 on Build GRCh38
rs570418881 Chr.9: 98056735-98056735 on Build GRCh38
rs779575112 Chr.9: 98056901-98056901 on Build GRCh38
rs760531732 Chr.9: 98056775-98056775 on Build GRCh38
rs1044556329 Chr.9: 98056752-98056752 on Build GRCh38
rs773288226 Chr.9: 98056779-98056779 on Build GRCh38
rs761026389 Chr.9: 98056955-98056955 on Build GRCh38
rs768320257 Chr.9: 98056921-98056921 on Build GRCh38
rs3199064 Chr.9: 98056788-98056788 on Build GRCh38
rs1054502962 Chr.9: 98056765-98056765 on Build GRCh38
rs998567877 Chr.9: 98056615-98056615 on Build GRCh38
rs749860074 Chr.9: 98056808-98056808 on Build GRCh38
rs775230449 Chr.9: 98056832-98056832 on Build GRCh38
rs956102727 Chr.9: 98056638-98056638 on Build GRCh38
rs529443863 Chr.9: 98056972-98056972 on Build GRCh38
rs1015670159 Chr.9: 98057036-98057036 on Build GRCh38
rs770606631 Chr.9: 98056762-98056762 on Build GRCh38
rs748416345 Chr.9: 98056797-98056797 on Build GRCh38
rs950332298 Chr.9: 98056982-98056982 on Build GRCh38
rs749808742 Chr.9: 98056862-98056862 on Build GRCh38
rs745376822 Chr.9: 98056812-98056812 on Build GRCh38
rs989209949 Chr.9: 98056663-98056663 on Build GRCh38
rs1032976326 Chr.9: 98056629-98056629 on Build GRCh38
rs141699820 Chr.9: 98056816-98056816 on Build GRCh38
rs774127195 Chr.9: 98056766-98056766 on Build GRCh38
rs1020822701 Chr.9: 98057044-98057044 on Build GRCh38
rs1008787752 Chr.9: 98056877-98056877 on Build GRCh38
rs955261188 Chr.9: 98056992-98056992 on Build GRCh38
rs966686977 Chr.9: 98056592-98056612 on Build GRCh38
rs369033671 Chr.9: 98056809-98056809 on Build GRCh38
rs920387701 Chr.9: 98056913-98056913 on Build GRCh38
rs3739670 Chr.9: 98056910-98056910 on Build GRCh38
rs767597953 Chr.9: 98056854-98056854 on Build GRCh38
rs762458937 Chr.9: 98057045-98057045 on Build GRCh38
rs556952606 Chr.9: 98056728-98056728 on Build GRCh38
rs540341211 Chr.9: 98056912-98056912 on Build GRCh38
rs3832624 Chr.9: 98056627-98056627 on Build GRCh38
rs973113500 Chr.9: 98056702-98056702 on Build GRCh38
rs950326119 Chr.9: 98056739-98056739 on Build GRCh38
rs943110091 Chr.9: 98056699-98056699 on Build GRCh38
rs905976179 Chr.9: 98056623-98056623 on Build GRCh38
rs929023802 Chr.9: 98056724-98056724 on Build GRCh38
rs1017476857 Chr.9: 98056611-98056623 on Build GRCh38
rs757772678 Chr.9: 98056876-98056876 on Build GRCh38
rs761020129 Chr.9: 98056787-98056788 on Build GRCh38
rs1008468296 Chr.9: 98056997-98056997 on Build GRCh38
rs937993978 Chr.9: 98056819-98056819 on Build GRCh38
rs1014454904 Chr.9: 98056600-98056600 on Build GRCh38
rs759321267 Chr.9: 98056770-98056770 on Build GRCh38
rs1010259894 Chr.9: 98056769-98056769 on Build GRCh38
rs776860726 Chr.9: 98056814-98056819 on Build GRCh38
rs532047788 Chr.9: 98057038-98057038 on Build GRCh38
rs1020897950 Chr.9: 98056666-98056666 on Build GRCh38
rs560256436 Chr.9: 98056937-98056937 on Build GRCh38
rs553155111 Chr.9: 98056748-98056748 on Build GRCh38
rs779670037 Chr.9: 98056979-98056979 on Build GRCh38
rs755076717 Chr.9: 98056853-98056853 on Build GRCh38
rs954767843 Chr.9: 98056672-98056672 on Build GRCh38
rs766298846 Chr.9: 98056788-98056789 on Build GRCh38
rs776473084 Chr.9: 98056928-98056928 on Build GRCh38
rs1048017043 Chr.9: 98056587-98056587 on Build GRCh38
rs1017453701 Chr.9: 98056906-98056906 on Build GRCh38
rs929551309 Chr.9: 98056586-98056586 on Build GRCh38
rs1038977965 Chr.9: 98057011-98057018 on Build GRCh38
rs778113732 Chr.9: 98056803-98056803 on Build GRCh38
rs779521942 Chr.9: 98056973-98056973 on Build GRCh38
rs757824058 Chr.9: 98056977-98056977 on Build GRCh38
rs569582351 Chr.9: 98056684-98056684 on Build GRCh38
rs539539952 Chr.9: 98056740-98056740 on Build GRCh38
rs755256468 Chr.9: 98056787-98056787 on Build GRCh38
rs1000203275 Chr.9: 98056627-98056627 on Build GRCh38
rs751762509 Chr.9: 98056785-98056785 on Build GRCh38
rs776406527 Chr.9: 98056845-98056845 on Build GRCh38
rs1027356301 Chr.9: 98056747-98056749 on Build GRCh38
rs945378493 Chr.9: 98056622-98056622 on Build GRCh38
rs753844033 Chr.9: 98056776-98056776 on Build GRCh38
rs987337285 Chr.9: 98056687-98056687 on Build GRCh38
rs901722630 Chr.9: 98057029-98057029 on Build GRCh38
rs374668704 Chr.9: 98056622-98056622 on Build GRCh38
rs964581926 Chr.9: 98056907-98056907 on Build GRCh38
rs1023070959 Chr.9: 98056605-98056605 on Build GRCh38

View Full Product Details