Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 105574832-105575334 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKTN
Entrez Gene ID
2218
Gene Name
fukutin
Gene Aliases
CMD1X, FCMD, LGMD2M, MDDGA4, MDDGB4, MDDGC4
UniGene
Hs.55777
Gene Chromosome Location
Chr.9: 105558117-105655950 on Build GRCh38
Transcripts
NM_001079802.1 NM_001198963.1 NM_006731.2 XM_006717014.2 XM_011518368.2 XM_011518369.2 XM_011518370.2 XM_011518371.2 XM_011518373.2 XM_011518374.2 XM_011518375.2 XM_011518376.2 XM_011518378.2 XM_011518379.2 XM_011518381.2 XM_011518387.2 XM_011518389.2 XM_011518391.2 XM_017014462.1 XM_017014463.1 XM_017014464.1 XM_017014465.1 XM_017014466.1 XM_017014467.1 XM_017014468.1 XM_017014469.1 XM_017014470.1 XM_017014471.1 XM_017014472.1 XM_017014474.1 XM_017014475.1 XM_017014476.1 XM_017014477.1 XM_017014478.1 XM_017014479.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5901594 c.23T>A p.V8E Chr.9: 105575055-105575055 on Build GRCh38
3902752 c.12C>G p.I4M Chr.9: 105575044-105575044 on Build GRCh38
4850775 c.94T>C p.L32L Chr.9: 105575126-105575126 on Build GRCh38
1643689 c.38_70del33 p.L13_Q24>* Chr.9: 105575070-105575102 on Build GRCh38
486870 c.35T>C p.L12P Chr.9: 105575067-105575067 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs549316366 Chr.9: 105575231-105575231 on Build GRCh38
rs755870328 Chr.9: 105575055-105575055 on Build GRCh38
rs757900916 Chr.9: 105575042-105575042 on Build GRCh38
rs1023232100 Chr.9: 105574892-105574892 on Build GRCh38
rs867971470 Chr.9: 105575111-105575111 on Build GRCh38
rs923484240 Chr.9: 105574972-105574972 on Build GRCh38
rs780450716 Chr.9: 105575183-105575183 on Build GRCh38
rs777766061 Chr.9: 105575176-105575176 on Build GRCh38
rs398123554 Chr.9: 105575173-105575176 on Build GRCh38
rs754468850 Chr.9: 105575143-105575143 on Build GRCh38
rs990303663 Chr.9: 105574967-105574967 on Build GRCh38
rs1054946778 Chr.9: 105575121-105575121 on Build GRCh38
rs755931347 Chr.9: 105575169-105575169 on Build GRCh38
rs974922723 Chr.9: 105575259-105575259 on Build GRCh38
rs921462265 Chr.9: 105574970-105574970 on Build GRCh38
rs954757160 Chr.9: 105575256-105575256 on Build GRCh38
rs751029878 Chr.9: 105575137-105575137 on Build GRCh38
rs555754136 Chr.9: 105574940-105574940 on Build GRCh38
rs138740640 Chr.9: 105575134-105575134 on Build GRCh38
rs939985733 Chr.9: 105575066-105575066 on Build GRCh38
rs942192637 Chr.9: 105575297-105575297 on Build GRCh38
rs751473818 Chr.9: 105575046-105575046 on Build GRCh38
rs745881231 Chr.9: 105575001-105575001 on Build GRCh38
rs747056648 Chr.9: 105575187-105575187 on Build GRCh38
rs969473930 Chr.9: 105574914-105574914 on Build GRCh38
rs1032025444 Chr.9: 105575230-105575230 on Build GRCh38
rs533603648 Chr.9: 105575124-105575124 on Build GRCh38
rs762110230 Chr.9: 105575026-105575026 on Build GRCh38
rs368981218 Chr.9: 105575054-105575054 on Build GRCh38
rs902435776 Chr.9: 105575180-105575180 on Build GRCh38
rs193206252 Chr.9: 105575027-105575027 on Build GRCh38
rs567763205 Chr.9: 105575257-105575257 on Build GRCh38
rs752577408 Chr.9: 105575154-105575154 on Build GRCh38
rs185348598 Chr.9: 105575028-105575028 on Build GRCh38
rs776133464 Chr.9: 105575012-105575012 on Build GRCh38
rs750082228 Chr.9: 105575035-105575035 on Build GRCh38
rs570332610 Chr.9: 105574887-105574887 on Build GRCh38
rs564632018 Chr.9: 105575053-105575053 on Build GRCh38
rs1028489081 Chr.9: 105575302-105575302 on Build GRCh38
rs772280499 Chr.9: 105575004-105575004 on Build GRCh38
rs115730708 Chr.9: 105574939-105574939 on Build GRCh38
rs956683866 Chr.9: 105574958-105574958 on Build GRCh38
rs145387221 Chr.9: 105575057-105575057 on Build GRCh38
rs201648601 Chr.9: 105575075-105575075 on Build GRCh38
rs112124536 Chr.9: 105575266-105575266 on Build GRCh38
rs369797361 Chr.9: 105575110-105575110 on Build GRCh38
rs966402869 Chr.9: 105574889-105574889 on Build GRCh38
rs1037406947 Chr.9: 105575039-105575039 on Build GRCh38
rs77013649 Chr.9: 105574988-105574988 on Build GRCh38
rs553546181 Chr.9: 105575000-105575000 on Build GRCh38
rs188804693 Chr.9: 105574956-105574956 on Build GRCh38
rs762170532 Chr.9: 105575098-105575098 on Build GRCh38
rs181305205 Chr.9: 105575177-105575177 on Build GRCh38
rs759773682 Chr.9: 105575144-105575145 on Build GRCh38
rs149033995 Chr.9: 105575073-105575073 on Build GRCh38
rs769161688 Chr.9: 105575094-105575094 on Build GRCh38
rs957800165 Chr.9: 105575233-105575233 on Build GRCh38
rs570495326 Chr.9: 105575152-105575152 on Build GRCh38
rs188900946 Chr.9: 105575107-105575107 on Build GRCh38
rs374203636 Chr.9: 105575115-105575115 on Build GRCh38
rs766642997 Chr.9: 105575095-105575095 on Build GRCh38
rs542084356 Chr.9: 105575018-105575018 on Build GRCh38
rs773335262 Chr.9: 105575244-105575244 on Build GRCh38
rs758657034 Chr.9: 105575181-105575181 on Build GRCh38
rs771143609 Chr.9: 105574987-105574987 on Build GRCh38
rs202047149 Chr.9: 105575062-105575062 on Build GRCh38
rs78794935 Chr.9: 105575074-105575074 on Build GRCh38
rs535351242 Chr.9: 105575308-105575308 on Build GRCh38
rs910840865 Chr.9: 105575267-105575267 on Build GRCh38
rs933581388 Chr.9: 105574981-105574981 on Build GRCh38
rs764337105 Chr.9: 105575015-105575015 on Build GRCh38
rs761317265 Chr.9: 105575013-105575013 on Build GRCh38
rs765929865 Chr.9: 105575045-105575045 on Build GRCh38
rs1030766933 Chr.9: 105575205-105575205 on Build GRCh38
rs988179549 Chr.9: 105574959-105574959 on Build GRCh38
rs768104853 Chr.9: 105575183-105575183 on Build GRCh38

View Full Product Details