Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 64412541-64413068 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD20A4
Entrez Gene ID
728747
Gene Name
ankyrin repeat domain 20 family member A4
Gene Aliases
-
UniGene
Hs.645502
Gene Chromosome Location
Chr.9: 64369563-64412691 on Build GRCh38
Transcripts
NM_001098805.1

SNP Detail

SNP ID Chromosome Location
rs138331612 Chr.9: 64412891-64412891 on Build GRCh38
rs941909976 Chr.9: 64412698-64412698 on Build GRCh38
rs71221420 Chr.9: 64412594-64412594 on Build GRCh38
rs557081060 Chr.9: 64412809-64412809 on Build GRCh38
rs878961209 Chr.9: 64413043-64413044 on Build GRCh38
rs970478860 Chr.9: 64412646-64412646 on Build GRCh38
rs973288797 Chr.9: 64412730-64412730 on Build GRCh38
rs78626733 Chr.9: 64412609-64412609 on Build GRCh38
rs62555350 Chr.9: 64412566-64412566 on Build GRCh38
rs1047036640 Chr.9: 64412826-64412826 on Build GRCh38
rs375899174 Chr.9: 64412713-64412713 on Build GRCh38
rs919206301 Chr.9: 64412766-64412766 on Build GRCh38
rs1025124674 Chr.9: 64412579-64412579 on Build GRCh38
rs575426608 Chr.9: 64413011-64413011 on Build GRCh38
rs375996767 Chr.9: 64413044-64413045 on Build GRCh38
rs929962522 Chr.9: 64412814-64412814 on Build GRCh38
rs371086738 Chr.9: 64412665-64412665 on Build GRCh38
rs77695146 Chr.9: 64412981-64412981 on Build GRCh38
rs142721300 Chr.9: 64412902-64412902 on Build GRCh38
rs112372164 Chr.9: 64412963-64412963 on Build GRCh38
rs71221418 Chr.9: 64412811-64412811 on Build GRCh38
rs980940987 Chr.9: 64412694-64412694 on Build GRCh38
rs370856188 Chr.9: 64412569-64412569 on Build GRCh38
rs891135269 Chr.9: 64412856-64412856 on Build GRCh38

View Full Product Details