Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 135694963-135695462 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SOHLH1
Entrez Gene ID
402381
Gene Name
spermatogenesis and oogenesis specific basic helix-loop-helix 1
Gene Aliases
C9orf157, NOHLH, SPATA27, TEB2, bA100C15.3, bHLHe80
UniGene
Hs.120464
Gene Chromosome Location
Chr.9: 135691862-135701316 on Build GRCh38
Transcripts
NM_001012415.2 NM_001101677.1 XM_005263403.3 XM_006717109.3 XM_011518698.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5693877 c.771C>A p.S257R Chr.9: 135695154-135695154 on Build GRCh38
5693878 c.771C>A p.S257R Chr.9: 135695154-135695154 on Build GRCh38
341549 c.687C>A p.P229P Chr.9: 135695238-135695238 on Build GRCh38
4138886 c.718C>T p.P240S Chr.9: 135695207-135695207 on Build GRCh38
5509451 c.741C>T p.F247F Chr.9: 135695184-135695184 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772260088 Chr.9: 135695178-135695178 on Build GRCh38
rs755384172 Chr.9: 135695216-135695216 on Build GRCh38
rs764906600 Chr.9: 135695117-135695117 on Build GRCh38
rs371104276 Chr.9: 135695155-135695155 on Build GRCh38
rs756533182 Chr.9: 135695154-135695154 on Build GRCh38
rs974113886 Chr.9: 135695033-135695033 on Build GRCh38
rs770040924 Chr.9: 135695095-135695095 on Build GRCh38
rs916982340 Chr.9: 135695143-135695143 on Build GRCh38
rs756523071 Chr.9: 135695210-135695210 on Build GRCh38
rs879549827 Chr.9: 135695120-135695121 on Build GRCh38
rs190387809 Chr.9: 135695005-135695005 on Build GRCh38
rs1000405099 Chr.9: 135695270-135695270 on Build GRCh38
rs767068224 Chr.9: 135695267-135695267 on Build GRCh38
rs756014216 Chr.9: 135695017-135695017 on Build GRCh38
rs760696005 Chr.9: 135695109-135695109 on Build GRCh38
rs760229228 Chr.9: 135695268-135695268 on Build GRCh38
rs751199646 Chr.9: 135695204-135695204 on Build GRCh38
rs768880996 Chr.9: 135695101-135695101 on Build GRCh38
rs980219473 Chr.9: 135695321-135695321 on Build GRCh38
rs864309645 Chr.9: 135695220-135695220 on Build GRCh38
rs151131296 Chr.9: 135695213-135695213 on Build GRCh38
rs753622955 Chr.9: 135695072-135695072 on Build GRCh38
rs771260967 Chr.9: 135695002-135695002 on Build GRCh38
rs771332897 Chr.9: 135694986-135694986 on Build GRCh38
rs747479486 Chr.9: 135695036-135695036 on Build GRCh38
rs550754812 Chr.9: 135695182-135695182 on Build GRCh38
rs754225807 Chr.9: 135695214-135695214 on Build GRCh38
rs753569465 Chr.9: 135695022-135695022 on Build GRCh38
rs747124243 Chr.9: 135695307-135695307 on Build GRCh38
rs555432506 Chr.9: 135695425-135695425 on Build GRCh38
rs781706567 Chr.9: 135695152-135695152 on Build GRCh38
rs1025824692 Chr.9: 135695234-135695253 on Build GRCh38
rs759790274 Chr.9: 135695006-135695006 on Build GRCh38
rs778297193 Chr.9: 135695034-135695034 on Build GRCh38
rs764239665 Chr.9: 135695061-135695061 on Build GRCh38
rs776873948 Chr.9: 135695003-135695003 on Build GRCh38
rs779771355 Chr.9: 135695380-135695380 on Build GRCh38
rs749431231 Chr.9: 135695097-135695097 on Build GRCh38
rs750962597 Chr.9: 135695209-135695209 on Build GRCh38
rs751002538 Chr.9: 135695139-135695139 on Build GRCh38
rs775492494 Chr.9: 135695246-135695246 on Build GRCh38
rs756754075 Chr.9: 135695149-135695149 on Build GRCh38
rs748353406 Chr.9: 135695230-135695230 on Build GRCh38
rs369119349 Chr.9: 135695436-135695436 on Build GRCh38
rs773708000 Chr.9: 135695000-135695000 on Build GRCh38
rs766293252 Chr.9: 135695069-135695069 on Build GRCh38
rs2483364 Chr.9: 135695374-135695374 on Build GRCh38
rs752985711 Chr.9: 135695289-135695289 on Build GRCh38
rs150307035 Chr.9: 135695261-135695261 on Build GRCh38
rs67947712 Chr.9: 135695121-135695121 on Build GRCh38
rs145861345 Chr.9: 135695057-135695057 on Build GRCh38
rs761082850 Chr.9: 135695001-135695001 on Build GRCh38
rs758489128 Chr.9: 135695296-135695296 on Build GRCh38
rs143085310 Chr.9: 135695166-135695166 on Build GRCh38
rs757276257 Chr.9: 135695346-135695346 on Build GRCh38
rs761360902 Chr.9: 135695206-135695206 on Build GRCh38
rs1036970694 Chr.9: 135695201-135695201 on Build GRCh38
rs1040276814 Chr.9: 135695386-135695386 on Build GRCh38
rs931681564 Chr.9: 135695340-135695340 on Build GRCh38
rs777909798 Chr.9: 135695301-135695301 on Build GRCh38
rs373023569 Chr.9: 135695051-135695051 on Build GRCh38
rs757961881 Chr.9: 135695081-135695081 on Build GRCh38
rs140624736 Chr.9: 135695163-135695163 on Build GRCh38
rs956301078 Chr.9: 135695285-135695285 on Build GRCh38
rs182554663 Chr.9: 135695026-135695026 on Build GRCh38
rs761407342 Chr.9: 135695263-135695263 on Build GRCh38
rs750269318 Chr.9: 135695363-135695363 on Build GRCh38
rs370813781 Chr.9: 135695037-135695037 on Build GRCh38
rs765653767 Chr.9: 135695286-135695286 on Build GRCh38
rs971058372 Chr.9: 135695319-135695319 on Build GRCh38
rs3119932 Chr.9: 135695120-135695120 on Build GRCh38
rs386739489 Chr.9: 135695120-135695121 on Build GRCh38
rs138464466 Chr.9: 135695322-135695322 on Build GRCh38
rs150490094 Chr.9: 135695257-135695257 on Build GRCh38
rs750364732 Chr.9: 135695014-135695014 on Build GRCh38
rs776030089 Chr.9: 135695030-135695034 on Build GRCh38
rs763801290 Chr.9: 135695194-135695194 on Build GRCh38
rs539454834 Chr.9: 135695404-135695404 on Build GRCh38
rs374532775 Chr.9: 135695162-135695162 on Build GRCh38
rs537177480 Chr.9: 135695424-135695424 on Build GRCh38
rs201483476 Chr.9: 135695052-135695052 on Build GRCh38
rs760642500 Chr.9: 135695068-135695068 on Build GRCh38
rs55674357 Chr.9: 135695094-135695094 on Build GRCh38
rs774516240 Chr.9: 135695105-135695105 on Build GRCh38
rs891540968 Chr.9: 135694995-135694995 on Build GRCh38
rs569647904 Chr.9: 135695422-135695422 on Build GRCh38
rs752266509 Chr.9: 135695079-135695079 on Build GRCh38
rs928815377 Chr.9: 135695247-135695247 on Build GRCh38
rs749540912 Chr.9: 135695157-135695157 on Build GRCh38
rs529080738 Chr.9: 135695364-135695364 on Build GRCh38
rs200800281 Chr.9: 135695113-135695113 on Build GRCh38
rs556508667 Chr.9: 135695090-135695090 on Build GRCh38
rs146613743 Chr.9: 135695153-135695153 on Build GRCh38
rs754730441 Chr.9: 135695073-135695073 on Build GRCh38
rs745829439 Chr.9: 135695232-135695232 on Build GRCh38
rs776928977 Chr.9: 135695042-135695042 on Build GRCh38
rs3118655 Chr.9: 135695008-135695008 on Build GRCh38
rs547376090 Chr.9: 135695373-135695373 on Build GRCh38
rs940011984 Chr.9: 135695191-135695191 on Build GRCh38
rs754604776 Chr.9: 135695025-135695025 on Build GRCh38
rs759760836 Chr.9: 135695004-135695004 on Build GRCh38
rs912928099 Chr.9: 135695269-135695269 on Build GRCh38
rs776667698 Chr.9: 135695181-135695181 on Build GRCh38
rs769549005 Chr.9: 135695245-135695245 on Build GRCh38
rs1049447746 Chr.9: 135695180-135695180 on Build GRCh38
rs779330888 Chr.9: 135695227-135695227 on Build GRCh38
rs773314110 Chr.9: 135695062-135695062 on Build GRCh38
rs527684978 Chr.9: 135695135-135695135 on Build GRCh38
rs775821964 Chr.9: 135695096-135695096 on Build GRCh38
rs1036298640 Chr.9: 135695428-135695428 on Build GRCh38
rs763339753 Chr.9: 135695058-135695059 on Build GRCh38
rs3119933 Chr.9: 135695121-135695121 on Build GRCh38
rs938967865 Chr.9: 135695254-135695254 on Build GRCh38
rs200025387 Chr.9: 135695374-135695380 on Build GRCh38
rs779957567 Chr.9: 135695021-135695021 on Build GRCh38
rs980677828 Chr.9: 135695237-135695237 on Build GRCh38
rs951962847 Chr.9: 135695339-135695339 on Build GRCh38
rs775086063 Chr.9: 135695184-135695184 on Build GRCh38
rs765001834 Chr.9: 135695078-135695078 on Build GRCh38
rs1050265476 Chr.9: 135695423-135695423 on Build GRCh38
rs371151926 Chr.9: 135695396-135695396 on Build GRCh38
rs748378902 Chr.9: 135695434-135695434 on Build GRCh38
rs754379420 Chr.9: 135695282-135695282 on Build GRCh38
rs757836656 Chr.9: 135695032-135695032 on Build GRCh38
rs766856400 Chr.9: 135695281-135695281 on Build GRCh38
rs377733974 Chr.9: 135695027-135695027 on Build GRCh38
rs2769033 Chr.9: 135695357-135695357 on Build GRCh38
rs369207483 Chr.9: 135695284-135695284 on Build GRCh38
rs759428205 Chr.9: 135695115-135695115 on Build GRCh38
rs370080218 Chr.9: 135695231-135695231 on Build GRCh38
rs868231548 Chr.9: 135695053-135695053 on Build GRCh38
rs761658435 Chr.9: 135695007-135695007 on Build GRCh38
rs367825361 Chr.9: 135695188-135695188 on Build GRCh38
rs551367525 Chr.9: 135695411-135695411 on Build GRCh38

View Full Product Details