Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 130075599-130076110 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR107
Entrez Gene ID
57720
Gene Name
G protein-coupled receptor 107
Gene Aliases
GCDRP, LUSTR1, bA138E2.2
UniGene
Hs.512461
Gene Chromosome Location
Chr.9: 130053706-130140169 on Build GRCh38
Transcripts
NM_001136557.1 NM_001136558.1 NM_001287346.1 NM_020960.4 XM_017014953.1 XM_017014954.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7284615 c.233A>G p.E78G Chr.9: 130075727-130075727 on Build GRCh38
6673406 c.255+1G>A p.? Chr.9: 130075750-130075750 on Build GRCh38
4407431 c.243C>T p.D81D Chr.9: 130075737-130075737 on Build GRCh38
6182794 c.188A>T p.K63M Chr.9: 130075682-130075682 on Build GRCh38
6182793 c.188A>T p.K63M Chr.9: 130075682-130075682 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs902516348 Chr.9: 130075954-130075954 on Build GRCh38
rs139020383 Chr.9: 130076026-130076026 on Build GRCh38
rs773203065 Chr.9: 130075778-130075778 on Build GRCh38
rs562094114 Chr.9: 130075837-130075837 on Build GRCh38
rs537886231 Chr.9: 130075633-130075633 on Build GRCh38
rs555526929 Chr.9: 130076084-130076084 on Build GRCh38
rs754534510 Chr.9: 130075766-130075766 on Build GRCh38
rs989556499 Chr.9: 130075970-130075970 on Build GRCh38
rs528452332 Chr.9: 130075650-130075650 on Build GRCh38
rs780939826 Chr.9: 130076017-130076017 on Build GRCh38
rs923645364 Chr.9: 130075822-130075822 on Build GRCh38
rs557763083 Chr.9: 130075642-130075642 on Build GRCh38
rs973017653 Chr.9: 130076065-130076065 on Build GRCh38
rs990966313 Chr.9: 130075935-130075935 on Build GRCh38
rs970947987 Chr.9: 130075939-130075939 on Build GRCh38
rs542439252 Chr.9: 130076072-130076072 on Build GRCh38
rs757760401 Chr.9: 130075757-130075757 on Build GRCh38
rs544235194 Chr.9: 130075852-130075852 on Build GRCh38
rs747417381 Chr.9: 130075628-130075628 on Build GRCh38
rs368890619 Chr.9: 130076058-130076058 on Build GRCh38
rs963803638 Chr.9: 130075933-130075933 on Build GRCh38
rs371130458 Chr.9: 130075748-130075748 on Build GRCh38
rs566810927 Chr.9: 130075878-130075878 on Build GRCh38
rs577845310 Chr.9: 130075680-130075680 on Build GRCh38
rs573397563 Chr.9: 130075805-130075805 on Build GRCh38
rs760385750 Chr.9: 130075781-130075782 on Build GRCh38
rs529126960 Chr.9: 130075881-130075881 on Build GRCh38
rs752679373 Chr.9: 130075743-130075743 on Build GRCh38
rs934967923 Chr.9: 130075963-130075963 on Build GRCh38
rs773115029 Chr.9: 130075689-130075689 on Build GRCh38
rs752098424 Chr.9: 130075756-130075756 on Build GRCh38
rs1017144016 Chr.9: 130076034-130076034 on Build GRCh38
rs141390611 Chr.9: 130075720-130075720 on Build GRCh38
rs914954839 Chr.9: 130075980-130075980 on Build GRCh38
rs1006688438 Chr.9: 130075857-130075857 on Build GRCh38
rs547074359 Chr.9: 130075875-130075875 on Build GRCh38
rs778915298 Chr.9: 130075784-130075784 on Build GRCh38
rs924212016 Chr.9: 130075943-130075943 on Build GRCh38
rs1029716387 Chr.9: 130076012-130076012 on Build GRCh38
rs770193583 Chr.9: 130075785-130075785 on Build GRCh38
rs749412366 Chr.9: 130075793-130075794 on Build GRCh38
rs901033966 Chr.9: 130076016-130076016 on Build GRCh38
rs950719364 Chr.9: 130075906-130075906 on Build GRCh38
rs765916024 Chr.9: 130075782-130075783 on Build GRCh38
rs376703202 Chr.9: 130075702-130075702 on Build GRCh38
rs779908757 Chr.9: 130075636-130075636 on Build GRCh38
rs998120667 Chr.9: 130075997-130075997 on Build GRCh38
rs79938857 Chr.9: 130075799-130075799 on Build GRCh38
rs558047601 Chr.9: 130075971-130075971 on Build GRCh38
rs567861797 Chr.9: 130075876-130075876 on Build GRCh38
rs73539696 Chr.9: 130075737-130075737 on Build GRCh38
rs368673169 Chr.9: 130075746-130075746 on Build GRCh38
rs780883508 Chr.9: 130075844-130075844 on Build GRCh38
rs192182817 Chr.9: 130076022-130076022 on Build GRCh38
rs1016681936 Chr.9: 130076027-130076027 on Build GRCh38
rs78098792 Chr.9: 130075801-130075801 on Build GRCh38
rs768548930 Chr.9: 130075799-130075800 on Build GRCh38
rs899248155 Chr.9: 130076056-130076056 on Build GRCh38
rs759619148 Chr.9: 130075739-130075739 on Build GRCh38
rs1045510728 Chr.9: 130075843-130075843 on Build GRCh38
rs773674300 Chr.9: 130075670-130075670 on Build GRCh38
rs747665898 Chr.9: 130075776-130075776 on Build GRCh38
rs7038342 Chr.9: 130075838-130075838 on Build GRCh38
rs150723332 Chr.9: 130075942-130075942 on Build GRCh38
rs949666138 Chr.9: 130075841-130075841 on Build GRCh38
rs191639012 Chr.9: 130075859-130075859 on Build GRCh38
rs746882999 Chr.9: 130075781-130075781 on Build GRCh38
rs11446412 Chr.9: 130075784-130075785 on Build GRCh38
rs780094676 Chr.9: 130075785-130075787 on Build GRCh38
rs768213512 Chr.9: 130075660-130075660 on Build GRCh38
rs949642056 Chr.9: 130075704-130075704 on Build GRCh38
rs182555234 Chr.9: 130075886-130075886 on Build GRCh38
rs771601173 Chr.9: 130075777-130075777 on Build GRCh38
rs10119103 Chr.9: 130075907-130075907 on Build GRCh38
rs766160806 Chr.9: 130075709-130075709 on Build GRCh38
rs781633378 Chr.9: 130075764-130075764 on Build GRCh38
rs935071266 Chr.9: 130075823-130075823 on Build GRCh38
rs551112007 Chr.9: 130075864-130075864 on Build GRCh38
rs374428340 Chr.9: 130075681-130075681 on Build GRCh38
rs201087716 Chr.9: 130075771-130075771 on Build GRCh38
rs887038156 Chr.9: 130076024-130076024 on Build GRCh38
rs1024265581 Chr.9: 130075938-130075938 on Build GRCh38
rs748942096 Chr.9: 130075657-130075657 on Build GRCh38
rs187214175 Chr.9: 130076015-130076015 on Build GRCh38
rs398012422 Chr.9: 130075800-130075801 on Build GRCh38
rs542050903 Chr.9: 130075806-130075806 on Build GRCh38
rs765381982 Chr.9: 130075740-130075740 on Build GRCh38
rs757231408 Chr.9: 130075922-130075922 on Build GRCh38
rs752520238 Chr.9: 130075785-130075786 on Build GRCh38
rs748958341 Chr.9: 130075765-130075765 on Build GRCh38
rs1016321690 Chr.9: 130075912-130075912 on Build GRCh38
rs942483312 Chr.9: 130075983-130075983 on Build GRCh38
rs778230660 Chr.9: 130075627-130075627 on Build GRCh38

View Full Product Details