Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 114131056-114131564 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC186
Entrez Gene ID
55088
Gene Name
coiled-coil domain containing 186
Gene Aliases
C10orf118
UniGene
Hs.159066
Gene Chromosome Location
Chr.10: 114120862-114174312 on Build GRCh38
Transcripts
NM_001321829.1 NM_018017.3 NR_135815.1 XM_011539915.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
380814 c.1923G>T p.L641F Chr.10: 114131325-114131325 on Build GRCh38
3358635 c.1924T>C p.L642L Chr.10: 114131324-114131324 on Build GRCh38
5836262 c.1926_1927delGA p.K643fs*15 Chr.10: 114131321-114131322 on Build GRCh38
5957205 c.1930G>C p.E644Q Chr.10: 114131318-114131318 on Build GRCh38
465288 c.1968A>G p.Q656Q Chr.10: 114131280-114131280 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs192737718 Chr.10: 114131123-114131123 on Build GRCh38
rs143034163 Chr.10: 114131211-114131211 on Build GRCh38
rs750395925 Chr.10: 114131110-114131110 on Build GRCh38
rs770799214 Chr.10: 114131365-114131365 on Build GRCh38
rs888616871 Chr.10: 114131299-114131299 on Build GRCh38
rs907384980 Chr.10: 114131535-114131535 on Build GRCh38
rs184667275 Chr.10: 114131536-114131536 on Build GRCh38
rs985957953 Chr.10: 114131498-114131498 on Build GRCh38
rs774983690 Chr.10: 114131380-114131380 on Build GRCh38
rs1041047897 Chr.10: 114131264-114131264 on Build GRCh38
rs758881972 Chr.10: 114131465-114131465 on Build GRCh38
rs754681060 Chr.10: 114131128-114131128 on Build GRCh38
rs760712044 Chr.10: 114131276-114131276 on Build GRCh38
rs781481159 Chr.10: 114131200-114131200 on Build GRCh38
rs745639342 Chr.10: 114131259-114131259 on Build GRCh38
rs201550586 Chr.10: 114131277-114131277 on Build GRCh38
rs1029504164 Chr.10: 114131426-114131426 on Build GRCh38
rs746942063 Chr.10: 114131359-114131359 on Build GRCh38
rs941008092 Chr.10: 114131242-114131242 on Build GRCh38
rs568614777 Chr.10: 114131507-114131507 on Build GRCh38
rs762724481 Chr.10: 114131386-114131386 on Build GRCh38
rs953226577 Chr.10: 114131497-114131497 on Build GRCh38
rs773114333 Chr.10: 114131349-114131349 on Build GRCh38
rs767297902 Chr.10: 114131096-114131096 on Build GRCh38
rs1045420134 Chr.10: 114131527-114131528 on Build GRCh38
rs1030918362 Chr.10: 114131103-114131103 on Build GRCh38
rs745883331 Chr.10: 114131309-114131309 on Build GRCh38
rs989359803 Chr.10: 114131131-114131131 on Build GRCh38
rs1022178778 Chr.10: 114131488-114131488 on Build GRCh38
rs375583148 Chr.10: 114131136-114131136 on Build GRCh38
rs570903969 Chr.10: 114131360-114131360 on Build GRCh38
rs762196893 Chr.10: 114131136-114131136 on Build GRCh38
rs967488732 Chr.10: 114131395-114131396 on Build GRCh38
rs752430429 Chr.10: 114131293-114131293 on Build GRCh38
rs764770535 Chr.10: 114131375-114131375 on Build GRCh38
rs985793714 Chr.10: 114131439-114131441 on Build GRCh38
rs370836670 Chr.10: 114131417-114131417 on Build GRCh38
rs758817093 Chr.10: 114131160-114131160 on Build GRCh38
rs760062676 Chr.10: 114131112-114131112 on Build GRCh38
rs763442184 Chr.10: 114131527-114131527 on Build GRCh38
rs941257843 Chr.10: 114131480-114131480 on Build GRCh38
rs906291059 Chr.10: 114131346-114131346 on Build GRCh38
rs535643018 Chr.10: 114131532-114131532 on Build GRCh38
rs778107200 Chr.10: 114131170-114131170 on Build GRCh38
rs944058983 Chr.10: 114131255-114131255 on Build GRCh38
rs550674466 Chr.10: 114131503-114131503 on Build GRCh38
rs764824037 Chr.10: 114131290-114131290 on Build GRCh38
rs771737855 Chr.10: 114131345-114131345 on Build GRCh38
rs955210916 Chr.10: 114131438-114131438 on Build GRCh38
rs868088410 Chr.10: 114131234-114131234 on Build GRCh38
rs759063400 Chr.10: 114131279-114131279 on Build GRCh38
rs34179750 Chr.10: 114131175-114131175 on Build GRCh38
rs192860566 Chr.10: 114131474-114131474 on Build GRCh38
rs756283159 Chr.10: 114131317-114131317 on Build GRCh38
rs749565293 Chr.10: 114131344-114131344 on Build GRCh38
rs115582124 Chr.10: 114131423-114131423 on Build GRCh38
rs752949470 Chr.10: 114131142-114131142 on Build GRCh38
rs73354398 Chr.10: 114131342-114131342 on Build GRCh38
rs759010330 Chr.10: 114131367-114131367 on Build GRCh38
rs771113442 Chr.10: 114131186-114131186 on Build GRCh38
rs907336413 Chr.10: 114131289-114131289 on Build GRCh38
rs142919411 Chr.10: 114131270-114131270 on Build GRCh38
rs896933849 Chr.10: 114131260-114131260 on Build GRCh38
rs199821888 Chr.10: 114131306-114131306 on Build GRCh38
rs368583662 Chr.10: 114131114-114131114 on Build GRCh38
rs775793063 Chr.10: 114131231-114131231 on Build GRCh38
rs758664195 Chr.10: 114131305-114131305 on Build GRCh38
rs911302198 Chr.10: 114131243-114131243 on Build GRCh38
rs138638702 Chr.10: 114131229-114131229 on Build GRCh38
rs377658502 Chr.10: 114131244-114131244 on Build GRCh38

View Full Product Details