Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 17089343-17089853 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIK3C2A
Entrez Gene ID
5286
Gene Name
phosphatidylinositol-4-phosphate 3-kinase catalytic subunit type 2 alpha
Gene Aliases
CPK, PI3-K-C2(ALPHA), PI3-K-C2A, PI3K-C2-alpha, PI3K-C2alpha
UniGene
Hs.175343
Gene Chromosome Location
Chr.11: 17086572-17207996 on Build GRCh38
Transcripts
NM_001321378.1 NM_001321380.1 NM_002645.3 XM_011520186.2 XM_017017924.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6746308 c.5047G>A p.A1683T Chr.11: 17089752-17089752 on Build GRCh38
925426 c.5033A>G p.Y1678C Chr.11: 17089766-17089766 on Build GRCh38
6131959 c.4954C>G p.L1652V Chr.11: 17089845-17089845 on Build GRCh38
2156641 c.4958G>A p.R1653Q Chr.11: 17089841-17089841 on Build GRCh38
5561702 c.4992T>C p.P1664P Chr.11: 17089807-17089807 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs957885462 Chr.11: 17089596-17089596 on Build GRCh38
rs562074786 Chr.11: 17089535-17089535 on Build GRCh38
rs566279890 Chr.11: 17089657-17089657 on Build GRCh38
rs948918653 Chr.11: 17089663-17089663 on Build GRCh38
rs187834222 Chr.11: 17089524-17089524 on Build GRCh38
rs573561160 Chr.11: 17089374-17089374 on Build GRCh38
rs1029709917 Chr.11: 17089506-17089506 on Build GRCh38
rs1047399694 Chr.11: 17089370-17089370 on Build GRCh38
rs146134726 Chr.11: 17089753-17089753 on Build GRCh38
rs956887768 Chr.11: 17089450-17089450 on Build GRCh38
rs768141825 Chr.11: 17089688-17089688 on Build GRCh38
rs753880826 Chr.11: 17089652-17089653 on Build GRCh38
rs201133316 Chr.11: 17089754-17089754 on Build GRCh38
rs548104917 Chr.11: 17089595-17089595 on Build GRCh38
rs903863131 Chr.11: 17089668-17089671 on Build GRCh38
rs928383563 Chr.11: 17089675-17089675 on Build GRCh38
rs374090887 Chr.11: 17089800-17089800 on Build GRCh38
rs147624382 Chr.11: 17089485-17089485 on Build GRCh38
rs375811749 Chr.11: 17089746-17089746 on Build GRCh38
rs10832733 Chr.11: 17089725-17089725 on Build GRCh38
rs747006507 Chr.11: 17089778-17089778 on Build GRCh38
rs376894515 Chr.11: 17089699-17089699 on Build GRCh38
rs750900343 Chr.11: 17089591-17089591 on Build GRCh38
rs1014178694 Chr.11: 17089481-17089481 on Build GRCh38
rs998881219 Chr.11: 17089814-17089814 on Build GRCh38
rs566921421 Chr.11: 17089749-17089749 on Build GRCh38
rs775994151 Chr.11: 17089694-17089694 on Build GRCh38
rs201587362 Chr.11: 17089665-17089665 on Build GRCh38
rs139012235 Chr.11: 17089793-17089793 on Build GRCh38
rs774501874 Chr.11: 17089767-17089774 on Build GRCh38
rs149159222 Chr.11: 17089680-17089680 on Build GRCh38
rs200196448 Chr.11: 17089777-17089777 on Build GRCh38
rs763255339 Chr.11: 17089722-17089722 on Build GRCh38
rs955427897 Chr.11: 17089508-17089508 on Build GRCh38
rs551705163 Chr.11: 17089695-17089695 on Build GRCh38
rs71844947 Chr.11: 17089662-17089663 on Build GRCh38
rs564568481 Chr.11: 17089505-17089505 on Build GRCh38
rs3802962 Chr.11: 17089480-17089480 on Build GRCh38
rs919007207 Chr.11: 17089625-17089625 on Build GRCh38
rs199758636 Chr.11: 17089667-17089667 on Build GRCh38
rs553040181 Chr.11: 17089434-17089434 on Build GRCh38
rs997264596 Chr.11: 17089490-17089490 on Build GRCh38
rs66708666 Chr.11: 17089651-17089652 on Build GRCh38
rs756398286 Chr.11: 17089745-17089745 on Build GRCh38
rs749470322 Chr.11: 17089704-17089704 on Build GRCh38
rs201880057 Chr.11: 17089818-17089818 on Build GRCh38
rs747576036 Chr.11: 17089802-17089802 on Build GRCh38
rs1042978447 Chr.11: 17089668-17089669 on Build GRCh38
rs185284146 Chr.11: 17089566-17089566 on Build GRCh38
rs560549719 Chr.11: 17089547-17089547 on Build GRCh38
rs963413651 Chr.11: 17089574-17089574 on Build GRCh38
rs751999259 Chr.11: 17089727-17089727 on Build GRCh38
rs754232217 Chr.11: 17089414-17089414 on Build GRCh38
rs192983808 Chr.11: 17089529-17089529 on Build GRCh38
rs569686458 Chr.11: 17089703-17089703 on Build GRCh38
rs936702097 Chr.11: 17089679-17089679 on Build GRCh38
rs746526234 Chr.11: 17089687-17089687 on Build GRCh38
rs987282655 Chr.11: 17089650-17089651 on Build GRCh38
rs887709299 Chr.11: 17089382-17089382 on Build GRCh38
rs142316561 Chr.11: 17089741-17089741 on Build GRCh38
rs990407725 Chr.11: 17089597-17089597 on Build GRCh38
rs1006119886 Chr.11: 17089473-17089473 on Build GRCh38
rs933127369 Chr.11: 17089648-17089648 on Build GRCh38
rs769511599 Chr.11: 17089808-17089808 on Build GRCh38
rs910438647 Chr.11: 17089666-17089667 on Build GRCh38
rs199845096 Chr.11: 17089821-17089821 on Build GRCh38
rs769027910 Chr.11: 17089723-17089726 on Build GRCh38
rs540920139 Chr.11: 17089381-17089381 on Build GRCh38
rs760034166 Chr.11: 17089732-17089732 on Build GRCh38

View Full Product Details