Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 20159059-20159576 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DBX1
Entrez Gene ID
120237
Gene Name
developing brain homeobox 1
Gene Aliases
-
UniGene
Hs.558604
Gene Chromosome Location
Chr.11: 20156214-20160324 on Build GRCh38
Transcripts
NM_001029865.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4396591 c.402C>T p.P134P Chr.11: 20159258-20159258 on Build GRCh38
925893 c.455C>A p.S152Y Chr.11: 20159205-20159205 on Build GRCh38
1684248 c.454_456delTCT p.S153delS Chr.11: 20159204-20159206 on Build GRCh38
4032031 c.412G>A p.A138T Chr.11: 20159248-20159248 on Build GRCh38
6640310 c.411C>T p.F137F Chr.11: 20159249-20159249 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752884555 Chr.11: 20159307-20159308 on Build GRCh38
rs79619657 Chr.11: 20159094-20159094 on Build GRCh38
rs565023029 Chr.11: 20159418-20159418 on Build GRCh38
rs1042601671 Chr.11: 20159425-20159425 on Build GRCh38
rs776611407 Chr.11: 20159270-20159270 on Build GRCh38
rs781586152 Chr.11: 20159212-20159212 on Build GRCh38
rs760454100 Chr.11: 20159152-20159152 on Build GRCh38
rs145387898 Chr.11: 20159259-20159259 on Build GRCh38
rs755320065 Chr.11: 20159192-20159192 on Build GRCh38
rs372162212 Chr.11: 20159314-20159314 on Build GRCh38
rs191712920 Chr.11: 20159415-20159415 on Build GRCh38
rs767463101 Chr.11: 20159508-20159508 on Build GRCh38
rs112752110 Chr.11: 20159509-20159509 on Build GRCh38
rs147543596 Chr.11: 20159229-20159229 on Build GRCh38
rs1018658922 Chr.11: 20159136-20159136 on Build GRCh38
rs112398988 Chr.11: 20159099-20159099 on Build GRCh38
rs923230679 Chr.11: 20159297-20159297 on Build GRCh38
rs370758221 Chr.11: 20159174-20159174 on Build GRCh38
rs915525364 Chr.11: 20159403-20159403 on Build GRCh38
rs900180428 Chr.11: 20159475-20159475 on Build GRCh38
rs764893319 Chr.11: 20159252-20159266 on Build GRCh38
rs575542054 Chr.11: 20159110-20159110 on Build GRCh38
rs73440478 Chr.11: 20159394-20159394 on Build GRCh38
rs938003491 Chr.11: 20159354-20159354 on Build GRCh38
rs985296331 Chr.11: 20159144-20159144 on Build GRCh38
rs752083337 Chr.11: 20159296-20159296 on Build GRCh38
rs778912827 Chr.11: 20159339-20159339 on Build GRCh38
rs546826747 Chr.11: 20159524-20159524 on Build GRCh38
rs754078533 Chr.11: 20159322-20159322 on Build GRCh38
rs768038892 Chr.11: 20159313-20159313 on Build GRCh38
rs749909452 Chr.11: 20159178-20159178 on Build GRCh38
rs777981308 Chr.11: 20159321-20159321 on Build GRCh38
rs139751615 Chr.11: 20159081-20159081 on Build GRCh38
rs544459209 Chr.11: 20159129-20159129 on Build GRCh38
rs762784070 Chr.11: 20159318-20159318 on Build GRCh38
rs1042938542 Chr.11: 20159121-20159121 on Build GRCh38
rs368653074 Chr.11: 20159276-20159276 on Build GRCh38
rs377025187 Chr.11: 20159319-20159319 on Build GRCh38
rs761732213 Chr.11: 20159273-20159273 on Build GRCh38
rs572361588 Chr.11: 20159323-20159323 on Build GRCh38
rs151039617 Chr.11: 20159290-20159290 on Build GRCh38
rs775299210 Chr.11: 20159151-20159151 on Build GRCh38
rs751532766 Chr.11: 20159334-20159335 on Build GRCh38
rs369045659 Chr.11: 20159553-20159553 on Build GRCh38
rs80216726 Chr.11: 20159481-20159481 on Build GRCh38
rs765862237 Chr.11: 20159295-20159295 on Build GRCh38
rs766853515 Chr.11: 20159183-20159183 on Build GRCh38
rs777877261 Chr.11: 20159242-20159242 on Build GRCh38
rs747004190 Chr.11: 20159266-20159266 on Build GRCh38
rs371630209 Chr.11: 20159281-20159281 on Build GRCh38
rs542394693 Chr.11: 20159350-20159350 on Build GRCh38
rs1033308721 Chr.11: 20159384-20159384 on Build GRCh38
rs772688517 Chr.11: 20159334-20159337 on Build GRCh38
rs768553723 Chr.11: 20159268-20159268 on Build GRCh38
rs764049840 Chr.11: 20159156-20159156 on Build GRCh38
rs774669055 Chr.11: 20159393-20159393 on Build GRCh38
rs1040120743 Chr.11: 20159473-20159473 on Build GRCh38
rs528848608 Chr.11: 20159309-20159309 on Build GRCh38
rs757348889 Chr.11: 20159331-20159331 on Build GRCh38
rs369552737 Chr.11: 20159204-20159206 on Build GRCh38
rs752884849 Chr.11: 20159228-20159228 on Build GRCh38
rs1008650524 Chr.11: 20159133-20159133 on Build GRCh38
rs559393788 Chr.11: 20159351-20159351 on Build GRCh38
rs761609964 Chr.11: 20159175-20159175 on Build GRCh38
rs188841521 Chr.11: 20159528-20159528 on Build GRCh38
rs144448479 Chr.11: 20159466-20159466 on Build GRCh38
rs139870931 Chr.11: 20159254-20159254 on Build GRCh38
rs1027542523 Chr.11: 20159261-20159261 on Build GRCh38
rs764810169 Chr.11: 20159176-20159176 on Build GRCh38
rs771993925 Chr.11: 20159140-20159140 on Build GRCh38
rs144494190 Chr.11: 20159335-20159336 on Build GRCh38
rs752072279 Chr.11: 20159187-20159187 on Build GRCh38
rs902288596 Chr.11: 20159132-20159132 on Build GRCh38
rs762394227 Chr.11: 20159181-20159181 on Build GRCh38
rs144687409 Chr.11: 20159200-20159202 on Build GRCh38
rs375302728 Chr.11: 20159249-20159249 on Build GRCh38
rs182718437 Chr.11: 20159445-20159445 on Build GRCh38

View Full Product Details