Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65096694-65097190 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS51
Entrez Gene ID
738
Gene Name
VPS51, GARP complex subunit
Gene Aliases
ANG2, ANG3, C11orf2, C11orf3, FFR
UniGene
Hs.277517
Gene Chromosome Location
Chr.11: 65096115-65111860 on Build GRCh38
Transcripts
NM_013265.3 NR_073519.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6281244 c.284G>A p.R95Q Chr.11: 65097053-65097053 on Build GRCh38
1194553 c.229C>A p.L77M Chr.11: 65096998-65096998 on Build GRCh38
6304002 c.272C>T p.T91M Chr.11: 65097041-65097041 on Build GRCh38
6336683 c.233G>C p.R78P Chr.11: 65097002-65097002 on Build GRCh38
458453 c.315G>C p.Q105H Chr.11: 65097084-65097084 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs898563057 Chr.11: 65096920-65096920 on Build GRCh38
rs909641535 Chr.11: 65096846-65096846 on Build GRCh38
rs1008937838 Chr.11: 65097117-65097117 on Build GRCh38
rs775334144 Chr.11: 65097025-65097025 on Build GRCh38
rs756337621 Chr.11: 65096971-65096971 on Build GRCh38
rs897374158 Chr.11: 65096856-65096856 on Build GRCh38
rs748671067 Chr.11: 65096849-65096849 on Build GRCh38
rs540082749 Chr.11: 65096845-65096845 on Build GRCh38
rs575262822 Chr.11: 65097087-65097087 on Build GRCh38
rs752462037 Chr.11: 65096998-65096998 on Build GRCh38
rs377434441 Chr.11: 65097076-65097076 on Build GRCh38
rs535199706 Chr.11: 65096979-65096979 on Build GRCh38
rs140845832 Chr.11: 65097010-65097010 on Build GRCh38
rs935204274 Chr.11: 65096783-65096783 on Build GRCh38
rs560956518 Chr.11: 65097104-65097106 on Build GRCh38
rs992781710 Chr.11: 65097086-65097086 on Build GRCh38
rs998479000 Chr.11: 65096860-65096860 on Build GRCh38
rs543884686 Chr.11: 65097139-65097139 on Build GRCh38
rs770416035 Chr.11: 65097011-65097011 on Build GRCh38
rs745479049 Chr.11: 65097023-65097023 on Build GRCh38
rs1051518233 Chr.11: 65096878-65096878 on Build GRCh38
rs1044202920 Chr.11: 65096844-65096844 on Build GRCh38
rs992875251 Chr.11: 65096794-65096794 on Build GRCh38
rs750521037 Chr.11: 65097097-65097097 on Build GRCh38
rs777440245 Chr.11: 65097142-65097142 on Build GRCh38
rs754072392 Chr.11: 65097133-65097133 on Build GRCh38
rs557697297 Chr.11: 65097145-65097145 on Build GRCh38
rs995703111 Chr.11: 65096964-65096964 on Build GRCh38
rs769592721 Chr.11: 65097024-65097024 on Build GRCh38
rs923887173 Chr.11: 65096777-65096777 on Build GRCh38
rs1045149884 Chr.11: 65096884-65096884 on Build GRCh38
rs758165452 Chr.11: 65097002-65097002 on Build GRCh38
rs573097377 Chr.11: 65096941-65096941 on Build GRCh38
rs1037221975 Chr.11: 65096855-65096855 on Build GRCh38
rs555025775 Chr.11: 65097016-65097016 on Build GRCh38
rs761952156 Chr.11: 65096950-65096950 on Build GRCh38
rs368995837 Chr.11: 65096986-65096986 on Build GRCh38
rs763347191 Chr.11: 65097037-65097037 on Build GRCh38
rs762244845 Chr.11: 65097078-65097078 on Build GRCh38
rs11548901 Chr.11: 65097123-65097123 on Build GRCh38
rs889835409 Chr.11: 65096928-65096928 on Build GRCh38
rs777761330 Chr.11: 65097004-65097004 on Build GRCh38
rs796882242 Chr.11: 65096731-65096731 on Build GRCh38
rs188037613 Chr.11: 65096951-65096951 on Build GRCh38
rs757290359 Chr.11: 65097166-65097166 on Build GRCh38
rs571059665 Chr.11: 65096826-65096826 on Build GRCh38
rs999961353 Chr.11: 65097000-65097000 on Build GRCh38
rs917949152 Chr.11: 65096811-65096811 on Build GRCh38
rs758197042 Chr.11: 65097138-65097138 on Build GRCh38
rs372618130 Chr.11: 65096988-65096988 on Build GRCh38
rs750588629 Chr.11: 65096954-65096954 on Build GRCh38
rs767645867 Chr.11: 65096953-65096953 on Build GRCh38
rs768972176 Chr.11: 65097043-65097043 on Build GRCh38
rs550925755 Chr.11: 65096819-65096819 on Build GRCh38
rs760788710 Chr.11: 65097119-65097119 on Build GRCh38
rs1056306488 Chr.11: 65096804-65096804 on Build GRCh38
rs887371601 Chr.11: 65097066-65097066 on Build GRCh38
rs377381714 Chr.11: 65096952-65096952 on Build GRCh38
rs553190774 Chr.11: 65096935-65096935 on Build GRCh38

View Full Product Details