Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65241865-65242358 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC22A20
Entrez Gene ID
440044
Gene Name
solute carrier family 22 member 20
Gene Aliases
Oat6
UniGene
Hs.532372
Gene Chromosome Location
Chr.11: 65213840-65243108 on Build GRCh38
Transcripts
NR_033396.1 XM_017017775.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7082036 c.1429G>A p.E477K Chr.11: 65242011-65242011 on Build GRCh38
4826041 c.1415G>C p.R472T Chr.11: 65241997-65241997 on Build GRCh38
930365 c.1444G>T p.E482* Chr.11: 65242026-65242026 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758554776 Chr.11: 65241988-65241988 on Build GRCh38
rs749150325 Chr.11: 65242031-65242031 on Build GRCh38
rs1055985496 Chr.11: 65241886-65241886 on Build GRCh38
rs909403836 Chr.11: 65242230-65242230 on Build GRCh38
rs546041208 Chr.11: 65242136-65242136 on Build GRCh38
rs771154975 Chr.11: 65242022-65242022 on Build GRCh38
rs1045623862 Chr.11: 65242052-65242052 on Build GRCh38
rs780449152 Chr.11: 65242096-65242096 on Build GRCh38
rs374553880 Chr.11: 65242030-65242030 on Build GRCh38
rs1018546367 Chr.11: 65242177-65242177 on Build GRCh38
rs964261076 Chr.11: 65242191-65242191 on Build GRCh38
rs142290911 Chr.11: 65242272-65242273 on Build GRCh38
rs144391337 Chr.11: 65242020-65242024 on Build GRCh38
rs905803948 Chr.11: 65242300-65242300 on Build GRCh38
rs765640310 Chr.11: 65242106-65242106 on Build GRCh38
rs774398925 Chr.11: 65242036-65242036 on Build GRCh38
rs11227164 Chr.11: 65242237-65242237 on Build GRCh38
rs771566814 Chr.11: 65242048-65242048 on Build GRCh38
rs758035829 Chr.11: 65241949-65241955 on Build GRCh38
rs981445136 Chr.11: 65242200-65242200 on Build GRCh38
rs772849205 Chr.11: 65242055-65242055 on Build GRCh38
rs185570785 Chr.11: 65241926-65241926 on Build GRCh38
rs374756976 Chr.11: 65242080-65242080 on Build GRCh38
rs1045511015 Chr.11: 65242297-65242297 on Build GRCh38
rs144802759 Chr.11: 65242284-65242284 on Build GRCh38
rs1050732427 Chr.11: 65242321-65242321 on Build GRCh38
rs371268323 Chr.11: 65242125-65242125 on Build GRCh38
rs760343943 Chr.11: 65242074-65242074 on Build GRCh38
rs988670643 Chr.11: 65242232-65242232 on Build GRCh38
rs541545891 Chr.11: 65242246-65242246 on Build GRCh38
rs760145662 Chr.11: 65241892-65241892 on Build GRCh38
rs972322752 Chr.11: 65242295-65242295 on Build GRCh38
rs979661755 Chr.11: 65242213-65242213 on Build GRCh38
rs764315877 Chr.11: 65242143-65242143 on Build GRCh38
rs935436381 Chr.11: 65241947-65241947 on Build GRCh38
rs138934452 Chr.11: 65242220-65242220 on Build GRCh38
rs890898184 Chr.11: 65242090-65242090 on Build GRCh38
rs370896455 Chr.11: 65242004-65242004 on Build GRCh38
rs994352821 Chr.11: 65242119-65242119 on Build GRCh38
rs967255198 Chr.11: 65242199-65242199 on Build GRCh38
rs1018135771 Chr.11: 65242121-65242121 on Build GRCh38
rs190288134 Chr.11: 65242218-65242218 on Build GRCh38
rs764216952 Chr.11: 65242168-65242168 on Build GRCh38
rs1050684290 Chr.11: 65242062-65242062 on Build GRCh38
rs777736441 Chr.11: 65242002-65242002 on Build GRCh38
rs905751535 Chr.11: 65242088-65242088 on Build GRCh38
rs763024524 Chr.11: 65242016-65242017 on Build GRCh38
rs763729604 Chr.11: 65241962-65241964 on Build GRCh38
rs933533749 Chr.11: 65242319-65242319 on Build GRCh38
rs73482955 Chr.11: 65242216-65242216 on Build GRCh38
rs770461438 Chr.11: 65241900-65241900 on Build GRCh38
rs142130193 Chr.11: 65242277-65242277 on Build GRCh38
rs896865547 Chr.11: 65242017-65242017 on Build GRCh38
rs919516998 Chr.11: 65242310-65242310 on Build GRCh38
rs768450816 Chr.11: 65242035-65242035 on Build GRCh38
rs942324799 Chr.11: 65241907-65241907 on Build GRCh38
rs758834084 Chr.11: 65242129-65242129 on Build GRCh38
rs898144151 Chr.11: 65242109-65242109 on Build GRCh38
rs775631807 Chr.11: 65242043-65242043 on Build GRCh38
rs368245574 Chr.11: 65242102-65242102 on Build GRCh38
rs761682873 Chr.11: 65242039-65242039 on Build GRCh38
rs753184337 Chr.11: 65241990-65241990 on Build GRCh38

View Full Product Details