Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66285296-66285803 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YIF1A
Entrez Gene ID
10897
Gene Name
Yip1 interacting factor homolog A, membrane trafficking protein
Gene Aliases
54TM, FinGER7, YIF1, YIF1P
UniGene
Hs.446445
Gene Chromosome Location
Chr.11: 66284580-66289167 on Build GRCh38
Transcripts
NM_001300861.1 NM_020470.2 XM_005273720.3 XM_017017139.1 XM_017017140.1 XM_017017141.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7229839 c.633A>G p.K211K Chr.11: 66285389-66285389 on Build GRCh38
138005 c.554C>T p.A185V Chr.11: 66285468-66285468 on Build GRCh38
7152704 c.448G>C p.V150L Chr.11: 66285738-66285738 on Build GRCh38
5360428 c.535delG p.V179fs*2 Chr.11: 66285487-66285487 on Build GRCh38
6306865 c.587G>A p.R196H Chr.11: 66285435-66285435 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763482265 Chr.11: 66285330-66285330 on Build GRCh38
rs750326919 Chr.11: 66285676-66285676 on Build GRCh38
rs758936122 Chr.11: 66285357-66285358 on Build GRCh38
rs146653044 Chr.11: 66285613-66285613 on Build GRCh38
rs757306848 Chr.11: 66285455-66285455 on Build GRCh38
rs546078849 Chr.11: 66285714-66285714 on Build GRCh38
rs984651344 Chr.11: 66285612-66285612 on Build GRCh38
rs550537586 Chr.11: 66285502-66285502 on Build GRCh38
rs927143849 Chr.11: 66285319-66285319 on Build GRCh38
rs754799758 Chr.11: 66285711-66285711 on Build GRCh38
rs764541780 Chr.11: 66285333-66285333 on Build GRCh38
rs373035564 Chr.11: 66285497-66285497 on Build GRCh38
rs372863895 Chr.11: 66285358-66285358 on Build GRCh38
rs771229106 Chr.11: 66285665-66285669 on Build GRCh38
rs915410057 Chr.11: 66285664-66285664 on Build GRCh38
rs536918442 Chr.11: 66285422-66285422 on Build GRCh38
rs750803807 Chr.11: 66285425-66285425 on Build GRCh38
rs769650237 Chr.11: 66285475-66285475 on Build GRCh38
rs556851961 Chr.11: 66285423-66285423 on Build GRCh38
rs137934642 Chr.11: 66285416-66285416 on Build GRCh38
rs749367920 Chr.11: 66285376-66285376 on Build GRCh38
rs548169858 Chr.11: 66285386-66285386 on Build GRCh38
rs763064658 Chr.11: 66285670-66285670 on Build GRCh38
rs745606472 Chr.11: 66285464-66285464 on Build GRCh38
rs369917563 Chr.11: 66285385-66285385 on Build GRCh38
rs956826334 Chr.11: 66285448-66285448 on Build GRCh38
rs774943040 Chr.11: 66285476-66285476 on Build GRCh38
rs746021032 Chr.11: 66285758-66285758 on Build GRCh38
rs201246428 Chr.11: 66285320-66285320 on Build GRCh38
rs768349464 Chr.11: 66285581-66285581 on Build GRCh38
rs752987639 Chr.11: 66285667-66285667 on Build GRCh38
rs747618755 Chr.11: 66285602-66285602 on Build GRCh38
rs1040429495 Chr.11: 66285628-66285628 on Build GRCh38
rs961499601 Chr.11: 66285556-66285556 on Build GRCh38
rs764072622 Chr.11: 66285766-66285766 on Build GRCh38
rs568782610 Chr.11: 66285652-66285652 on Build GRCh38
rs755802633 Chr.11: 66285537-66285537 on Build GRCh38
rs150672448 Chr.11: 66285531-66285531 on Build GRCh38
rs770291833 Chr.11: 66285756-66285756 on Build GRCh38
rs781271279 Chr.11: 66285670-66285670 on Build GRCh38
rs778222387 Chr.11: 66285378-66285396 on Build GRCh38
rs756330560 Chr.11: 66285430-66285430 on Build GRCh38
rs374633036 Chr.11: 66285687-66285687 on Build GRCh38
rs570757759 Chr.11: 66285433-66285433 on Build GRCh38
rs371223379 Chr.11: 66285739-66285739 on Build GRCh38
rs148222670 Chr.11: 66285440-66285440 on Build GRCh38
rs753676072 Chr.11: 66285707-66285707 on Build GRCh38
rs777306132 Chr.11: 66285745-66285745 on Build GRCh38
rs369211299 Chr.11: 66285527-66285527 on Build GRCh38
rs370078273 Chr.11: 66285681-66285681 on Build GRCh38
rs568077792 Chr.11: 66285761-66285761 on Build GRCh38
rs777399288 Chr.11: 66285733-66285733 on Build GRCh38
rs2282568 Chr.11: 66285361-66285361 on Build GRCh38
rs747314467 Chr.11: 66285488-66285489 on Build GRCh38
rs766638690 Chr.11: 66285617-66285617 on Build GRCh38
rs768888878 Chr.11: 66285377-66285377 on Build GRCh38
rs201862801 Chr.11: 66285480-66285480 on Build GRCh38
rs748928771 Chr.11: 66285580-66285580 on Build GRCh38
rs575335774 Chr.11: 66285573-66285573 on Build GRCh38
rs773912505 Chr.11: 66285493-66285493 on Build GRCh38
rs773535411 Chr.11: 66285611-66285611 on Build GRCh38
rs779970511 Chr.11: 66285567-66285567 on Build GRCh38
rs368767101 Chr.11: 66285334-66285334 on Build GRCh38
rs768054964 Chr.11: 66285498-66285498 on Build GRCh38
rs775784131 Chr.11: 66285758-66285758 on Build GRCh38
rs771478102 Chr.11: 66285731-66285731 on Build GRCh38
rs141955007 Chr.11: 66285436-66285436 on Build GRCh38
rs762845314 Chr.11: 66285618-66285618 on Build GRCh38
rs34297861 Chr.11: 66285460-66285460 on Build GRCh38
rs777794755 Chr.11: 66285380-66285380 on Build GRCh38
rs1050600491 Chr.11: 66285699-66285699 on Build GRCh38
rs754002854 Chr.11: 66285521-66285521 on Build GRCh38
rs202184738 Chr.11: 66285752-66285752 on Build GRCh38
rs779694835 Chr.11: 66285700-66285700 on Build GRCh38
rs773820851 Chr.11: 66285594-66285594 on Build GRCh38
rs747449469 Chr.11: 66285738-66285738 on Build GRCh38
rs764009134 Chr.11: 66285657-66285657 on Build GRCh38
rs748949016 Chr.11: 66285479-66285479 on Build GRCh38
rs763611495 Chr.11: 66285400-66285400 on Build GRCh38
rs559938696 Chr.11: 66285715-66285715 on Build GRCh38
rs769368772 Chr.11: 66285390-66285390 on Build GRCh38
rs754112960 Chr.11: 66285435-66285435 on Build GRCh38
rs145901576 Chr.11: 66285439-66285439 on Build GRCh38
rs771634242 Chr.11: 66285607-66285607 on Build GRCh38
rs766770020 Chr.11: 66285506-66285506 on Build GRCh38
rs376934575 Chr.11: 66285726-66285726 on Build GRCh38
rs757563807 Chr.11: 66285336-66285336 on Build GRCh38
rs140256356 Chr.11: 66285757-66285757 on Build GRCh38
rs761854899 Chr.11: 66285772-66285772 on Build GRCh38
rs985589887 Chr.11: 66285347-66285347 on Build GRCh38
rs940474108 Chr.11: 66285623-66285623 on Build GRCh38
rs762316597 Chr.11: 66285397-66285397 on Build GRCh38
rs757681308 Chr.11: 66285478-66285478 on Build GRCh38
rs951824542 Chr.11: 66285599-66285599 on Build GRCh38
rs755306836 Chr.11: 66285525-66285525 on Build GRCh38
rs765513647 Chr.11: 66285665-66285665 on Build GRCh38
rs377063479 Chr.11: 66285512-66285512 on Build GRCh38
rs768154963 Chr.11: 66285491-66285491 on Build GRCh38
rs977854411 Chr.11: 66285697-66285697 on Build GRCh38
rs758579591 Chr.11: 66285528-66285528 on Build GRCh38
rs773801488 Chr.11: 66285501-66285501 on Build GRCh38
rs74555530 Chr.11: 66285673-66285673 on Build GRCh38
rs1049247608 Chr.11: 66285339-66285339 on Build GRCh38
rs998572964 Chr.11: 66285604-66285606 on Build GRCh38

View Full Product Details