Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67437528-67438020 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPRCAP
Entrez Gene ID
5790
Gene Name
protein tyrosine phosphatase, receptor type C associated protein
Gene Aliases
CD45-AP, LPAP
UniGene
Hs.155975
Gene Chromosome Location
Chr.11: 67435510-67437682 on Build GRCh38
Transcripts
NM_005608.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5469739 c.3+9C>T p.? Chr.11: 67437608-67437608 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs773859701 Chr.11: 67437573-67437573 on Build GRCh38
rs1038833484 Chr.11: 67437872-67437872 on Build GRCh38
rs978176982 Chr.11: 67437681-67437681 on Build GRCh38
rs771618598 Chr.11: 67437669-67437669 on Build GRCh38
rs376109006 Chr.11: 67437642-67437642 on Build GRCh38
rs756388414 Chr.11: 67437632-67437632 on Build GRCh38
rs770236193 Chr.11: 67437568-67437568 on Build GRCh38
rs1054492207 Chr.11: 67437740-67437740 on Build GRCh38
rs773346021 Chr.11: 67437796-67437797 on Build GRCh38
rs1026110671 Chr.11: 67437825-67437825 on Build GRCh38
rs894550791 Chr.11: 67437745-67437745 on Build GRCh38
rs974562434 Chr.11: 67437567-67437567 on Build GRCh38
rs1010443846 Chr.11: 67437757-67437757 on Build GRCh38
rs550878107 Chr.11: 67437841-67437841 on Build GRCh38
rs902448404 Chr.11: 67437988-67437988 on Build GRCh38
rs764213205 Chr.11: 67437607-67437607 on Build GRCh38
rs907066634 Chr.11: 67437763-67437763 on Build GRCh38
rs748478948 Chr.11: 67437867-67437867 on Build GRCh38
rs781536204 Chr.11: 67437625-67437625 on Build GRCh38
rs749828019 Chr.11: 67437572-67437572 on Build GRCh38
rs992963339 Chr.11: 67437751-67437751 on Build GRCh38
rs948957916 Chr.11: 67437565-67437565 on Build GRCh38
rs772801423 Chr.11: 67437671-67437671 on Build GRCh38
rs1054128284 Chr.11: 67437934-67437934 on Build GRCh38
rs552036455 Chr.11: 67437961-67437963 on Build GRCh38
rs755432223 Chr.11: 67437793-67437793 on Build GRCh38
rs376586456 Chr.11: 67437590-67437590 on Build GRCh38
rs765879641 Chr.11: 67437631-67437631 on Build GRCh38
rs750093964 Chr.11: 67437572-67437609 on Build GRCh38
rs755779738 Chr.11: 67437628-67437629 on Build GRCh38
rs945828027 Chr.11: 67437985-67437985 on Build GRCh38
rs960448706 Chr.11: 67437696-67437696 on Build GRCh38
rs760363164 Chr.11: 67437598-67437598 on Build GRCh38
rs1010757636 Chr.11: 67437911-67437911 on Build GRCh38
rs747624666 Chr.11: 67437654-67437654 on Build GRCh38
rs753678445 Chr.11: 67437604-67437604 on Build GRCh38
rs528671701 Chr.11: 67437725-67437725 on Build GRCh38
rs370351242 Chr.11: 67437597-67437597 on Build GRCh38
rs561403125 Chr.11: 67437687-67437687 on Build GRCh38
rs869736 Chr.11: 67437991-67437991 on Build GRCh38
rs964203218 Chr.11: 67437852-67437852 on Build GRCh38
rs962697970 Chr.11: 67437813-67437813 on Build GRCh38
rs757480088 Chr.11: 67437619-67437619 on Build GRCh38
rs962730445 Chr.11: 67437556-67437556 on Build GRCh38
rs1022373941 Chr.11: 67437672-67437672 on Build GRCh38
rs925571637 Chr.11: 67437876-67437876 on Build GRCh38
rs146337170 Chr.11: 67437840-67437840 on Build GRCh38
rs879227172 Chr.11: 67437571-67437571 on Build GRCh38
rs3216798 Chr.11: 67437899-67437899 on Build GRCh38
rs186536987 Chr.11: 67437771-67437771 on Build GRCh38
rs753399697 Chr.11: 67437671-67437675 on Build GRCh38
rs544974953 Chr.11: 67437611-67437611 on Build GRCh38
rs756947153 Chr.11: 67437724-67437724 on Build GRCh38
rs891550388 Chr.11: 67437753-67437753 on Build GRCh38
rs761368949 Chr.11: 67437580-67437580 on Build GRCh38
rs543124999 Chr.11: 67437675-67437675 on Build GRCh38
rs1001436161 Chr.11: 67437788-67437788 on Build GRCh38
rs569281170 Chr.11: 67437944-67437944 on Build GRCh38
rs780504756 Chr.11: 67437756-67437756 on Build GRCh38
rs370892336 Chr.11: 67437643-67437643 on Build GRCh38
rs935629418 Chr.11: 67437889-67437889 on Build GRCh38
rs373848571 Chr.11: 67437637-67437637 on Build GRCh38
rs779461827 Chr.11: 67437649-67437649 on Build GRCh38
rs772488622 Chr.11: 67437978-67437978 on Build GRCh38
rs1034754391 Chr.11: 67437695-67437695 on Build GRCh38
rs771704733 Chr.11: 67437588-67437588 on Build GRCh38
rs1010810107 Chr.11: 67437655-67437655 on Build GRCh38
rs535853943 Chr.11: 67437846-67437846 on Build GRCh38
rs547335865 Chr.11: 67437758-67437758 on Build GRCh38
rs900380958 Chr.11: 67437883-67437883 on Build GRCh38
rs565343043 Chr.11: 67437797-67437797 on Build GRCh38
rs750726419 Chr.11: 67437627-67437627 on Build GRCh38
rs568796867 Chr.11: 67437820-67437820 on Build GRCh38

View Full Product Details