Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 132139734-132140222 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX51
Entrez Gene ID
317781
Gene Name
DEAD-box helicase 51
Gene Aliases
-
UniGene
Hs.445168
Gene Chromosome Location
Chr.12: 132136594-132144335 on Build GRCh38
Transcripts
NM_175066.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5655720 c.1839G>A p.Q613Q Chr.12: 132139861-132139861 on Build GRCh38
5727656 c.1718G>A p.G573D Chr.12: 132140155-132140155 on Build GRCh38
6505384 c.1703G>A p.G568D Chr.12: 132140170-132140170 on Build GRCh38
4440338 c.1748_1750delCCC p.P583delP Chr.12: 132140123-132140125 on Build GRCh38
1360198 c.1796C>T p.A599V Chr.12: 132139904-132139904 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763478398 Chr.12: 132139762-132139762 on Build GRCh38
rs73164956 Chr.12: 132140150-132140150 on Build GRCh38
rs778468381 Chr.12: 132139841-132139841 on Build GRCh38
rs768272737 Chr.12: 132139797-132139797 on Build GRCh38
rs926416692 Chr.12: 132139979-132139979 on Build GRCh38
rs1044111714 Chr.12: 132140016-132140016 on Build GRCh38
rs531852048 Chr.12: 132140130-132140130 on Build GRCh38
rs939218524 Chr.12: 132139782-132139782 on Build GRCh38
rs756453156 Chr.12: 132139908-132139908 on Build GRCh38
rs75236767 Chr.12: 132139765-132139765 on Build GRCh38
rs764352242 Chr.12: 132140048-132140048 on Build GRCh38
rs562690136 Chr.12: 132140085-132140085 on Build GRCh38
rs371266706 Chr.12: 132140017-132140017 on Build GRCh38
rs369388129 Chr.12: 132139848-132139848 on Build GRCh38
rs891270243 Chr.12: 132140042-132140042 on Build GRCh38
rs544858811 Chr.12: 132139984-132139984 on Build GRCh38
rs750919779 Chr.12: 132140068-132140068 on Build GRCh38
rs755656121 Chr.12: 132139820-132139821 on Build GRCh38
rs762033125 Chr.12: 132140106-132140106 on Build GRCh38
rs368978932 Chr.12: 132140204-132140204 on Build GRCh38
rs781031787 Chr.12: 132140089-132140089 on Build GRCh38
rs568996389 Chr.12: 132139821-132139821 on Build GRCh38
rs865926838 Chr.12: 132140029-132140029 on Build GRCh38
rs747897041 Chr.12: 132140184-132140184 on Build GRCh38
rs749982802 Chr.12: 132140174-132140174 on Build GRCh38
rs201693192 Chr.12: 132139973-132139973 on Build GRCh38
rs559632831 Chr.12: 132139836-132139836 on Build GRCh38
rs745694319 Chr.12: 132139958-132139958 on Build GRCh38
rs779890164 Chr.12: 132139794-132139794 on Build GRCh38
rs764262231 Chr.12: 132139890-132139890 on Build GRCh38
rs555475958 Chr.12: 132140178-132140178 on Build GRCh38
rs371612714 Chr.12: 132140177-132140177 on Build GRCh38
rs752352713 Chr.12: 132140080-132140080 on Build GRCh38
rs767607890 Chr.12: 132140107-132140107 on Build GRCh38
rs375018885 Chr.12: 132140166-132140166 on Build GRCh38
rs759688588 Chr.12: 132139824-132139824 on Build GRCh38
rs748941981 Chr.12: 132139795-132139795 on Build GRCh38
rs375884335 Chr.12: 132140188-132140188 on Build GRCh38
rs775640375 Chr.12: 132139963-132139963 on Build GRCh38
rs111929757 Chr.12: 132140115-132140115 on Build GRCh38
rs781677972 Chr.12: 132139851-132139851 on Build GRCh38
rs766822796 Chr.12: 132139972-132139972 on Build GRCh38
rs766740835 Chr.12: 132140122-132140122 on Build GRCh38
rs181335926 Chr.12: 132140030-132140030 on Build GRCh38
rs771549191 Chr.12: 132139870-132139870 on Build GRCh38
rs1052558648 Chr.12: 132139960-132139960 on Build GRCh38
rs768161730 Chr.12: 132140063-132140063 on Build GRCh38
rs371530170 Chr.12: 132140148-132140148 on Build GRCh38
rs527446214 Chr.12: 132140092-132140092 on Build GRCh38
rs1018578088 Chr.12: 132139754-132139754 on Build GRCh38
rs780089893 Chr.12: 132140176-132140176 on Build GRCh38
rs945345455 Chr.12: 132140014-132140014 on Build GRCh38
rs757006044 Chr.12: 132139944-132139945 on Build GRCh38
rs746189167 Chr.12: 132139853-132139853 on Build GRCh38
rs749026114 Chr.12: 132140099-132140099 on Build GRCh38
rs773662471 Chr.12: 132140110-132140110 on Build GRCh38
rs768549780 Chr.12: 132140139-132140139 on Build GRCh38
rs148337712 Chr.12: 132140086-132140086 on Build GRCh38
rs774667409 Chr.12: 132140105-132140105 on Build GRCh38
rs892442031 Chr.12: 132139803-132139803 on Build GRCh38
rs537678709 Chr.12: 132139873-132139873 on Build GRCh38
rs778843273 Chr.12: 132139780-132139780 on Build GRCh38
rs763781080 Chr.12: 132140126-132140126 on Build GRCh38
rs751933021 Chr.12: 132139959-132139960 on Build GRCh38
rs751269359 Chr.12: 132140165-132140165 on Build GRCh38
rs778592245 Chr.12: 132139859-132139859 on Build GRCh38
rs995778269 Chr.12: 132140076-132140076 on Build GRCh38
rs772345507 Chr.12: 132140187-132140187 on Build GRCh38
rs373307850 Chr.12: 132139895-132139895 on Build GRCh38
rs765581676 Chr.12: 132140163-132140163 on Build GRCh38
rs754038692 Chr.12: 132139775-132139775 on Build GRCh38
rs11246941 Chr.12: 132140043-132140043 on Build GRCh38
rs972454083 Chr.12: 132140022-132140022 on Build GRCh38
rs550340511 Chr.12: 132140175-132140175 on Build GRCh38
rs61756267 Chr.12: 132139769-132139769 on Build GRCh38
rs1027702039 Chr.12: 132140082-132140082 on Build GRCh38
rs777475798 Chr.12: 132139848-132139849 on Build GRCh38
rs753419593 Chr.12: 132139850-132139851 on Build GRCh38
rs773084442 Chr.12: 132139807-132139807 on Build GRCh38
rs777184659 Chr.12: 132140198-132140198 on Build GRCh38
rs770226978 Chr.12: 132140202-132140202 on Build GRCh38
rs376513403 Chr.12: 132139778-132139778 on Build GRCh38
rs770881420 Chr.12: 132140196-132140196 on Build GRCh38
rs377534239 Chr.12: 132140127-132140127 on Build GRCh38
rs770882991 Chr.12: 132139941-132139941 on Build GRCh38
rs548387793 Chr.12: 132139952-132139952 on Build GRCh38
rs146467868 Chr.12: 132140070-132140070 on Build GRCh38
rs552270104 Chr.12: 132139789-132139789 on Build GRCh38
rs117634899 Chr.12: 132140028-132140028 on Build GRCh38
rs150150422 Chr.12: 132140156-132140156 on Build GRCh38
rs200096047 Chr.12: 132139923-132139923 on Build GRCh38
rs374416628 Chr.12: 132140133-132140133 on Build GRCh38
rs766611304 Chr.12: 132139774-132139774 on Build GRCh38
rs61729149 Chr.12: 132139879-132139879 on Build GRCh38
rs1024206337 Chr.12: 132139990-132139990 on Build GRCh38
rs760602783 Chr.12: 132139876-132139876 on Build GRCh38
rs200190355 Chr.12: 132140169-132140169 on Build GRCh38
rs777548969 Chr.12: 132140001-132140001 on Build GRCh38
rs372467556 Chr.12: 132139930-132139930 on Build GRCh38
rs750785272 Chr.12: 132139907-132139907 on Build GRCh38
rs369048191 Chr.12: 132139906-132139906 on Build GRCh38
rs568166359 Chr.12: 132140091-132140091 on Build GRCh38
rs752937577 Chr.12: 132139888-132139888 on Build GRCh38
rs757974282 Chr.12: 132139931-132139931 on Build GRCh38
rs547979140 Chr.12: 132139878-132139878 on Build GRCh38
rs747158794 Chr.12: 132140195-132140195 on Build GRCh38
rs768201638 Chr.12: 132139854-132139854 on Build GRCh38
rs189746991 Chr.12: 132139985-132139985 on Build GRCh38
rs777254148 Chr.12: 132139936-132139936 on Build GRCh38
rs774411306 Chr.12: 132139764-132139764 on Build GRCh38
rs904028777 Chr.12: 132139940-132139940 on Build GRCh38
rs769599491 Chr.12: 132139959-132139959 on Build GRCh38
rs760023898 Chr.12: 132140200-132140200 on Build GRCh38
rs760956660 Chr.12: 132139937-132139937 on Build GRCh38
rs760608020 Chr.12: 132140155-132140155 on Build GRCh38
rs765432474 Chr.12: 132139829-132139829 on Build GRCh38
rs1016242526 Chr.12: 132139897-132139897 on Build GRCh38
rs771938253 Chr.12: 132139805-132139805 on Build GRCh38
rs61729150 Chr.12: 132139889-132139889 on Build GRCh38
rs769032219 Chr.12: 132140103-132140103 on Build GRCh38
rs758062417 Chr.12: 132140081-132140081 on Build GRCh38
rs779084339 Chr.12: 132139952-132139958 on Build GRCh38
rs1001421656 Chr.12: 132140003-132140003 on Build GRCh38
rs545772582 Chr.12: 132140072-132140072 on Build GRCh38
rs765399911 Chr.12: 132139885-132139885 on Build GRCh38
rs367552438 Chr.12: 132140140-132140140 on Build GRCh38
rs780739881 Chr.12: 132140129-132140129 on Build GRCh38
rs576200798 Chr.12: 132139980-132139980 on Build GRCh38
rs539483790 Chr.12: 132140153-132140153 on Build GRCh38
rs73164954 Chr.12: 132139773-132139773 on Build GRCh38
rs370529011 Chr.12: 132140132-132140132 on Build GRCh38
rs371856587 Chr.12: 132140162-132140162 on Build GRCh38
rs532512944 Chr.12: 132139784-132139784 on Build GRCh38
rs775367770 Chr.12: 132140098-132140098 on Build GRCh38
rs757760361 Chr.12: 132139849-132139849 on Build GRCh38
rs772105564 Chr.12: 132140152-132140152 on Build GRCh38
rs370972760 Chr.12: 132140088-132140088 on Build GRCh38
rs774579369 Chr.12: 132140051-132140051 on Build GRCh38
rs761234659 Chr.12: 132140117-132140117 on Build GRCh38
rs771397237 Chr.12: 132139793-132139793 on Build GRCh38
rs765712733 Chr.12: 132140062-132140062 on Build GRCh38
rs761945168 Chr.12: 132140052-132140052 on Build GRCh38
rs778384310 Chr.12: 132139785-132139785 on Build GRCh38
rs747637753 Chr.12: 132139860-132139860 on Build GRCh38
rs761324385 Chr.12: 132139798-132139798 on Build GRCh38
rs752750950 Chr.12: 132139830-132139830 on Build GRCh38
rs760512765 Chr.12: 132139815-132139815 on Build GRCh38
rs376087306 Chr.12: 132139945-132139945 on Build GRCh38

View Full Product Details