Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 39762429-39762928 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IKZF3
Entrez Gene ID
22806
Gene Name
IKAROS family zinc finger 3
Gene Aliases
AIO, AIOLOS, ZNFN1A3
UniGene
Hs.371680
Gene Chromosome Location
Chr.17: 39757715-39864188 on Build GRCh38
Transcripts
NM_001257408.1 NM_001257409.1 NM_001257410.1 NM_001257411.1 NM_001257412.1 NM_001257413.1 NM_001257414.1 NM_001284514.1 NM_001284515.1 NM_001284516.1 NM_012481.4 NM_183228.2 NM_183229.2 NM_183230.2 NM_183231.2 NM_183232.2

SNP Detail

SNP ID Chromosome Location
rs892490097 Chr.17: 39762690-39762690 on Build GRCh38
rs917586113 Chr.17: 39762879-39762879 on Build GRCh38
rs565685444 Chr.17: 39762613-39762613 on Build GRCh38
rs955506789 Chr.17: 39762753-39762753 on Build GRCh38
rs973810881 Chr.17: 39762819-39762819 on Build GRCh38
rs552906771 Chr.17: 39762829-39762829 on Build GRCh38
rs1030320471 Chr.17: 39762749-39762749 on Build GRCh38
rs373836351 Chr.17: 39762743-39762743 on Build GRCh38
rs192224858 Chr.17: 39762529-39762529 on Build GRCh38
rs548457495 Chr.17: 39762658-39762658 on Build GRCh38
rs949669069 Chr.17: 39762904-39762904 on Build GRCh38
rs918191078 Chr.17: 39762500-39762500 on Build GRCh38
rs534794387 Chr.17: 39762798-39762798 on Build GRCh38
rs537231310 Chr.17: 39762751-39762751 on Build GRCh38
rs929619166 Chr.17: 39762459-39762459 on Build GRCh38
rs991613894 Chr.17: 39762892-39762896 on Build GRCh38
rs1046708219 Chr.17: 39762493-39762493 on Build GRCh38
rs951148301 Chr.17: 39762844-39762844 on Build GRCh38
rs1045677528 Chr.17: 39762533-39762533 on Build GRCh38
rs558344523 Chr.17: 39762771-39762771 on Build GRCh38
rs903107553 Chr.17: 39762620-39762620 on Build GRCh38
rs962280595 Chr.17: 39762805-39762805 on Build GRCh38
rs935371124 Chr.17: 39762642-39762642 on Build GRCh38
rs1052854698 Chr.17: 39762664-39762664 on Build GRCh38
rs915700807 Chr.17: 39762537-39762538 on Build GRCh38
rs559496291 Chr.17: 39762609-39762610 on Build GRCh38
rs146664873 Chr.17: 39762616-39762616 on Build GRCh38
rs574765831 Chr.17: 39762832-39762832 on Build GRCh38
rs566935765 Chr.17: 39762499-39762499 on Build GRCh38
rs889252390 Chr.17: 39762762-39762762 on Build GRCh38
rs997984084 Chr.17: 39762703-39762703 on Build GRCh38
rs569876570 Chr.17: 39762663-39762663 on Build GRCh38
rs982888285 Chr.17: 39762865-39762865 on Build GRCh38
rs570580767 Chr.17: 39762782-39762782 on Build GRCh38
rs71355411 Chr.17: 39762867-39762867 on Build GRCh38
rs761993850 Chr.17: 39762766-39762766 on Build GRCh38
rs750505408 Chr.17: 39762536-39762536 on Build GRCh38
rs959830959 Chr.17: 39762888-39762888 on Build GRCh38
rs780169743 Chr.17: 39762588-39762588 on Build GRCh38
rs961665587 Chr.17: 39762504-39762504 on Build GRCh38
rs971784922 Chr.17: 39762848-39762848 on Build GRCh38
rs915748922 Chr.17: 39762895-39762895 on Build GRCh38
rs1055651420 Chr.17: 39762537-39762537 on Build GRCh38
rs1053844473 Chr.17: 39762657-39762657 on Build GRCh38

View Full Product Details