Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 39762934-39763470 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IKZF3
Entrez Gene ID
22806
Gene Name
IKAROS family zinc finger 3
Gene Aliases
AIO, AIOLOS, ZNFN1A3
UniGene
Hs.371680
Gene Chromosome Location
Chr.17: 39757715-39864188 on Build GRCh38
Transcripts
NM_001257408.1 NM_001257409.1 NM_001257410.1 NM_001257411.1 NM_001257412.1 NM_001257413.1 NM_001257414.1 NM_001284514.1 NM_001284515.1 NM_001284516.1 NM_012481.4 NM_183228.2 NM_183229.2 NM_183230.2 NM_183231.2 NM_183232.2

SNP Detail

SNP ID Chromosome Location
rs546386636 Chr.17: 39763167-39763167 on Build GRCh38
rs758548134 Chr.17: 39763341-39763341 on Build GRCh38
rs549090033 Chr.17: 39763342-39763342 on Build GRCh38
rs1050440836 Chr.17: 39763412-39763412 on Build GRCh38
rs571250885 Chr.17: 39763179-39763179 on Build GRCh38
rs576968384 Chr.17: 39763219-39763219 on Build GRCh38
rs892584821 Chr.17: 39763040-39763040 on Build GRCh38
rs1052237784 Chr.17: 39763070-39763070 on Build GRCh38
rs951242189 Chr.17: 39763238-39763238 on Build GRCh38
rs1018123596 Chr.17: 39763117-39763117 on Build GRCh38
rs563551713 Chr.17: 39763105-39763105 on Build GRCh38
rs557611415 Chr.17: 39763021-39763021 on Build GRCh38
rs541903936 Chr.17: 39762995-39762995 on Build GRCh38
rs781232826 Chr.17: 39763084-39763084 on Build GRCh38
rs1009646446 Chr.17: 39763062-39763062 on Build GRCh38
rs559447294 Chr.17: 39763364-39763365 on Build GRCh38
rs995327133 Chr.17: 39763187-39763187 on Build GRCh38
rs968428945 Chr.17: 39763256-39763256 on Build GRCh38
rs994127620 Chr.17: 39763136-39763136 on Build GRCh38
rs559534124 Chr.17: 39763405-39763405 on Build GRCh38
rs978591750 Chr.17: 39763286-39763286 on Build GRCh38
rs1052461528 Chr.17: 39762962-39762962 on Build GRCh38
rs889200196 Chr.17: 39763067-39763067 on Build GRCh38
rs923298687 Chr.17: 39763337-39763337 on Build GRCh38
rs1050899518 Chr.17: 39763005-39763005 on Build GRCh38
rs1022706056 Chr.17: 39763239-39763239 on Build GRCh38
rs1035370791 Chr.17: 39763166-39763166 on Build GRCh38
rs766575957 Chr.17: 39763014-39763014 on Build GRCh38
rs982511703 Chr.17: 39763308-39763308 on Build GRCh38
rs138745067 Chr.17: 39763433-39763433 on Build GRCh38
rs1008574910 Chr.17: 39763103-39763103 on Build GRCh38
rs67919571 Chr.17: 39763098-39763098 on Build GRCh38
rs924421199 Chr.17: 39763298-39763298 on Build GRCh38
rs933354605 Chr.17: 39763392-39763392 on Build GRCh38
rs991169058 Chr.17: 39763203-39763203 on Build GRCh38
rs970393761 Chr.17: 39763316-39763316 on Build GRCh38
rs529249551 Chr.17: 39763312-39763312 on Build GRCh38
rs570067668 Chr.17: 39762985-39762985 on Build GRCh38
rs936801032 Chr.17: 39762999-39762999 on Build GRCh38
rs537479167 Chr.17: 39762967-39762967 on Build GRCh38
rs768893887 Chr.17: 39762999-39763000 on Build GRCh38
rs751497743 Chr.17: 39763023-39763023 on Build GRCh38
rs183734888 Chr.17: 39763329-39763329 on Build GRCh38
rs910668673 Chr.17: 39763427-39763427 on Build GRCh38

View Full Product Details