Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 145926364-145925849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBM8A
Entrez Gene ID
9939
Gene Name
RNA binding motif protein 8A
Gene Aliases
BOV-1A, BOV-1B, BOV-1C, C1DELq21.1, DEL1q21.1, MDS014, RBM8, RBM8B, TAR, Y14, ZNRP, ZRNP1
UniGene
Hs.591455
Gene Chromosome Location
Chr.1: 145921556-145927536 on Build GRCh38
Transcripts
NM_005105.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
125796 c.511C>T p.R171W Chr.1: 145925896-145925896 on Build GRCh38
462852 c.491G>T p.R164I Chr.1: 145925916-145925916 on Build GRCh38
6459224 c.433A>C p.S145R Chr.1: 145926087-145926087 on Build GRCh38
4915147 c.361T>C p.Y121H Chr.1: 145926159-145926159 on Build GRCh38
674809 c.518G>T p.R173L Chr.1: 145925889-145925889 on Build GRCh38
View More

View Full Product Details