Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154829454-154829959 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMIM9
Entrez Gene ID
100132963
Gene Name
small integral membrane protein 9
Gene Aliases
CXorf68
UniGene
Hs.220762
Gene Chromosome Location
Chr.X: 154823348-154834662 on Build GRCh38
Transcripts
NM_001162936.1

SNP Detail

SNP ID Chromosome Location
rs781806798 Chr.X: 154829582-154829582 on Build GRCh38
rs782144530 Chr.X: 154829593-154829593 on Build GRCh38
rs782272298 Chr.X: 154829716-154829716 on Build GRCh38
rs781814240 Chr.X: 154829711-154829711 on Build GRCh38
rs782225641 Chr.X: 154829534-154829534 on Build GRCh38
rs782105980 Chr.X: 154829507-154829507 on Build GRCh38
rs782044602 Chr.X: 154829754-154829754 on Build GRCh38
rs782441881 Chr.X: 154829537-154829537 on Build GRCh38
rs55675924 Chr.X: 154829794-154829794 on Build GRCh38
rs904965647 Chr.X: 154829485-154829485 on Build GRCh38
rs782701288 Chr.X: 154829571-154829572 on Build GRCh38
rs143007329 Chr.X: 154829818-154829818 on Build GRCh38
rs782576603 Chr.X: 154829641-154829641 on Build GRCh38
rs781988755 Chr.X: 154829497-154829497 on Build GRCh38
rs782400103 Chr.X: 154829487-154829487 on Build GRCh38
rs782637504 Chr.X: 154829918-154829918 on Build GRCh38
rs192315779 Chr.X: 154829607-154829607 on Build GRCh38
rs915191656 Chr.X: 154829904-154829904 on Build GRCh38
rs782802569 Chr.X: 154829696-154829696 on Build GRCh38
rs1035154434 Chr.X: 154829512-154829512 on Build GRCh38
rs782731460 Chr.X: 154829760-154829760 on Build GRCh38
rs911719522 Chr.X: 154829774-154829774 on Build GRCh38
rs1044780415 Chr.X: 154829483-154829483 on Build GRCh38
rs987268784 Chr.X: 154829773-154829773 on Build GRCh38
rs782414507 Chr.X: 154829855-154829855 on Build GRCh38
rs782507630 Chr.X: 154829874-154829874 on Build GRCh38
rs975450692 Chr.X: 154829744-154829744 on Build GRCh38
rs1004866293 Chr.X: 154829617-154829617 on Build GRCh38
rs988444258 Chr.X: 154829866-154829866 on Build GRCh38
rs1017227590 Chr.X: 154829678-154829678 on Build GRCh38

View Full Product Details