Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.Y: 19459870-19460367 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCORP1
Entrez Gene ID
286554
Gene Name
BCL6 corepressor pseudogene 1
Gene Aliases
BCORL2
UniGene
Hs.211713
Gene Chromosome Location
Chr.Y: 19455431-19503153 on Build GRCh38
Transcripts
NR_002923.2 NR_033732.1

SNP Detail

SNP ID Chromosome Location
rs752470163 Chr.Y: 19460267-19460267 on Build GRCh38
rs891875526 Chr.Y: 19459994-19459994 on Build GRCh38
rs1022200829 Chr.Y: 19460045-19460045 on Build GRCh38
rs34948194 Chr.Y: 19460120-19460120 on Build GRCh38
rs766814820 Chr.Y: 19460210-19460210 on Build GRCh38
rs767397903 Chr.Y: 19460218-19460218 on Build GRCh38
rs980302813 Chr.Y: 19460199-19460199 on Build GRCh38
rs1010358151 Chr.Y: 19460036-19460036 on Build GRCh38

View Full Product Details