Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.Y: 19480068-19480574 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCORP1
Entrez Gene ID
286554
Gene Name
BCL6 corepressor pseudogene 1
Gene Aliases
BCORL2
UniGene
Hs.211713
Gene Chromosome Location
Chr.Y: 19455431-19503153 on Build GRCh38
Transcripts
NR_002923.2 NR_033732.1

SNP Detail

SNP ID Chromosome Location
rs34724285 Chr.Y: 19480410-19480410 on Build GRCh38
rs921882281 Chr.Y: 19480119-19480119 on Build GRCh38
rs893952948 Chr.Y: 19480387-19480387 on Build GRCh38
rs773348348 Chr.Y: 19480142-19480142 on Build GRCh38
rs1051769025 Chr.Y: 19480193-19480193 on Build GRCh38
rs1012424675 Chr.Y: 19480398-19480398 on Build GRCh38
rs923979528 Chr.Y: 19480261-19480261 on Build GRCh38
rs868789088 Chr.Y: 19480505-19480505 on Build GRCh38
rs760872504 Chr.Y: 19480099-19480099 on Build GRCh38
rs935405626 Chr.Y: 19480313-19480313 on Build GRCh38
rs1053823539 Chr.Y: 19480381-19480381 on Build GRCh38

View Full Product Details