Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 129840547-129841083 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM49B
Entrez Gene ID
51571
Gene Name
family with sequence similarity 49 member B
Gene Aliases
BM-009, L1
UniGene
Hs.126941
Gene Chromosome Location
Chr.8: 129839593-130016697 on Build GRCh38
Transcripts
NM_001256763.1 NM_016623.4 NR_046359.1 NR_046360.1 NR_046361.1

SNP Detail

SNP ID Chromosome Location
rs954679196 Chr.8: 129841054-129841054 on Build GRCh38
rs1045745766 Chr.8: 129840744-129840744 on Build GRCh38
rs1038406126 Chr.8: 129841021-129841021 on Build GRCh38
rs544370911 Chr.8: 129840712-129840712 on Build GRCh38
rs893306905 Chr.8: 129840828-129840828 on Build GRCh38
rs963697679 Chr.8: 129840641-129840641 on Build GRCh38
rs963645642 Chr.8: 129840891-129840891 on Build GRCh38
rs1006347155 Chr.8: 129840995-129840995 on Build GRCh38
rs984608194 Chr.8: 129840591-129840591 on Build GRCh38
rs750431242 Chr.8: 129840821-129840821 on Build GRCh38
rs747417936 Chr.8: 129841007-129841007 on Build GRCh38
rs539191773 Chr.8: 129840780-129840780 on Build GRCh38
rs780235638 Chr.8: 129841006-129841006 on Build GRCh38
rs1047255522 Chr.8: 129840812-129840812 on Build GRCh38
rs751145342 Chr.8: 129840794-129840794 on Build GRCh38
rs927256547 Chr.8: 129840749-129840749 on Build GRCh38
rs956709070 Chr.8: 129840667-129840667 on Build GRCh38
rs950850399 Chr.8: 129840583-129840583 on Build GRCh38
rs141908848 Chr.8: 129840703-129840703 on Build GRCh38
rs933001207 Chr.8: 129840675-129840675 on Build GRCh38
rs917782724 Chr.8: 129840984-129840984 on Build GRCh38
rs191598146 Chr.8: 129840954-129840954 on Build GRCh38
rs964555603 Chr.8: 129840998-129840998 on Build GRCh38
rs1013087082 Chr.8: 129840837-129840837 on Build GRCh38
rs16904157 Chr.8: 129840645-129840645 on Build GRCh38
rs988033286 Chr.8: 129840695-129840695 on Build GRCh38
rs766924433 Chr.8: 129840622-129840622 on Build GRCh38
rs527336159 Chr.8: 129840607-129840607 on Build GRCh38
rs182438271 Chr.8: 129840904-129840904 on Build GRCh38
rs139826181 Chr.8: 129840615-129840615 on Build GRCh38
rs562658190 Chr.8: 129840871-129840871 on Build GRCh38
rs71570502 Chr.8: 129840654-129840655 on Build GRCh38
rs937338822 Chr.8: 129840753-129840753 on Build GRCh38
rs950768487 Chr.8: 129840940-129840940 on Build GRCh38
rs978154130 Chr.8: 129841056-129841057 on Build GRCh38
rs1007907057 Chr.8: 129840836-129840836 on Build GRCh38
rs955184701 Chr.8: 129840601-129840601 on Build GRCh38
rs956684771 Chr.8: 129840932-129840932 on Build GRCh38
rs1017991068 Chr.8: 129840851-129840851 on Build GRCh38
rs1045054933 Chr.8: 129840852-129840852 on Build GRCh38
rs1016072207 Chr.8: 129840997-129840997 on Build GRCh38
rs1038597537 Chr.8: 129840776-129840776 on Build GRCh38
rs758644483 Chr.8: 129840935-129840935 on Build GRCh38
rs1003265471 Chr.8: 129840909-129840909 on Build GRCh38
rs541729652 Chr.8: 129840962-129840962 on Build GRCh38
rs139373792 Chr.8: 129840965-129840965 on Build GRCh38
rs557555853 Chr.8: 129840880-129840880 on Build GRCh38
rs970567832 Chr.8: 129840993-129840993 on Build GRCh38
rs937322428 Chr.8: 129841014-129841014 on Build GRCh38
rs73710949 Chr.8: 129840916-129840916 on Build GRCh38
rs925229553 Chr.8: 129840660-129840660 on Build GRCh38
rs187961028 Chr.8: 129840931-129840931 on Build GRCh38
rs371953787 Chr.8: 129840636-129840636 on Build GRCh38

View Full Product Details