Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42679883-42680380 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCR10
Entrez Gene ID
2826
Gene Name
C-C motif chemokine receptor 10
Gene Aliases
GPR2
UniGene
Hs.744542
Gene Chromosome Location
Chr.17: 42679402-42681827 on Build GRCh38
Transcripts
NM_016602.2 XM_017025238.1

Gene Symbol
CNTNAP1
Entrez Gene ID
8506
Gene Name
contactin associated protein 1
Gene Aliases
CASPR, CNTNAP, NRXN4, P190
UniGene
Hs.408730
Gene Chromosome Location
Chr.17: 42679963-42699993 on Build GRCh38
Transcripts
NM_016602.2 XM_017025238.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6623182 c.471C>T p.R157R Chr.17: 42680171-42680171 on Build GRCh38
5967067 c.691G>A p.G231S Chr.17: 42679951-42679951 on Build GRCh38
4495657 c.458C>T p.S153F Chr.17: 42680184-42680184 on Build GRCh38
6217162 c.487G>A p.V163I Chr.17: 42680155-42680155 on Build GRCh38
4670275 c.571T>C p.C191R Chr.17: 42680071-42680071 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs955181145 Chr.17: 42679994-42679994 on Build GRCh38
rs543480592 Chr.17: 42680025-42680025 on Build GRCh38
rs369737969 Chr.17: 42680172-42680172 on Build GRCh38
rs768842357 Chr.17: 42680212-42680212 on Build GRCh38
rs752063093 Chr.17: 42679923-42679923 on Build GRCh38
rs1050788014 Chr.17: 42680053-42680053 on Build GRCh38
rs763018086 Chr.17: 42680092-42680092 on Build GRCh38
rs536224679 Chr.17: 42680004-42680004 on Build GRCh38
rs571983254 Chr.17: 42679991-42679991 on Build GRCh38
rs756514733 Chr.17: 42680149-42680149 on Build GRCh38
rs774645592 Chr.17: 42679940-42679940 on Build GRCh38
rs761085990 Chr.17: 42680249-42680249 on Build GRCh38
rs772919615 Chr.17: 42680037-42680037 on Build GRCh38
rs1031510359 Chr.17: 42679960-42679960 on Build GRCh38
rs904077847 Chr.17: 42680350-42680350 on Build GRCh38
rs34866491 Chr.17: 42680177-42680177 on Build GRCh38
rs941058472 Chr.17: 42680084-42680084 on Build GRCh38
rs994745346 Chr.17: 42679907-42679907 on Build GRCh38
rs989071227 Chr.17: 42679990-42679990 on Build GRCh38
rs985398470 Chr.17: 42680009-42680009 on Build GRCh38
rs760548149 Chr.17: 42680073-42680073 on Build GRCh38
rs1034282079 Chr.17: 42680302-42680302 on Build GRCh38
rs564865053 Chr.17: 42680029-42680029 on Build GRCh38
rs776284725 Chr.17: 42680199-42680199 on Build GRCh38
rs752345698 Chr.17: 42680307-42680307 on Build GRCh38
rs766828913 Chr.17: 42680262-42680262 on Build GRCh38
rs764224857 Chr.17: 42680098-42680098 on Build GRCh38
rs760332513 Chr.17: 42680025-42680025 on Build GRCh38
rs768286532 Chr.17: 42680012-42680012 on Build GRCh38
rs768044830 Chr.17: 42680129-42680129 on Build GRCh38
rs377520676 Chr.17: 42680152-42680152 on Build GRCh38
rs777785778 Chr.17: 42680197-42680197 on Build GRCh38
rs766901487 Chr.17: 42680063-42680063 on Build GRCh38
rs750772322 Chr.17: 42679961-42679961 on Build GRCh38
rs756396744 Chr.17: 42679978-42679978 on Build GRCh38
rs754667679 Chr.17: 42680173-42680173 on Build GRCh38
rs868757027 Chr.17: 42679903-42679903 on Build GRCh38
rs868771750 Chr.17: 42679998-42679998 on Build GRCh38
rs957502629 Chr.17: 42680316-42680316 on Build GRCh38
rs753702518 Chr.17: 42680173-42680177 on Build GRCh38
rs749568925 Chr.17: 42680200-42680200 on Build GRCh38
rs762350487 Chr.17: 42680122-42680122 on Build GRCh38
rs1001277959 Chr.17: 42680357-42680357 on Build GRCh38
rs764099692 Chr.17: 42679922-42679922 on Build GRCh38
rs867757435 Chr.17: 42679987-42679987 on Build GRCh38
rs1012365278 Chr.17: 42680290-42680290 on Build GRCh38
rs773778989 Chr.17: 42680229-42680229 on Build GRCh38
rs763957331 Chr.17: 42680078-42680083 on Build GRCh38
rs990005028 Chr.17: 42680359-42680359 on Build GRCh38
rs1023822174 Chr.17: 42679934-42679934 on Build GRCh38
rs543649831 Chr.17: 42680225-42680225 on Build GRCh38
rs754715052 Chr.17: 42680268-42680268 on Build GRCh38
rs752706648 Chr.17: 42680021-42680022 on Build GRCh38
rs533628082 Chr.17: 42680114-42680114 on Build GRCh38
rs896754410 Chr.17: 42679914-42679914 on Build GRCh38
rs868232399 Chr.17: 42680088-42680088 on Build GRCh38
rs781592415 Chr.17: 42679948-42679948 on Build GRCh38
rs956775983 Chr.17: 42679988-42679988 on Build GRCh38
rs201963186 Chr.17: 42680308-42680308 on Build GRCh38
rs757938393 Chr.17: 42680181-42680181 on Build GRCh38
rs188794826 Chr.17: 42679981-42679981 on Build GRCh38
rs777080789 Chr.17: 42680339-42680339 on Build GRCh38
rs761904768 Chr.17: 42680218-42680218 on Build GRCh38
rs867124292 Chr.17: 42680248-42680248 on Build GRCh38
rs377734034 Chr.17: 42680264-42680264 on Build GRCh38
rs750846806 Chr.17: 42680141-42680141 on Build GRCh38
rs553745746 Chr.17: 42679944-42679944 on Build GRCh38
rs748835076 Chr.17: 42680005-42680005 on Build GRCh38
rs981155097 Chr.17: 42680275-42680275 on Build GRCh38
rs756878862 Chr.17: 42680264-42680267 on Build GRCh38
rs200791327 Chr.17: 42680198-42680198 on Build GRCh38
rs1012481264 Chr.17: 42679925-42679925 on Build GRCh38
rs1024979374 Chr.17: 42680242-42680242 on Build GRCh38
rs898720566 Chr.17: 42680183-42680183 on Build GRCh38
rs774518591 Chr.17: 42680216-42680216 on Build GRCh38
rs770582830 Chr.17: 42680076-42680076 on Build GRCh38
rs772110876 Chr.17: 42680036-42680036 on Build GRCh38
rs753596890 Chr.17: 42680160-42680160 on Build GRCh38
rs765010392 Chr.17: 42680297-42680297 on Build GRCh38
rs775790846 Chr.17: 42680089-42680089 on Build GRCh38
rs751960077 Chr.17: 42680156-42680156 on Build GRCh38
rs1013623634 Chr.17: 42680211-42680211 on Build GRCh38
rs769070530 Chr.17: 42679909-42679909 on Build GRCh38
rs867798743 Chr.17: 42680083-42680083 on Build GRCh38
rs763116531 Chr.17: 42679904-42679904 on Build GRCh38

View Full Product Details