Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 29333619-29334160 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP4
Entrez Gene ID
1846
Gene Name
dual specificity phosphatase 4
Gene Aliases
HVH2, MKP-2, MKP2, TYP
UniGene
Hs.417962
Gene Chromosome Location
Chr.8: 29333062-29350750 on Build GRCh38
Transcripts
NM_001394.6 NM_057158.3 XM_011544428.2

SNP Detail

SNP ID Chromosome Location
rs555492915 Chr.8: 29334083-29334083 on Build GRCh38
rs889104215 Chr.8: 29333668-29333668 on Build GRCh38
rs1009548247 Chr.8: 29333850-29333850 on Build GRCh38
rs1048680791 Chr.8: 29334006-29334006 on Build GRCh38
rs28539936 Chr.8: 29334121-29334121 on Build GRCh38
rs926767033 Chr.8: 29333686-29333686 on Build GRCh38
rs148003091 Chr.8: 29333697-29333697 on Build GRCh38
rs757021592 Chr.8: 29333846-29333846 on Build GRCh38
rs146221623 Chr.8: 29333667-29333667 on Build GRCh38
rs1049434656 Chr.8: 29333808-29333808 on Build GRCh38
rs543745457 Chr.8: 29333650-29333650 on Build GRCh38
rs949747428 Chr.8: 29333743-29333743 on Build GRCh38
rs112637547 Chr.8: 29333736-29333736 on Build GRCh38
rs929064471 Chr.8: 29333990-29333990 on Build GRCh38
rs567571597 Chr.8: 29333964-29333964 on Build GRCh38
rs892523944 Chr.8: 29334122-29334122 on Build GRCh38
rs1048714353 Chr.8: 29333725-29333725 on Build GRCh38
rs901665279 Chr.8: 29333799-29333799 on Build GRCh38
rs575829992 Chr.8: 29333881-29333881 on Build GRCh38
rs971087658 Chr.8: 29334048-29334048 on Build GRCh38
rs659481 Chr.8: 29334113-29334113 on Build GRCh38
rs1022318999 Chr.8: 29333867-29333867 on Build GRCh38
rs919087081 Chr.8: 29333753-29333753 on Build GRCh38
rs535809201 Chr.8: 29334049-29334049 on Build GRCh38
rs557163431 Chr.8: 29333872-29333872 on Build GRCh38
rs141664200 Chr.8: 29333741-29333741 on Build GRCh38
rs918971970 Chr.8: 29333965-29333965 on Build GRCh38
rs966072253 Chr.8: 29333694-29333694 on Build GRCh38
rs10100467 Chr.8: 29333707-29333707 on Build GRCh38
rs375984116 Chr.8: 29334085-29334085 on Build GRCh38
rs1034279856 Chr.8: 29333970-29333970 on Build GRCh38
rs994755665 Chr.8: 29333640-29333640 on Build GRCh38
rs957009627 Chr.8: 29333841-29333841 on Build GRCh38
rs1040342494 Chr.8: 29333864-29333864 on Build GRCh38
rs1019383276 Chr.8: 29333689-29333689 on Build GRCh38
rs960894553 Chr.8: 29333947-29333947 on Build GRCh38
rs10100468 Chr.8: 29333714-29333714 on Build GRCh38
rs779815802 Chr.8: 29333773-29333773 on Build GRCh38
rs901765913 Chr.8: 29334090-29334090 on Build GRCh38
rs981004126 Chr.8: 29333913-29333913 on Build GRCh38
rs1021039253 Chr.8: 29333698-29333698 on Build GRCh38
rs919034740 Chr.8: 29334053-29334053 on Build GRCh38
rs537334544 Chr.8: 29333798-29333798 on Build GRCh38
rs930408030 Chr.8: 29334061-29334061 on Build GRCh38
rs980909100 Chr.8: 29333674-29333674 on Build GRCh38
rs569050842 Chr.8: 29334000-29334000 on Build GRCh38
rs550943678 Chr.8: 29333824-29333824 on Build GRCh38
rs945407180 Chr.8: 29334126-29334126 on Build GRCh38
rs908868112 Chr.8: 29334020-29334020 on Build GRCh38
rs572475749 Chr.8: 29334087-29334087 on Build GRCh38
rs374027765 Chr.8: 29333793-29333793 on Build GRCh38
rs926904379 Chr.8: 29333920-29333920 on Build GRCh38
rs968227198 Chr.8: 29333705-29333705 on Build GRCh38
rs528193968 Chr.8: 29333719-29333719 on Build GRCh38
rs997799127 Chr.8: 29333823-29333823 on Build GRCh38
rs868129008 Chr.8: 29333871-29333871 on Build GRCh38
rs1009643568 Chr.8: 29334123-29334123 on Build GRCh38
rs1036156113 Chr.8: 29333752-29333752 on Build GRCh38
rs959619901 Chr.8: 29333977-29333977 on Build GRCh38
rs997319735 Chr.8: 29334100-29334100 on Build GRCh38

View Full Product Details