Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 29334434-29334943 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP4
Entrez Gene ID
1846
Gene Name
dual specificity phosphatase 4
Gene Aliases
HVH2, MKP-2, MKP2, TYP
UniGene
Hs.417962
Gene Chromosome Location
Chr.8: 29333062-29350750 on Build GRCh38
Transcripts
NM_001394.6 NM_057158.3 XM_011544428.2

SNP Detail

SNP ID Chromosome Location
rs1013628004 Chr.8: 29334669-29334669 on Build GRCh38
rs377318579 Chr.8: 29334880-29334880 on Build GRCh38
rs567533322 Chr.8: 29334618-29334618 on Build GRCh38
rs536572167 Chr.8: 29334651-29334651 on Build GRCh38
rs897383410 Chr.8: 29334602-29334602 on Build GRCh38
rs903917565 Chr.8: 29334605-29334605 on Build GRCh38
rs879096554 Chr.8: 29334631-29334631 on Build GRCh38
rs923119924 Chr.8: 29334718-29334718 on Build GRCh38
rs1025947344 Chr.8: 29334611-29334611 on Build GRCh38
rs1055246217 Chr.8: 29334645-29334645 on Build GRCh38
rs752275861 Chr.8: 29334652-29334652 on Build GRCh38
rs550805468 Chr.8: 29334600-29334600 on Build GRCh38
rs936727924 Chr.8: 29334527-29334527 on Build GRCh38
rs950882695 Chr.8: 29334508-29334508 on Build GRCh38
rs989130134 Chr.8: 29334588-29334588 on Build GRCh38
rs951841334 Chr.8: 29334637-29334637 on Build GRCh38
rs537212664 Chr.8: 29334893-29334893 on Build GRCh38
rs964876722 Chr.8: 29334717-29334717 on Build GRCh38
rs937183289 Chr.8: 29334909-29334909 on Build GRCh38
rs751541095 Chr.8: 29334834-29334834 on Build GRCh38
rs1014196596 Chr.8: 29334573-29334573 on Build GRCh38
rs964557141 Chr.8: 29334686-29334686 on Build GRCh38
rs867493517 Chr.8: 29334646-29334646 on Build GRCh38
rs886700511 Chr.8: 29334688-29334688 on Build GRCh38
rs762461128 Chr.8: 29334696-29334696 on Build GRCh38
rs117991304 Chr.8: 29334902-29334902 on Build GRCh38
rs1042340065 Chr.8: 29334472-29334472 on Build GRCh38
rs922916209 Chr.8: 29334546-29334546 on Build GRCh38
rs1015927838 Chr.8: 29334510-29334510 on Build GRCh38
rs6998333 Chr.8: 29334572-29334572 on Build GRCh38
rs190520436 Chr.8: 29334666-29334666 on Build GRCh38
rs938092881 Chr.8: 29334643-29334643 on Build GRCh38
rs996309725 Chr.8: 29334728-29334728 on Build GRCh38
rs750232218 Chr.8: 29334901-29334901 on Build GRCh38
rs773627821 Chr.8: 29334797-29334797 on Build GRCh38
rs1026592321 Chr.8: 29334488-29334488 on Build GRCh38
rs550186262 Chr.8: 29334878-29334878 on Build GRCh38
rs551231036 Chr.8: 29334498-29334498 on Build GRCh38
rs773603573 Chr.8: 29334509-29334509 on Build GRCh38
rs1014242413 Chr.8: 29334463-29334463 on Build GRCh38
rs577723196 Chr.8: 29334835-29334835 on Build GRCh38
rs947167144 Chr.8: 29334593-29334593 on Build GRCh38
rs532635458 Chr.8: 29334870-29334870 on Build GRCh38
rs945312448 Chr.8: 29334460-29334460 on Build GRCh38
rs925303267 Chr.8: 29334803-29334803 on Build GRCh38
rs1030119344 Chr.8: 29334760-29334760 on Build GRCh38
rs74690328 Chr.8: 29334900-29334900 on Build GRCh38
rs75900028 Chr.8: 29334694-29334694 on Build GRCh38
rs530677687 Chr.8: 29334592-29334592 on Build GRCh38
rs964448935 Chr.8: 29334513-29334513 on Build GRCh38
rs1055200228 Chr.8: 29334837-29334837 on Build GRCh38
rs535113142 Chr.8: 29334776-29334776 on Build GRCh38
rs1026974667 Chr.8: 29334670-29334670 on Build GRCh38
rs764935680 Chr.8: 29334576-29334576 on Build GRCh38
rs755798853 Chr.8: 29334921-29334921 on Build GRCh38
rs974350519 Chr.8: 29334530-29334530 on Build GRCh38
rs528177531 Chr.8: 29334464-29334464 on Build GRCh38
rs994429948 Chr.8: 29334607-29334607 on Build GRCh38
rs936698742 Chr.8: 29334813-29334813 on Build GRCh38
rs988501001 Chr.8: 29334853-29334853 on Build GRCh38
rs140762538 Chr.8: 29334784-29334784 on Build GRCh38

View Full Product Details