Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 29337242-29337682 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP4
Entrez Gene ID
1846
Gene Name
dual specificity phosphatase 4
Gene Aliases
HVH2, MKP-2, MKP2, TYP
UniGene
Hs.417962
Gene Chromosome Location
Chr.8: 29333062-29350750 on Build GRCh38
Transcripts
NM_001394.6 NM_057158.3 XM_011544428.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2789108 c.550C>T p.R184C Chr.8: 29337388-29337388 on Build GRCh38
2789107 c.583C>T p.R195W Chr.8: 29337355-29337355 on Build GRCh38
7365672 c.813C>A p.D271E Chr.8: 29337398-29337398 on Build GRCh38
5409396 c.867C>T p.T289T Chr.8: 29337344-29337344 on Build GRCh38
1151106 c.672C>A p.S224R Chr.8: 29337266-29337266 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755206848 Chr.8: 29337392-29337392 on Build GRCh38
rs754204824 Chr.8: 29337355-29337355 on Build GRCh38
rs375453930 Chr.8: 29337388-29337388 on Build GRCh38
rs1004262894 Chr.8: 29337393-29337393 on Build GRCh38
rs912444482 Chr.8: 29337556-29337556 on Build GRCh38
rs986666956 Chr.8: 29337467-29337467 on Build GRCh38
rs777470475 Chr.8: 29337394-29337394 on Build GRCh38
rs117758124 Chr.8: 29337356-29337356 on Build GRCh38
rs763446218 Chr.8: 29337454-29337454 on Build GRCh38
rs1019511802 Chr.8: 29337470-29337470 on Build GRCh38
rs367986568 Chr.8: 29337550-29337550 on Build GRCh38
rs760374334 Chr.8: 29337329-29337329 on Build GRCh38
rs776888629 Chr.8: 29337437-29337437 on Build GRCh38
rs1042262865 Chr.8: 29337284-29337284 on Build GRCh38
rs146191698 Chr.8: 29337407-29337407 on Build GRCh38
rs914671391 Chr.8: 29337604-29337604 on Build GRCh38
rs1044423282 Chr.8: 29337633-29337633 on Build GRCh38
rs182220993 Chr.8: 29337598-29337598 on Build GRCh38
rs771017098 Chr.8: 29337406-29337406 on Build GRCh38
rs775054923 Chr.8: 29337423-29337423 on Build GRCh38
rs7842999 Chr.8: 29337492-29337492 on Build GRCh38
rs758412658 Chr.8: 29337386-29337386 on Build GRCh38
rs555964828 Chr.8: 29337462-29337462 on Build GRCh38
rs149074309 Chr.8: 29337290-29337290 on Build GRCh38
rs770960298 Chr.8: 29337283-29337283 on Build GRCh38
rs898199432 Chr.8: 29337645-29337645 on Build GRCh38
rs17059930 Chr.8: 29337609-29337609 on Build GRCh38
rs373637449 Chr.8: 29337305-29337305 on Build GRCh38
rs764420873 Chr.8: 29337350-29337350 on Build GRCh38
rs995199777 Chr.8: 29337476-29337476 on Build GRCh38
rs750523072 Chr.8: 29337385-29337385 on Build GRCh38
rs749054331 Chr.8: 29337405-29337405 on Build GRCh38
rs961554235 Chr.8: 29337475-29337475 on Build GRCh38
rs112876436 Chr.8: 29337442-29337442 on Build GRCh38
rs769407772 Chr.8: 29337270-29337270 on Build GRCh38
rs190722868 Chr.8: 29337575-29337575 on Build GRCh38
rs923749534 Chr.8: 29337593-29337593 on Build GRCh38
rs768319639 Chr.8: 29337434-29337434 on Build GRCh38
rs980218827 Chr.8: 29337519-29337519 on Build GRCh38
rs116751946 Chr.8: 29337440-29337440 on Build GRCh38
rs771553603 Chr.8: 29337408-29337408 on Build GRCh38
rs1037066663 Chr.8: 29337639-29337639 on Build GRCh38
rs914614613 Chr.8: 29337317-29337317 on Build GRCh38
rs576877318 Chr.8: 29337426-29337426 on Build GRCh38
rs1015728886 Chr.8: 29337472-29337472 on Build GRCh38
rs985146306 Chr.8: 29337523-29337523 on Build GRCh38
rs548330426 Chr.8: 29337441-29337441 on Build GRCh38
rs965334546 Chr.8: 29337561-29337561 on Build GRCh38
rs1028109078 Chr.8: 29337453-29337453 on Build GRCh38
rs907419897 Chr.8: 29337647-29337647 on Build GRCh38
rs927434867 Chr.8: 29337541-29337541 on Build GRCh38
rs773054971 Chr.8: 29337273-29337273 on Build GRCh38
rs745783011 Chr.8: 29337455-29337477 on Build GRCh38
rs144675383 Chr.8: 29337312-29337312 on Build GRCh38
rs372203752 Chr.8: 29337387-29337387 on Build GRCh38
rs766427233 Chr.8: 29337456-29337456 on Build GRCh38
rs771654410 Chr.8: 29337522-29337522 on Build GRCh38
rs755187350 Chr.8: 29337463-29337463 on Build GRCh38
rs912488794 Chr.8: 29337268-29337268 on Build GRCh38
rs749288021 Chr.8: 29337274-29337274 on Build GRCh38
rs540275706 Chr.8: 29337600-29337600 on Build GRCh38
rs138566700 Chr.8: 29337319-29337319 on Build GRCh38
rs141546461 Chr.8: 29337365-29337365 on Build GRCh38
rs145258672 Chr.8: 29337518-29337518 on Build GRCh38

View Full Product Details