Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 73679552-73680098 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHIC1
Entrez Gene ID
53344
Gene Name
cysteine rich hydrophobic domain 1
Gene Aliases
BRX
UniGene
Hs.496323
Gene Chromosome Location
Chr.X: 73563045-73687109 on Build GRCh38
Transcripts
NM_001039840.2 NM_001300884.1 XM_017029583.1

SNP Detail

SNP ID Chromosome Location
rs867076605 Chr.X: 73679680-73679680 on Build GRCh38
rs771892526 Chr.X: 73679965-73679965 on Build GRCh38
rs990556023 Chr.X: 73679591-73679591 on Build GRCh38
rs914625537 Chr.X: 73679854-73679854 on Build GRCh38
rs1044381823 Chr.X: 73679880-73679880 on Build GRCh38
rs186805548 Chr.X: 73679813-73679813 on Build GRCh38
rs752724362 Chr.X: 73679769-73679769 on Build GRCh38
rs947458812 Chr.X: 73679868-73679868 on Build GRCh38
rs1056961424 Chr.X: 73680045-73680045 on Build GRCh38
rs777053866 Chr.X: 73679700-73679700 on Build GRCh38
rs919484814 Chr.X: 73680007-73680007 on Build GRCh38
rs923540043 Chr.X: 73679738-73679738 on Build GRCh38
rs766075118 Chr.X: 73679825-73679842 on Build GRCh38
rs867509861 Chr.X: 73679698-73679698 on Build GRCh38
rs929302802 Chr.X: 73680034-73680034 on Build GRCh38
rs1002858823 Chr.X: 73679994-73679994 on Build GRCh38
rs1051755559 Chr.X: 73680059-73680059 on Build GRCh38
rs963555716 Chr.X: 73679865-73679865 on Build GRCh38
rs760060800 Chr.X: 73680056-73680056 on Build GRCh38
rs905888148 Chr.X: 73679920-73679920 on Build GRCh38
rs771427603 Chr.X: 73679609-73679609 on Build GRCh38
rs976318311 Chr.X: 73679733-73679733 on Build GRCh38
rs745638570 Chr.X: 73679846-73679855 on Build GRCh38
rs1053728489 Chr.X: 73679840-73679840 on Build GRCh38
rs934811844 Chr.X: 73679827-73679827 on Build GRCh38
rs910882742 Chr.X: 73679645-73679645 on Build GRCh38
rs762495509 Chr.X: 73679739-73679739 on Build GRCh38
rs1001295554 Chr.X: 73679610-73679610 on Build GRCh38
rs183149182 Chr.X: 73679799-73679799 on Build GRCh38
rs774883393 Chr.X: 73679627-73679627 on Build GRCh38
rs964930265 Chr.X: 73679710-73679710 on Build GRCh38
rs781391555 Chr.X: 73679765-73679765 on Build GRCh38
rs755009621 Chr.X: 73679583-73679583 on Build GRCh38
rs1018969761 Chr.X: 73680071-73680083 on Build GRCh38
rs916105364 Chr.X: 73679811-73679811 on Build GRCh38

View Full Product Details