Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 15735204-15735760 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBC1D26
Entrez Gene ID
353149
Gene Name
TBC1 domain family member 26
Gene Aliases
-
UniGene
Hs.434123
Gene Chromosome Location
Chr.17: 15732277-15744784 on Build GRCh38
Transcripts
NM_178571.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4129604 c.101A>G p.D34G Chr.17: 15735622-15735622 on Build GRCh38
3733502 c.3G>A p.M1I Chr.17: 15735351-15735351 on Build GRCh38
4000025 c.147C>T p.L49L Chr.17: 15735668-15735668 on Build GRCh38
6023303 c.41G>A p.G14E Chr.17: 15735389-15735389 on Build GRCh38
4000026 c.147C>T p.L49L Chr.17: 15735668-15735668 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779917707 Chr.17: 15735427-15735427 on Build GRCh38
rs745627452 Chr.17: 15735317-15735317 on Build GRCh38
rs201159605 Chr.17: 15735581-15735581 on Build GRCh38
rs556973606 Chr.17: 15735600-15735600 on Build GRCh38
rs780697917 Chr.17: 15735434-15735434 on Build GRCh38
rs557119938 Chr.17: 15735232-15735232 on Build GRCh38
rs1040321622 Chr.17: 15735267-15735267 on Build GRCh38
rs375419872 Chr.17: 15735463-15735463 on Build GRCh38
rs879036503 Chr.17: 15735282-15735282 on Build GRCh38
rs367990858 Chr.17: 15735330-15735330 on Build GRCh38
rs765793574 Chr.17: 15735335-15735336 on Build GRCh38
rs758284041 Chr.17: 15735319-15735319 on Build GRCh38
rs750647744 Chr.17: 15735569-15735569 on Build GRCh38
rs182760119 Chr.17: 15735251-15735251 on Build GRCh38
rs752265454 Chr.17: 15735668-15735668 on Build GRCh38
rs748337844 Chr.17: 15735238-15735238 on Build GRCh38
rs73978520 Chr.17: 15735368-15735368 on Build GRCh38
rs754918263 Chr.17: 15735607-15735607 on Build GRCh38
rs780875595 Chr.17: 15735360-15735360 on Build GRCh38
rs542217894 Chr.17: 15735264-15735264 on Build GRCh38
rs567852346 Chr.17: 15735565-15735565 on Build GRCh38
rs754527643 Chr.17: 15735430-15735430 on Build GRCh38
rs562193446 Chr.17: 15735269-15735269 on Build GRCh38
rs202051234 Chr.17: 15735622-15735622 on Build GRCh38
rs546185482 Chr.17: 15735247-15735247 on Build GRCh38
rs764237834 Chr.17: 15735382-15735382 on Build GRCh38
rs62072376 Chr.17: 15735472-15735472 on Build GRCh38
rs573174294 Chr.17: 15735258-15735258 on Build GRCh38
rs756811086 Chr.17: 15735713-15735713 on Build GRCh38
rs371546891 Chr.17: 15735437-15735437 on Build GRCh38
rs776180360 Chr.17: 15735456-15735456 on Build GRCh38
rs201255243 Chr.17: 15735693-15735693 on Build GRCh38
rs751479870 Chr.17: 15735429-15735429 on Build GRCh38
rs776050557 Chr.17: 15735372-15735372 on Build GRCh38
rs186622578 Chr.17: 15735281-15735281 on Build GRCh38
rs772459728 Chr.17: 15735452-15735452 on Build GRCh38
rs751320186 Chr.17: 15735587-15735587 on Build GRCh38
rs547766707 Chr.17: 15735507-15735507 on Build GRCh38
rs760885675 Chr.17: 15735380-15735380 on Build GRCh38
rs201828456 Chr.17: 15735369-15735369 on Build GRCh38
rs577510691 Chr.17: 15735234-15735234 on Build GRCh38
rs985922984 Chr.17: 15735480-15735480 on Build GRCh38
rs779976893 Chr.17: 15735347-15735347 on Build GRCh38
rs371878474 Chr.17: 15735358-15735358 on Build GRCh38
rs769384152 Chr.17: 15735363-15735363 on Build GRCh38
rs200269154 Chr.17: 15735727-15735727 on Build GRCh38
rs200204898 Chr.17: 15735438-15735438 on Build GRCh38
rs747824782 Chr.17: 15735362-15735362 on Build GRCh38
rs369260191 Chr.17: 15735471-15735471 on Build GRCh38
rs761295151 Chr.17: 15735462-15735462 on Build GRCh38
rs762708547 Chr.17: 15735579-15735579 on Build GRCh38
rs746592338 Chr.17: 15735354-15735354 on Build GRCh38
rs753929084 Chr.17: 15735390-15735390 on Build GRCh38
rs765442914 Chr.17: 15735392-15735392 on Build GRCh38
rs534342357 Chr.17: 15735502-15735502 on Build GRCh38
rs577379140 Chr.17: 15735699-15735699 on Build GRCh38
rs748948865 Chr.17: 15735708-15735708 on Build GRCh38
rs758211118 Chr.17: 15735420-15735420 on Build GRCh38
rs750191939 Chr.17: 15735411-15735411 on Build GRCh38
rs369790985 Chr.17: 15735442-15735442 on Build GRCh38
rs548141614 Chr.17: 15735370-15735370 on Build GRCh38
rs778604226 Chr.17: 15735316-15735316 on Build GRCh38

View Full Product Details