Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 46615654-46616187 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HARBI1
Entrez Gene ID
283254
Gene Name
harbinger transposase derived 1
Gene Aliases
C11orf77
UniGene
Hs.714463
Gene Chromosome Location
Chr.11: 46603027-46617651 on Build GRCh38
Transcripts
NM_173811.3 XM_011520025.1 XM_011520026.2 XM_011520027.1 XM_011520028.2 XM_011520029.2 XM_017017602.1 XM_017017603.1 XM_017017604.1 XM_017017605.1 XM_017017606.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6132757 c.105G>C p.L35F Chr.11: 46616133-46616133 on Build GRCh38
4033039 c.423G>A p.M141I Chr.11: 46615815-46615815 on Build GRCh38
5980075 c.580C>T p.P194S Chr.11: 46615658-46615658 on Build GRCh38
6025154 c.470C>T p.A157V Chr.11: 46615768-46615768 on Build GRCh38
3986179 c.420G>T p.G140G Chr.11: 46615818-46615818 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773083925 Chr.11: 46615992-46615992 on Build GRCh38
rs759855163 Chr.11: 46615924-46615924 on Build GRCh38
rs909329739 Chr.11: 46615921-46615921 on Build GRCh38
rs146331633 Chr.11: 46615918-46615918 on Build GRCh38
rs749659169 Chr.11: 46615851-46615851 on Build GRCh38
rs142862456 Chr.11: 46616022-46616022 on Build GRCh38
rs536492639 Chr.11: 46615959-46615959 on Build GRCh38
rs763931346 Chr.11: 46615811-46615811 on Build GRCh38
rs755011918 Chr.11: 46615842-46615842 on Build GRCh38
rs770603042 Chr.11: 46615770-46615770 on Build GRCh38
rs1035815398 Chr.11: 46615710-46615710 on Build GRCh38
rs375003116 Chr.11: 46616155-46616155 on Build GRCh38
rs780902440 Chr.11: 46616093-46616093 on Build GRCh38
rs775259761 Chr.11: 46615809-46615809 on Build GRCh38
rs139581343 Chr.11: 46616110-46616110 on Build GRCh38
rs757926662 Chr.11: 46616070-46616070 on Build GRCh38
rs761683338 Chr.11: 46615904-46615904 on Build GRCh38
rs141436294 Chr.11: 46615858-46615858 on Build GRCh38
rs183446599 Chr.11: 46615896-46615896 on Build GRCh38
rs117030196 Chr.11: 46616113-46616113 on Build GRCh38
rs932563509 Chr.11: 46615845-46615845 on Build GRCh38
rs768672905 Chr.11: 46616128-46616128 on Build GRCh38
rs763197607 Chr.11: 46615823-46615823 on Build GRCh38
rs756640162 Chr.11: 46615849-46615849 on Build GRCh38
rs781266899 Chr.11: 46615976-46615976 on Build GRCh38
rs751591792 Chr.11: 46615720-46615720 on Build GRCh38
rs148523303 Chr.11: 46615847-46615847 on Build GRCh38
rs756192790 Chr.11: 46615806-46615806 on Build GRCh38
rs765476628 Chr.11: 46616047-46616047 on Build GRCh38
rs764226942 Chr.11: 46616039-46616039 on Build GRCh38
rs746902117 Chr.11: 46615872-46615872 on Build GRCh38
rs145989752 Chr.11: 46616042-46616042 on Build GRCh38
rs1011302103 Chr.11: 46615726-46615726 on Build GRCh38
rs776508559 Chr.11: 46615903-46615903 on Build GRCh38
rs779837091 Chr.11: 46616075-46616075 on Build GRCh38
rs775298236 Chr.11: 46615674-46615674 on Build GRCh38
rs766524874 Chr.11: 46615704-46615704 on Build GRCh38
rs974113361 Chr.11: 46616031-46616031 on Build GRCh38
rs746885615 Chr.11: 46615673-46615673 on Build GRCh38
rs772345259 Chr.11: 46615996-46615996 on Build GRCh38
rs751861091 Chr.11: 46615840-46615840 on Build GRCh38
rs776722319 Chr.11: 46616133-46616133 on Build GRCh38
rs747294798 Chr.11: 46616039-46616040 on Build GRCh38
rs778073952 Chr.11: 46615850-46615850 on Build GRCh38
rs760731188 Chr.11: 46616020-46616020 on Build GRCh38
rs143406663 Chr.11: 46615820-46615820 on Build GRCh38
rs766489500 Chr.11: 46615989-46615989 on Build GRCh38
rs962664670 Chr.11: 46615818-46615818 on Build GRCh38
rs539145552 Chr.11: 46616080-46616080 on Build GRCh38
rs553116745 Chr.11: 46615975-46615975 on Build GRCh38
rs745401140 Chr.11: 46615870-46615870 on Build GRCh38
rs778684067 Chr.11: 46616012-46616013 on Build GRCh38
rs938192727 Chr.11: 46616032-46616032 on Build GRCh38
rs149210113 Chr.11: 46615736-46615736 on Build GRCh38
rs914599692 Chr.11: 46616167-46616167 on Build GRCh38
rs745573978 Chr.11: 46615856-46615856 on Build GRCh38
rs138485412 Chr.11: 46615695-46615695 on Build GRCh38
rs756846129 Chr.11: 46615852-46615852 on Build GRCh38
rs371776479 Chr.11: 46615931-46615931 on Build GRCh38
rs115211185 Chr.11: 46615993-46615993 on Build GRCh38
rs755452054 Chr.11: 46615732-46615732 on Build GRCh38
rs758098724 Chr.11: 46615969-46615969 on Build GRCh38
rs1034125649 Chr.11: 46615702-46615702 on Build GRCh38
rs750650747 Chr.11: 46615694-46615694 on Build GRCh38
rs754962032 Chr.11: 46615998-46615999 on Build GRCh38
rs367552499 Chr.11: 46615819-46615819 on Build GRCh38
rs758875912 Chr.11: 46615686-46615686 on Build GRCh38
rs577458840 Chr.11: 46615938-46615938 on Build GRCh38
rs749994307 Chr.11: 46615967-46615967 on Build GRCh38
rs748006645 Chr.11: 46616014-46616015 on Build GRCh38
rs373717082 Chr.11: 46615743-46615743 on Build GRCh38
rs367948574 Chr.11: 46615945-46615945 on Build GRCh38
rs571242324 Chr.11: 46615746-46615746 on Build GRCh38
rs148294520 Chr.11: 46615831-46615831 on Build GRCh38
rs374607995 Chr.11: 46615940-46615940 on Build GRCh38
rs771717143 Chr.11: 46616016-46616017 on Build GRCh38
rs997588658 Chr.11: 46616122-46616122 on Build GRCh38
rs762405252 Chr.11: 46616045-46616045 on Build GRCh38
rs372565562 Chr.11: 46616095-46616095 on Build GRCh38
rs771670880 Chr.11: 46615803-46615803 on Build GRCh38
rs760542239 Chr.11: 46615678-46615678 on Build GRCh38
rs367932622 Chr.11: 46615911-46615911 on Build GRCh38
rs759785243 Chr.11: 46616012-46616012 on Build GRCh38
rs767679662 Chr.11: 46615844-46615844 on Build GRCh38
rs779382512 Chr.11: 46615963-46615963 on Build GRCh38
rs747272767 Chr.11: 46616126-46616126 on Build GRCh38
rs754400630 Chr.11: 46615950-46615950 on Build GRCh38
rs751319655 Chr.11: 46616078-46616078 on Build GRCh38
rs779362941 Chr.11: 46615974-46615974 on Build GRCh38
rs762158904 Chr.11: 46616151-46616151 on Build GRCh38
rs575249184 Chr.11: 46616118-46616118 on Build GRCh38
rs908316854 Chr.11: 46615786-46615786 on Build GRCh38
rs1005066028 Chr.11: 46615980-46615980 on Build GRCh38
rs780177490 Chr.11: 46615929-46615931 on Build GRCh38
rs745384168 Chr.11: 46615789-46615789 on Build GRCh38
rs764019374 Chr.11: 46615679-46615679 on Build GRCh38
rs749360535 Chr.11: 46615939-46615940 on Build GRCh38
rs769693436 Chr.11: 46615914-46615914 on Build GRCh38
rs773846623 Chr.11: 46615777-46615777 on Build GRCh38
rs775686124 Chr.11: 46616124-46616124 on Build GRCh38
rs893955524 Chr.11: 46616062-46616062 on Build GRCh38
rs763131121 Chr.11: 46615944-46615944 on Build GRCh38
rs772932628 Chr.11: 46616066-46616066 on Build GRCh38
rs774584740 Chr.11: 46616008-46616008 on Build GRCh38
rs748893856 Chr.11: 46615762-46615762 on Build GRCh38
rs750803133 Chr.11: 46616057-46616057 on Build GRCh38
rs777367207 Chr.11: 46616111-46616111 on Build GRCh38
rs953251761 Chr.11: 46615878-46615878 on Build GRCh38
rs748132707 Chr.11: 46615978-46615978 on Build GRCh38
rs906115753 Chr.11: 46616084-46616084 on Build GRCh38

View Full Product Details