Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 109139194-109139738 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACACB
Entrez Gene ID
32
Gene Name
acetyl-CoA carboxylase beta
Gene Aliases
ACC2, ACCB, HACC275
UniGene
Hs.234898
Gene Chromosome Location
Chr.12: 109112666-109268226 on Build GRCh38
Transcripts
NM_001093.3 XM_005253876.4 XM_011538259.2 XM_011538263.2 XM_011538264.2 XM_011538265.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4392016 c.128C>A p.P43Q Chr.12: 109139533-109139533 on Build GRCh38
1283211 c.171A>C p.T57T Chr.12: 109139576-109139576 on Build GRCh38
278566 c.324C>T p.D108D Chr.12: 109139729-109139729 on Build GRCh38
1739937 c.173C>T p.P58L Chr.12: 109139578-109139578 on Build GRCh38
370422 c.309_310CC>TA p.P103>? Chr.12: 109139714-109139715 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs578034031 Chr.12: 109139324-109139324 on Build GRCh38
rs533766108 Chr.12: 109139679-109139679 on Build GRCh38
rs748511143 Chr.12: 109139642-109139643 on Build GRCh38
rs745834531 Chr.12: 109139657-109139657 on Build GRCh38
rs370340277 Chr.12: 109139443-109139443 on Build GRCh38
rs888333193 Chr.12: 109139277-109139277 on Build GRCh38
rs1045044463 Chr.12: 109139318-109139318 on Build GRCh38
rs201767549 Chr.12: 109139711-109139711 on Build GRCh38
rs769981426 Chr.12: 109139665-109139665 on Build GRCh38
rs922357415 Chr.12: 109139663-109139663 on Build GRCh38
rs376458298 Chr.12: 109139373-109139373 on Build GRCh38
rs571766506 Chr.12: 109139606-109139606 on Build GRCh38
rs1036164921 Chr.12: 109139367-109139367 on Build GRCh38
rs767325347 Chr.12: 109139527-109139527 on Build GRCh38
rs371386144 Chr.12: 109139661-109139661 on Build GRCh38
rs757194840 Chr.12: 109139651-109139651 on Build GRCh38
rs760884462 Chr.12: 109139486-109139486 on Build GRCh38
rs905134619 Chr.12: 109139550-109139550 on Build GRCh38
rs773226498 Chr.12: 109139602-109139602 on Build GRCh38
rs555738682 Chr.12: 109139699-109139699 on Build GRCh38
rs748081662 Chr.12: 109139387-109139387 on Build GRCh38
rs777716398 Chr.12: 109139459-109139459 on Build GRCh38
rs376872449 Chr.12: 109139431-109139431 on Build GRCh38
rs769916566 Chr.12: 109139391-109139391 on Build GRCh38
rs769946341 Chr.12: 109139578-109139578 on Build GRCh38
rs368045407 Chr.12: 109139627-109139627 on Build GRCh38
rs765824239 Chr.12: 109139415-109139415 on Build GRCh38
rs767936750 Chr.12: 109139686-109139686 on Build GRCh38
rs140156252 Chr.12: 109139667-109139667 on Build GRCh38
rs753548861 Chr.12: 109139703-109139703 on Build GRCh38
rs144316349 Chr.12: 109139455-109139455 on Build GRCh38
rs769849813 Chr.12: 109139590-109139590 on Build GRCh38
rs780859563 Chr.12: 109139383-109139383 on Build GRCh38
rs774959869 Chr.12: 109139671-109139671 on Build GRCh38
rs761835144 Chr.12: 109139498-109139498 on Build GRCh38
rs11065655 Chr.12: 109139660-109139660 on Build GRCh38
rs763378476 Chr.12: 109139510-109139510 on Build GRCh38
rs763702236 Chr.12: 109139701-109139701 on Build GRCh38
rs1003407293 Chr.12: 109139338-109139338 on Build GRCh38
rs551468976 Chr.12: 109139664-109139664 on Build GRCh38
rs748370787 Chr.12: 109139579-109139579 on Build GRCh38
rs755735357 Chr.12: 109139628-109139628 on Build GRCh38
rs1046741460 Chr.12: 109139270-109139270 on Build GRCh38
rs780843060 Chr.12: 109139446-109139446 on Build GRCh38
rs766749273 Chr.12: 109139512-109139512 on Build GRCh38
rs747698774 Chr.12: 109139452-109139452 on Build GRCh38
rs191728558 Chr.12: 109139326-109139326 on Build GRCh38
rs531976358 Chr.12: 109139545-109139545 on Build GRCh38
rs761186241 Chr.12: 109139708-109139708 on Build GRCh38
rs550116625 Chr.12: 109139568-109139568 on Build GRCh38
rs749213609 Chr.12: 109139475-109139475 on Build GRCh38
rs754359144 Chr.12: 109139265-109139265 on Build GRCh38
rs778648797 Chr.12: 109139574-109139574 on Build GRCh38
rs755417058 Chr.12: 109139303-109139303 on Build GRCh38
rs745709988 Chr.12: 109139576-109139576 on Build GRCh38
rs929610160 Chr.12: 109139259-109139259 on Build GRCh38
rs922401906 Chr.12: 109139253-109139253 on Build GRCh38
rs756710601 Chr.12: 109139561-109139561 on Build GRCh38
rs139767464 Chr.12: 109139539-109139539 on Build GRCh38
rs79484712 Chr.12: 109139424-109139424 on Build GRCh38
rs768907972 Chr.12: 109139492-109139492 on Build GRCh38
rs776063981 Chr.12: 109139687-109139687 on Build GRCh38
rs770652656 Chr.12: 109139400-109139400 on Build GRCh38
rs143135974 Chr.12: 109139618-109139618 on Build GRCh38
rs562661753 Chr.12: 109139280-109139280 on Build GRCh38
rs753987065 Chr.12: 109139414-109139414 on Build GRCh38
rs373682906 Chr.12: 109139408-109139408 on Build GRCh38
rs763514954 Chr.12: 109139604-109139604 on Build GRCh38
rs750878973 Chr.12: 109139509-109139509 on Build GRCh38
rs781472343 Chr.12: 109139642-109139642 on Build GRCh38
rs370897180 Chr.12: 109139394-109139394 on Build GRCh38
rs1033644454 Chr.12: 109139551-109139551 on Build GRCh38
rs755458548 Chr.12: 109139454-109139454 on Build GRCh38
rs775740191 Chr.12: 109139402-109139402 on Build GRCh38
rs754732999 Chr.12: 109139445-109139445 on Build GRCh38
rs1010623826 Chr.12: 109139379-109139379 on Build GRCh38
rs146888683 Chr.12: 109139534-109139534 on Build GRCh38
rs757532146 Chr.12: 109139409-109139409 on Build GRCh38
rs773313448 Chr.12: 109139666-109139666 on Build GRCh38
rs778771224 Chr.12: 109139655-109139655 on Build GRCh38
rs142445607 Chr.12: 109139533-109139533 on Build GRCh38
rs200591816 Chr.12: 109139668-109139668 on Build GRCh38
rs776672844 Chr.12: 109139497-109139497 on Build GRCh38
rs747603571 Chr.12: 109139313-109139313 on Build GRCh38
rs371940807 Chr.12: 109139639-109139639 on Build GRCh38
rs375344890 Chr.12: 109139614-109139614 on Build GRCh38
rs757016767 Chr.12: 109139677-109139677 on Build GRCh38
rs753327617 Chr.12: 109139649-109139649 on Build GRCh38
rs527664804 Chr.12: 109139525-109139525 on Build GRCh38
rs762853678 Chr.12: 109139396-109139396 on Build GRCh38
rs770693620 Chr.12: 109139485-109139485 on Build GRCh38
rs866351563 Chr.12: 109139616-109139616 on Build GRCh38

View Full Product Details