Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 24138539-24139115 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CABYR
Entrez Gene ID
26256
Gene Name
calcium binding tyrosine phosphorylation regulated
Gene Aliases
CABYRa, CABYRc, CABYRc/d, CABYRe, CBP86, CT88, FSP-2, FSP2
UniGene
Hs.511983
Gene Chromosome Location
Chr.18: 24138956-24161600 on Build GRCh38
Transcripts
NM_001308231.1 NM_012189.3 NM_138643.2 NM_153768.2 NM_153769.2 NM_153770.2 XM_005258247.2 XM_017025706.1

SNP Detail

SNP ID Chromosome Location
rs919539532 Chr.18: 24139068-24139068 on Build GRCh38
rs1012008562 Chr.18: 24138986-24138986 on Build GRCh38
rs530643257 Chr.18: 24138810-24138810 on Build GRCh38
rs115019087 Chr.18: 24138625-24138625 on Build GRCh38
rs866393176 Chr.18: 24139048-24139048 on Build GRCh38
rs973395808 Chr.18: 24138983-24138983 on Build GRCh38
rs980811442 Chr.18: 24138697-24138697 on Build GRCh38
rs1045617360 Chr.18: 24139042-24139042 on Build GRCh38
rs139130022 Chr.18: 24138633-24138633 on Build GRCh38
rs780792689 Chr.18: 24139078-24139078 on Build GRCh38
rs915367965 Chr.18: 24138867-24138867 on Build GRCh38
rs11876575 Chr.18: 24138980-24138980 on Build GRCh38
rs930884446 Chr.18: 24139070-24139070 on Build GRCh38
rs1025158153 Chr.18: 24138987-24139001 on Build GRCh38
rs929484765 Chr.18: 24138815-24138815 on Build GRCh38
rs9675662 Chr.18: 24138981-24138981 on Build GRCh38
rs1024117812 Chr.18: 24138923-24138923 on Build GRCh38
rs896804649 Chr.18: 24138947-24138947 on Build GRCh38
rs1050634363 Chr.18: 24138584-24138584 on Build GRCh38
rs975390964 Chr.18: 24139004-24139004 on Build GRCh38
rs950773849 Chr.18: 24138994-24138994 on Build GRCh38
rs912060969 Chr.18: 24138911-24138911 on Build GRCh38
rs956729669 Chr.18: 24139027-24139027 on Build GRCh38
rs1045246690 Chr.18: 24139088-24139088 on Build GRCh38
rs777831371 Chr.18: 24138996-24138996 on Build GRCh38
rs75141028 Chr.18: 24138836-24138836 on Build GRCh38
rs1038718709 Chr.18: 24138873-24138873 on Build GRCh38
rs1001493584 Chr.18: 24139049-24139049 on Build GRCh38
rs568183034 Chr.18: 24139015-24139015 on Build GRCh38
rs906358898 Chr.18: 24139092-24139092 on Build GRCh38
rs953436045 Chr.18: 24139046-24139046 on Build GRCh38
rs947760700 Chr.18: 24139085-24139085 on Build GRCh38
rs546914046 Chr.18: 24138916-24138916 on Build GRCh38
rs929665492 Chr.18: 24138951-24138951 on Build GRCh38
rs891624924 Chr.18: 24138598-24138598 on Build GRCh38
rs1048056822 Chr.18: 24139077-24139077 on Build GRCh38
rs1022262204 Chr.18: 24139044-24139044 on Build GRCh38
rs548790214 Chr.18: 24139009-24139009 on Build GRCh38
rs896831474 Chr.18: 24138617-24138617 on Build GRCh38
rs943672262 Chr.18: 24138998-24138998 on Build GRCh38
rs957618000 Chr.18: 24139096-24139096 on Build GRCh38
rs1023473657 Chr.18: 24138729-24138729 on Build GRCh38
rs973894185 Chr.18: 24138804-24138804 on Build GRCh38
rs932070668 Chr.18: 24138887-24138887 on Build GRCh38
rs557025066 Chr.18: 24139053-24139053 on Build GRCh38
rs943604045 Chr.18: 24138846-24138846 on Build GRCh38
rs746839018 Chr.18: 24139007-24139007 on Build GRCh38
rs559685526 Chr.18: 24138585-24138585 on Build GRCh38
rs553562853 Chr.18: 24138880-24138881 on Build GRCh38
rs551858631 Chr.18: 24139016-24139016 on Build GRCh38
rs1053285214 Chr.18: 24138888-24138888 on Build GRCh38
rs949770846 Chr.18: 24139038-24139038 on Build GRCh38
rs190467787 Chr.18: 24138607-24138607 on Build GRCh38
rs974897088 Chr.18: 24139087-24139087 on Build GRCh38
rs1031372794 Chr.18: 24139017-24139017 on Build GRCh38
rs761000967 Chr.18: 24138682-24138682 on Build GRCh38
rs891508968 Chr.18: 24138891-24138891 on Build GRCh38
rs371432073 Chr.18: 24139028-24139029 on Build GRCh38
rs780595448 Chr.18: 24138753-24138754 on Build GRCh38
rs905809116 Chr.18: 24138929-24138929 on Build GRCh38
rs755246516 Chr.18: 24138793-24138794 on Build GRCh38
rs912044013 Chr.18: 24139051-24139051 on Build GRCh38
rs989517853 Chr.18: 24138865-24138865 on Build GRCh38
rs78688037 Chr.18: 24138852-24138852 on Build GRCh38
rs1037043744 Chr.18: 24138925-24138925 on Build GRCh38
rs2278663 Chr.18: 24138841-24138841 on Build GRCh38
rs890669453 Chr.18: 24138598-24138598 on Build GRCh38
rs1003826336 Chr.18: 24139095-24139095 on Build GRCh38
rs1034085093 Chr.18: 24138789-24138789 on Build GRCh38
rs766010220 Chr.18: 24138717-24138717 on Build GRCh38
rs1008932791 Chr.18: 24139072-24139072 on Build GRCh38
rs117082687 Chr.18: 24138799-24138799 on Build GRCh38
rs533074510 Chr.18: 24138903-24138903 on Build GRCh38
rs1040687267 Chr.18: 24138609-24138609 on Build GRCh38
rs373950699 Chr.18: 24139080-24139080 on Build GRCh38
rs1026763893 Chr.18: 24138639-24138639 on Build GRCh38
rs986228798 Chr.18: 24138910-24138910 on Build GRCh38
rs537559831 Chr.18: 24139036-24139036 on Build GRCh38
rs770747019 Chr.18: 24139054-24139054 on Build GRCh38

View Full Product Details