Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 14148099-14148697 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRL1
Entrez Gene ID
22859
Gene Name
adhesion G protein-coupled receptor L1
Gene Aliases
CIRL1, CL1, LEC2, LPHN1
UniGene
Hs.94229
Gene Chromosome Location
Chr.19: 14147737-14206245 on Build GRCh38
Transcripts
NM_001008701.2 NM_014921.4 XM_005259818.3 XM_011527796.2 XM_011527798.2 XM_011527801.2 XM_017026475.1 XM_017026476.1 XM_017026477.1 XM_017026478.1 XM_017026479.1 XM_017026480.1 NR_045214.1

Gene Symbol
LOC100507373
Entrez Gene ID
100507373
Gene Name
uncharacterized LOC100507373
Gene Aliases
-
UniGene
Hs.720437
Gene Chromosome Location
Chr.19: 14137152-14171264 on Build GRCh38
Transcripts
NM_001008701.2 NM_014921.4 XM_005259818.3 XM_011527796.2 XM_011527798.2 XM_011527801.2 XM_017026475.1 XM_017026476.1 XM_017026477.1 XM_017026478.1 XM_017026479.1 XM_017026480.1 NR_045214.1

SNP Detail

SNP ID Chromosome Location
rs1009865808 Chr.19: 14148249-14148249 on Build GRCh38
rs978979103 Chr.19: 14148530-14148530 on Build GRCh38
rs939492160 Chr.19: 14148507-14148507 on Build GRCh38
rs190172662 Chr.19: 14148621-14148621 on Build GRCh38
rs947567990 Chr.19: 14148312-14148312 on Build GRCh38
rs930887944 Chr.19: 14148160-14148160 on Build GRCh38
rs144853952 Chr.19: 14148313-14148313 on Build GRCh38
rs868564859 Chr.19: 14148617-14148617 on Build GRCh38
rs567043431 Chr.19: 14148309-14148309 on Build GRCh38
rs754290438 Chr.19: 14148378-14148379 on Build GRCh38
rs926906392 Chr.19: 14148489-14148489 on Build GRCh38
rs762318181 Chr.19: 14148399-14148399 on Build GRCh38
rs536945423 Chr.19: 14148377-14148377 on Build GRCh38
rs558552974 Chr.19: 14148420-14148420 on Build GRCh38
rs917646801 Chr.19: 14148567-14148567 on Build GRCh38
rs905051005 Chr.19: 14148193-14148193 on Build GRCh38
rs539168133 Chr.19: 14148253-14148253 on Build GRCh38
rs760516718 Chr.19: 14148378-14148378 on Build GRCh38
rs747773982 Chr.19: 14148360-14148360 on Build GRCh38
rs949091719 Chr.19: 14148589-14148589 on Build GRCh38
rs925034818 Chr.19: 14148318-14148318 on Build GRCh38
rs746274553 Chr.19: 14148241-14148241 on Build GRCh38
rs766150764 Chr.19: 14148488-14148488 on Build GRCh38
rs1056637615 Chr.19: 14148512-14148512 on Build GRCh38
rs866477475 Chr.19: 14148295-14148295 on Build GRCh38
rs781708192 Chr.19: 14148161-14148161 on Build GRCh38
rs1019683844 Chr.19: 14148440-14148440 on Build GRCh38
rs550193390 Chr.19: 14148165-14148165 on Build GRCh38
rs988318447 Chr.19: 14148550-14148550 on Build GRCh38
rs975667543 Chr.19: 14148259-14148259 on Build GRCh38
rs1009114603 Chr.19: 14148639-14148639 on Build GRCh38
rs972157627 Chr.19: 14148432-14148432 on Build GRCh38
rs995316489 Chr.19: 14148214-14148214 on Build GRCh38
rs905339027 Chr.19: 14148181-14148181 on Build GRCh38
rs760142393 Chr.19: 14148334-14148334 on Build GRCh38
rs1048338542 Chr.19: 14148657-14148659 on Build GRCh38
rs759452883 Chr.19: 14148628-14148628 on Build GRCh38
rs930667619 Chr.19: 14148460-14148460 on Build GRCh38
rs771669663 Chr.19: 14148369-14148369 on Build GRCh38
rs949061730 Chr.19: 14148358-14148358 on Build GRCh38
rs982807235 Chr.19: 14148244-14148244 on Build GRCh38
rs576750599 Chr.19: 14148595-14148595 on Build GRCh38
rs1003740603 Chr.19: 14148196-14148196 on Build GRCh38
rs1000095606 Chr.19: 14148310-14148310 on Build GRCh38
rs1044713100 Chr.19: 14148597-14148597 on Build GRCh38
rs757496334 Chr.19: 14148142-14148142 on Build GRCh38
rs73509752 Chr.19: 14148599-14148599 on Build GRCh38
rs904177275 Chr.19: 14148570-14148570 on Build GRCh38
rs924806678 Chr.19: 14148531-14148531 on Build GRCh38
rs920591945 Chr.19: 14148454-14148454 on Build GRCh38
rs1057254937 Chr.19: 14148227-14148227 on Build GRCh38
rs554575970 Chr.19: 14148335-14148335 on Build GRCh38
rs889043527 Chr.19: 14148627-14148627 on Build GRCh38
rs1052182003 Chr.19: 14148323-14148323 on Build GRCh38
rs965400842 Chr.19: 14148248-14148248 on Build GRCh38
rs571526002 Chr.19: 14148212-14148212 on Build GRCh38
rs1004740534 Chr.19: 14148419-14148419 on Build GRCh38
rs547219141 Chr.19: 14148232-14148232 on Build GRCh38
rs148162456 Chr.19: 14148372-14148372 on Build GRCh38
rs1020073846 Chr.19: 14148246-14148246 on Build GRCh38
rs756524426 Chr.19: 14148252-14148252 on Build GRCh38
rs920813593 Chr.19: 14148151-14148151 on Build GRCh38
rs776501556 Chr.19: 14148540-14148540 on Build GRCh38
rs768052512 Chr.19: 14148319-14148319 on Build GRCh38
rs968900975 Chr.19: 14148514-14148514 on Build GRCh38
rs547876851 Chr.19: 14148269-14148269 on Build GRCh38
rs1027084597 Chr.19: 14148231-14148231 on Build GRCh38
rs780481970 Chr.19: 14148263-14148263 on Build GRCh38
rs965785001 Chr.19: 14148449-14148449 on Build GRCh38
rs1041513548 Chr.19: 14148548-14148548 on Build GRCh38
rs895521761 Chr.19: 14148528-14148528 on Build GRCh38

View Full Product Details