Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 41941261-41941665 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCND3
Entrez Gene ID
896
Gene Name
cyclin D3
Gene Aliases
-
UniGene
Hs.534307
Gene Chromosome Location
Chr.6: 41934933-42048894 on Build GRCh38
Transcripts
NM_001136017.3 NM_001136125.2 NM_001136126.2 NM_001287427.1 NM_001287434.1 NM_001760.4 XM_011514971.1 XM_011514972.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4997747 c.97C>T p.R33C Chr.6: 41941553-41941553 on Build GRCh38
1754810 c.150G>A p.R50R Chr.6: 41941500-41941500 on Build GRCh38
6513955 c.24C>T p.G8G Chr.6: 41941626-41941626 on Build GRCh38
4888062 c.109C>A p.R37S Chr.6: 41941541-41941541 on Build GRCh38
6495115 c.63G>T p.L21L Chr.6: 41941587-41941587 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755019224 Chr.6: 41941428-41941428 on Build GRCh38
rs368995905 Chr.6: 41941312-41941313 on Build GRCh38
rs1017720006 Chr.6: 41941350-41941350 on Build GRCh38
rs867977624 Chr.6: 41941573-41941573 on Build GRCh38
rs776692321 Chr.6: 41941475-41941475 on Build GRCh38
rs761079148 Chr.6: 41941532-41941532 on Build GRCh38
rs573762582 Chr.6: 41941388-41941388 on Build GRCh38
rs1044535473 Chr.6: 41941432-41941432 on Build GRCh38
rs941153314 Chr.6: 41941436-41941436 on Build GRCh38
rs553896566 Chr.6: 41941374-41941374 on Build GRCh38
rs993120161 Chr.6: 41941607-41941607 on Build GRCh38
rs756643151 Chr.6: 41941424-41941424 on Build GRCh38
rs189586831 Chr.6: 41941407-41941407 on Build GRCh38
rs764873715 Chr.6: 41941482-41941482 on Build GRCh38
rs534048771 Chr.6: 41941306-41941306 on Build GRCh38
rs577743238 Chr.6: 41941281-41941281 on Build GRCh38
rs752370290 Chr.6: 41941402-41941402 on Build GRCh38
rs998134479 Chr.6: 41941510-41941510 on Build GRCh38
rs749418010 Chr.6: 41941427-41941427 on Build GRCh38
rs556229606 Chr.6: 41941550-41941550 on Build GRCh38
rs746833245 Chr.6: 41941643-41941643 on Build GRCh38
rs746832372 Chr.6: 41941585-41941585 on Build GRCh38
rs758087276 Chr.6: 41941409-41941409 on Build GRCh38
rs768238448 Chr.6: 41941613-41941613 on Build GRCh38
rs375385226 Chr.6: 41941480-41941480 on Build GRCh38
rs957618987 Chr.6: 41941399-41941399 on Build GRCh38
rs371124796 Chr.6: 41941443-41941443 on Build GRCh38
rs766833972 Chr.6: 41941536-41941536 on Build GRCh38
rs775204715 Chr.6: 41941605-41941605 on Build GRCh38
rs150526537 Chr.6: 41941587-41941587 on Build GRCh38
rs1033224695 Chr.6: 41941396-41941396 on Build GRCh38
rs762507772 Chr.6: 41941610-41941610 on Build GRCh38
rs770832153 Chr.6: 41941446-41941446 on Build GRCh38
rs545054088 Chr.6: 41941620-41941620 on Build GRCh38
rs778922487 Chr.6: 41941435-41941435 on Build GRCh38
rs200538938 Chr.6: 41941415-41941415 on Build GRCh38
rs981945157 Chr.6: 41941431-41941431 on Build GRCh38
rs778042653 Chr.6: 41941584-41941584 on Build GRCh38
rs967655006 Chr.6: 41941336-41941336 on Build GRCh38
rs755426354 Chr.6: 41941552-41941552 on Build GRCh38
rs376321266 Chr.6: 41941499-41941499 on Build GRCh38
rs748389291 Chr.6: 41941439-41941439 on Build GRCh38
rs745825762 Chr.6: 41941599-41941599 on Build GRCh38
rs1006519914 Chr.6: 41941535-41941535 on Build GRCh38
rs969256609 Chr.6: 41941376-41941376 on Build GRCh38
rs572116433 Chr.6: 41941554-41941554 on Build GRCh38
rs924832888 Chr.6: 41941403-41941403 on Build GRCh38
rs545476294 Chr.6: 41941530-41941530 on Build GRCh38
rs917248590 Chr.6: 41941612-41941612 on Build GRCh38
rs1036879059 Chr.6: 41941461-41941461 on Build GRCh38
rs956471154 Chr.6: 41941594-41941594 on Build GRCh38
rs888904737 Chr.6: 41941623-41941623 on Build GRCh38
rs775094377 Chr.6: 41941488-41941488 on Build GRCh38
rs778979671 Chr.6: 41941553-41941553 on Build GRCh38
rs758615400 Chr.6: 41941581-41941581 on Build GRCh38
rs927809612 Chr.6: 41941442-41941442 on Build GRCh38
rs116713943 Chr.6: 41941627-41941627 on Build GRCh38
rs772889403 Chr.6: 41941445-41941445 on Build GRCh38
rs769785626 Chr.6: 41941601-41941601 on Build GRCh38
rs889483163 Chr.6: 41941518-41941518 on Build GRCh38
rs780592965 Chr.6: 41941426-41941426 on Build GRCh38

View Full Product Details