Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 80513878-80514447 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSPAN14
Entrez Gene ID
81619
Gene Name
tetraspanin 14
Gene Aliases
DC-TM4F2, TM4SF14
UniGene
Hs.718943
Gene Chromosome Location
Chr.10: 80454271-80522635 on Build GRCh38
Transcripts
NM_001128309.1 NM_030927.2 XM_006717992.3 XM_011540219.2 XM_011540220.2 XM_011540221.2 XM_011540222.2 XM_017016707.1 XM_017016708.1 XM_017016709.1 XM_017016710.1 XM_017016711.1 XM_017016712.1 XM_017016713.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3734278 c.491A>C p.K164T Chr.10: 80514023-80514023 on Build GRCh38
920814 c.605A>G p.Y202C Chr.10: 80514047-80514047 on Build GRCh38
3734277 c.581A>C p.K194T Chr.10: 80514023-80514023 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs991230200 Chr.10: 80514195-80514195 on Build GRCh38
rs949651816 Chr.10: 80514327-80514327 on Build GRCh38
rs889491601 Chr.10: 80513979-80513979 on Build GRCh38
rs1022142074 Chr.10: 80514324-80514324 on Build GRCh38
rs368734913 Chr.10: 80513998-80513998 on Build GRCh38
rs769639994 Chr.10: 80514307-80514307 on Build GRCh38
rs774735228 Chr.10: 80514095-80514095 on Build GRCh38
rs966417485 Chr.10: 80514351-80514351 on Build GRCh38
rs371144258 Chr.10: 80513983-80513983 on Build GRCh38
rs546869257 Chr.10: 80514240-80514240 on Build GRCh38
rs35574505 Chr.10: 80513947-80513948 on Build GRCh38
rs139086777 Chr.10: 80514012-80514012 on Build GRCh38
rs966469843 Chr.10: 80513925-80513925 on Build GRCh38
rs918135929 Chr.10: 80514312-80514312 on Build GRCh38
rs201688164 Chr.10: 80513984-80513984 on Build GRCh38
rs911503131 Chr.10: 80514076-80514076 on Build GRCh38
rs113545776 Chr.10: 80514228-80514228 on Build GRCh38
rs772948374 Chr.10: 80514379-80514379 on Build GRCh38
rs771781333 Chr.10: 80514103-80514103 on Build GRCh38
rs185519928 Chr.10: 80514113-80514113 on Build GRCh38
rs1032749426 Chr.10: 80514146-80514146 on Build GRCh38
rs554841349 Chr.10: 80514425-80514425 on Build GRCh38
rs1010722939 Chr.10: 80514290-80514290 on Build GRCh38
rs764125981 Chr.10: 80513997-80513997 on Build GRCh38
rs778357301 Chr.10: 80514075-80514075 on Build GRCh38
rs764105135 Chr.10: 80514390-80514391 on Build GRCh38
rs978123041 Chr.10: 80513953-80513953 on Build GRCh38
rs748288768 Chr.10: 80514047-80514047 on Build GRCh38
rs771420375 Chr.10: 80514094-80514094 on Build GRCh38
rs1048517000 Chr.10: 80513972-80513972 on Build GRCh38
rs918630080 Chr.10: 80514118-80514118 on Build GRCh38
rs985543235 Chr.10: 80514388-80514388 on Build GRCh38
rs999275591 Chr.10: 80514355-80514355 on Build GRCh38
rs781220549 Chr.10: 80514038-80514038 on Build GRCh38
rs760056536 Chr.10: 80513994-80513994 on Build GRCh38
rs778909750 Chr.10: 80514017-80514017 on Build GRCh38
rs931045261 Chr.10: 80514354-80514354 on Build GRCh38
rs1029866473 Chr.10: 80514373-80514373 on Build GRCh38
rs1055040841 Chr.10: 80514256-80514256 on Build GRCh38
rs756703418 Chr.10: 80513941-80513941 on Build GRCh38
rs1001962245 Chr.10: 80514202-80514202 on Build GRCh38
rs1010043449 Chr.10: 80514073-80514073 on Build GRCh38
rs780800949 Chr.10: 80513980-80513986 on Build GRCh38
rs150070619 Chr.10: 80514100-80514100 on Build GRCh38
rs979569085 Chr.10: 80514128-80514128 on Build GRCh38
rs758657157 Chr.10: 80514072-80514072 on Build GRCh38
rs918778932 Chr.10: 80514353-80514353 on Build GRCh38
rs755292347 Chr.10: 80514029-80514029 on Build GRCh38
rs931083887 Chr.10: 80513955-80513955 on Build GRCh38
rs780448743 Chr.10: 80514239-80514239 on Build GRCh38
rs750239953 Chr.10: 80514019-80514019 on Build GRCh38
rs1046266666 Chr.10: 80514138-80514138 on Build GRCh38
rs530537887 Chr.10: 80514050-80514050 on Build GRCh38
rs749848129 Chr.10: 80514080-80514080 on Build GRCh38
rs749235529 Chr.10: 80514395-80514395 on Build GRCh38
rs535878849 Chr.10: 80514149-80514149 on Build GRCh38
rs930068766 Chr.10: 80514132-80514132 on Build GRCh38
rs566657340 Chr.10: 80514268-80514268 on Build GRCh38
rs966763331 Chr.10: 80514107-80514107 on Build GRCh38
rs962934237 Chr.10: 80514415-80514415 on Build GRCh38
rs1048178402 Chr.10: 80514366-80514366 on Build GRCh38

View Full Product Details