Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 20652143-20652739 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDOST
Entrez Gene ID
1650
Gene Name
dolichyl-diphosphooligosaccharide--protein glycosyltransferase non-catalytic subunit
Gene Aliases
AGER1, CDG1R, OKSWcl45, OST, OST48, WBP1
UniGene
Hs.523145
Gene Chromosome Location
Chr.1: 20651767-20661544 on Build GRCh38
Transcripts
NM_005216.4 NR_046507.1

Gene Symbol
PINK1-AS
Entrez Gene ID
100861548
Gene Name
PINK1 antisense RNA
Gene Aliases
NAPINK1, PINK1-AS1, PINK1AS
UniGene
Hs.710259
Gene Chromosome Location
Chr.1: 20642657-20652193 on Build GRCh38
Transcripts
NM_005216.4 NR_046507.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5341863 c.1335C>T p.V445V Chr.1: 20652415-20652415 on Build GRCh38
6852973 c.1150G>A p.V384M Chr.1: 20652692-20652692 on Build GRCh38
1929081 c.1133A>G p.Q378R Chr.1: 20652709-20652709 on Build GRCh38
463873 c.1147G>A p.D383N Chr.1: 20652695-20652695 on Build GRCh38
1929060 c.1288G>A p.A430T Chr.1: 20652462-20652462 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781678887 Chr.1: 20652335-20652336 on Build GRCh38
rs770836120 Chr.1: 20652613-20652613 on Build GRCh38
rs886045864 Chr.1: 20652695-20652695 on Build GRCh38
rs770264207 Chr.1: 20652333-20652333 on Build GRCh38
rs748707299 Chr.1: 20652332-20652332 on Build GRCh38
rs748686232 Chr.1: 20652374-20652375 on Build GRCh38
rs587776874 Chr.1: 20652464-20652485 on Build GRCh38
rs761280123 Chr.1: 20652430-20652430 on Build GRCh38
rs369696003 Chr.1: 20652361-20652361 on Build GRCh38
rs761348979 Chr.1: 20652484-20652484 on Build GRCh38
rs757415954 Chr.1: 20652585-20652585 on Build GRCh38
rs1020375060 Chr.1: 20652262-20652262 on Build GRCh38
rs768487044 Chr.1: 20652414-20652414 on Build GRCh38
rs138277251 Chr.1: 20652269-20652269 on Build GRCh38
rs767736586 Chr.1: 20652519-20652519 on Build GRCh38
rs931915308 Chr.1: 20652457-20652457 on Build GRCh38
rs749626636 Chr.1: 20652397-20652397 on Build GRCh38
rs763273068 Chr.1: 20652334-20652334 on Build GRCh38
rs142441124 Chr.1: 20652478-20652478 on Build GRCh38
rs748246542 Chr.1: 20652368-20652368 on Build GRCh38
rs764414378 Chr.1: 20652636-20652636 on Build GRCh38
rs1032130396 Chr.1: 20652327-20652327 on Build GRCh38
rs370226725 Chr.1: 20652394-20652394 on Build GRCh38
rs753945605 Chr.1: 20652642-20652642 on Build GRCh38
rs749823019 Chr.1: 20652473-20652473 on Build GRCh38
rs1051884 Chr.1: 20652193-20652193 on Build GRCh38
rs1011294204 Chr.1: 20652257-20652257 on Build GRCh38
rs759303850 Chr.1: 20652619-20652619 on Build GRCh38
rs200636246 Chr.1: 20652522-20652522 on Build GRCh38
rs901496245 Chr.1: 20652175-20652175 on Build GRCh38
rs117634838 Chr.1: 20652406-20652406 on Build GRCh38
rs764066661 Chr.1: 20652526-20652526 on Build GRCh38
rs771423836 Chr.1: 20652399-20652399 on Build GRCh38
rs530990753 Chr.1: 20652503-20652503 on Build GRCh38
rs376894317 Chr.1: 20652463-20652463 on Build GRCh38
rs757807492 Chr.1: 20652545-20652545 on Build GRCh38
rs770477094 Chr.1: 20652378-20652379 on Build GRCh38
rs746272803 Chr.1: 20652479-20652479 on Build GRCh38
rs762670507 Chr.1: 20652446-20652446 on Build GRCh38
rs767848609 Chr.1: 20652428-20652428 on Build GRCh38
rs149576051 Chr.1: 20652523-20652523 on Build GRCh38
rs781312902 Chr.1: 20652456-20652456 on Build GRCh38
rs566726706 Chr.1: 20652604-20652605 on Build GRCh38
rs754996957 Chr.1: 20652359-20652359 on Build GRCh38
rs745516952 Chr.1: 20652533-20652535 on Build GRCh38
rs776939873 Chr.1: 20652331-20652331 on Build GRCh38
rs1047320919 Chr.1: 20652246-20652246 on Build GRCh38
rs899463821 Chr.1: 20652290-20652290 on Build GRCh38
rs923235061 Chr.1: 20652418-20652418 on Build GRCh38
rs755694319 Chr.1: 20652690-20652690 on Build GRCh38
rs769550415 Chr.1: 20652588-20652588 on Build GRCh38
rs1019291114 Chr.1: 20652272-20652272 on Build GRCh38
rs6893 Chr.1: 20652339-20652339 on Build GRCh38
rs779611800 Chr.1: 20652551-20652551 on Build GRCh38
rs750359512 Chr.1: 20652442-20652442 on Build GRCh38
rs756177600 Chr.1: 20652462-20652462 on Build GRCh38
rs370808251 Chr.1: 20652439-20652439 on Build GRCh38
rs1051804 Chr.1: 20652292-20652292 on Build GRCh38
rs537385227 Chr.1: 20652267-20652267 on Build GRCh38
rs751328230 Chr.1: 20652517-20652517 on Build GRCh38
rs561443647 Chr.1: 20652552-20652552 on Build GRCh38
rs760192704 Chr.1: 20652337-20652337 on Build GRCh38
rs527983100 Chr.1: 20652280-20652281 on Build GRCh38
rs758721537 Chr.1: 20652568-20652568 on Build GRCh38
rs569862570 Chr.1: 20652247-20652247 on Build GRCh38
rs1036335973 Chr.1: 20652576-20652576 on Build GRCh38
rs148883998 Chr.1: 20652490-20652490 on Build GRCh38
rs771896096 Chr.1: 20652541-20652542 on Build GRCh38
rs747769976 Chr.1: 20652575-20652575 on Build GRCh38
rs754036321 Chr.1: 20652528-20652528 on Build GRCh38
rs374032054 Chr.1: 20652425-20652425 on Build GRCh38
rs772707199 Chr.1: 20652600-20652600 on Build GRCh38
rs996505869 Chr.1: 20652296-20652296 on Build GRCh38
rs765762722 Chr.1: 20652210-20652210 on Build GRCh38
rs1004299514 Chr.1: 20652714-20652714 on Build GRCh38
rs1009708478 Chr.1: 20652634-20652634 on Build GRCh38
rs752770102 Chr.1: 20652459-20652459 on Build GRCh38
rs774300302 Chr.1: 20652616-20652616 on Build GRCh38
rs747477003 Chr.1: 20652672-20652672 on Build GRCh38
rs145940009 Chr.1: 20652643-20652643 on Build GRCh38
rs746186938 Chr.1: 20652559-20652559 on Build GRCh38
rs775410757 Chr.1: 20652631-20652631 on Build GRCh38
rs748840954 Chr.1: 20652606-20652606 on Build GRCh38
rs750211906 Chr.1: 20652353-20652353 on Build GRCh38
rs931204168 Chr.1: 20652213-20652213 on Build GRCh38
rs751418274 Chr.1: 20652450-20652450 on Build GRCh38
rs776171831 Chr.1: 20652483-20652483 on Build GRCh38
rs760390881 Chr.1: 20652412-20652412 on Build GRCh38
rs898568954 Chr.1: 20652180-20652180 on Build GRCh38
rs138561924 Chr.1: 20652656-20652656 on Build GRCh38
rs775911987 Chr.1: 20652323-20652323 on Build GRCh38
rs769192887 Chr.1: 20652489-20652489 on Build GRCh38
rs758213706 Chr.1: 20652354-20652354 on Build GRCh38
rs201469687 Chr.1: 20652345-20652345 on Build GRCh38
rs892861868 Chr.1: 20652230-20652230 on Build GRCh38
rs770428927 Chr.1: 20652709-20652709 on Build GRCh38
rs764406890 Chr.1: 20652385-20652385 on Build GRCh38
rs777455786 Chr.1: 20652693-20652693 on Build GRCh38
rs893569505 Chr.1: 20652533-20652533 on Build GRCh38
rs1003533489 Chr.1: 20652278-20652278 on Build GRCh38
rs766177749 Chr.1: 20652447-20652447 on Build GRCh38
rs767226385 Chr.1: 20652625-20652625 on Build GRCh38
rs767227234 Chr.1: 20652335-20652335 on Build GRCh38
rs780832225 Chr.1: 20652572-20652572 on Build GRCh38
rs931693537 Chr.1: 20652176-20652176 on Build GRCh38
rs780174398 Chr.1: 20652397-20652399 on Build GRCh38
rs771622498 Chr.1: 20652475-20652475 on Build GRCh38
rs1020454817 Chr.1: 20652638-20652638 on Build GRCh38
rs762867601 Chr.1: 20652500-20652500 on Build GRCh38
rs780209777 Chr.1: 20652356-20652356 on Build GRCh38
rs74526704 Chr.1: 20652655-20652655 on Build GRCh38
rs768756395 Chr.1: 20652330-20652330 on Build GRCh38
rs544661380 Chr.1: 20652417-20652417 on Build GRCh38
rs780269585 Chr.1: 20652669-20652669 on Build GRCh38
rs369025294 Chr.1: 20652481-20652481 on Build GRCh38
rs755436581 Chr.1: 20652329-20652330 on Build GRCh38
rs373329310 Chr.1: 20652518-20652518 on Build GRCh38
rs894703465 Chr.1: 20652578-20652578 on Build GRCh38
rs1051670 Chr.1: 20652451-20652451 on Build GRCh38
rs139861731 Chr.1: 20652696-20652696 on Build GRCh38
rs375122135 Chr.1: 20652346-20652346 on Build GRCh38

View Full Product Details