Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27325686-27326285 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF3C2
Entrez Gene ID
2976
Gene Name
general transcription factor IIIC subunit 2
Gene Aliases
TFIIIC-BETA, TFIIIC110
UniGene
Hs.745168
Gene Chromosome Location
Chr.2: 27325849-27357034 on Build GRCh38
Transcripts
NM_001035521.2 NM_001318909.1 NM_001521.3 XM_005264272.4 XM_011532801.2

SNP Detail

SNP ID Chromosome Location
rs1045997381 Chr.2: 27325990-27325990 on Build GRCh38
rs183874653 Chr.2: 27326238-27326238 on Build GRCh38
rs931276780 Chr.2: 27326139-27326139 on Build GRCh38
rs893169835 Chr.2: 27326081-27326081 on Build GRCh38
rs191286683 Chr.2: 27326171-27326171 on Build GRCh38
rs779386890 Chr.2: 27326195-27326195 on Build GRCh38
rs926982229 Chr.2: 27325957-27325957 on Build GRCh38
rs1008344437 Chr.2: 27325724-27325724 on Build GRCh38
rs541605351 Chr.2: 27326121-27326121 on Build GRCh38
rs746108276 Chr.2: 27326210-27326210 on Build GRCh38
rs968769160 Chr.2: 27325897-27325897 on Build GRCh38
rs1028253193 Chr.2: 27326109-27326109 on Build GRCh38
rs751814241 Chr.2: 27326098-27326098 on Build GRCh38
rs957425058 Chr.2: 27325723-27325723 on Build GRCh38
rs186706145 Chr.2: 27326053-27326053 on Build GRCh38
rs1049736662 Chr.2: 27326142-27326142 on Build GRCh38
rs1037981367 Chr.2: 27326106-27326106 on Build GRCh38
rs767312163 Chr.2: 27326247-27326248 on Build GRCh38
rs959764933 Chr.2: 27326036-27326036 on Build GRCh38
rs571125203 Chr.2: 27325804-27325806 on Build GRCh38
rs370980814 Chr.2: 27325806-27325806 on Build GRCh38
rs979807839 Chr.2: 27325951-27325951 on Build GRCh38
rs371789691 Chr.2: 27326097-27326097 on Build GRCh38
rs867771982 Chr.2: 27326188-27326188 on Build GRCh38
rs747664480 Chr.2: 27326026-27326027 on Build GRCh38
rs972549549 Chr.2: 27325859-27325859 on Build GRCh38
rs1053215159 Chr.2: 27326069-27326069 on Build GRCh38
rs920541537 Chr.2: 27326120-27326120 on Build GRCh38
rs894742977 Chr.2: 27325921-27325921 on Build GRCh38
rs1010459373 Chr.2: 27325831-27325831 on Build GRCh38
rs543486486 Chr.2: 27325812-27325812 on Build GRCh38
rs760410910 Chr.2: 27326211-27326211 on Build GRCh38
rs889889704 Chr.2: 27325718-27325718 on Build GRCh38
rs939831703 Chr.2: 27325854-27325854 on Build GRCh38
rs749346507 Chr.2: 27326158-27326158 on Build GRCh38
rs778187398 Chr.2: 27326051-27326051 on Build GRCh38
rs986611621 Chr.2: 27325839-27325839 on Build GRCh38
rs940534316 Chr.2: 27326105-27326105 on Build GRCh38
rs1022267892 Chr.2: 27326221-27326221 on Build GRCh38
rs990520538 Chr.2: 27325741-27325741 on Build GRCh38
rs919663371 Chr.2: 27325881-27325881 on Build GRCh38
rs759042573 Chr.2: 27325778-27325778 on Build GRCh38
rs780182474 Chr.2: 27325883-27325883 on Build GRCh38
rs953942696 Chr.2: 27325813-27325813 on Build GRCh38
rs1010428211 Chr.2: 27326217-27326217 on Build GRCh38
rs781230694 Chr.2: 27326205-27326205 on Build GRCh38
rs757644805 Chr.2: 27326204-27326204 on Build GRCh38
rs1021881505 Chr.2: 27325864-27325864 on Build GRCh38
rs746292008 Chr.2: 27326197-27326197 on Build GRCh38
rs1054770950 Chr.2: 27325918-27325922 on Build GRCh38
rs1002219001 Chr.2: 27326261-27326261 on Build GRCh38
rs954034863 Chr.2: 27326164-27326164 on Build GRCh38
rs547906023 Chr.2: 27326242-27326242 on Build GRCh38

View Full Product Details