Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 206480819-206481359 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM23
Entrez Gene ID
8745
Gene Name
ADAM metallopeptidase domain 23
Gene Aliases
MDC-3, MDC3
UniGene
Hs.370287
Gene Chromosome Location
Chr.2: 206443544-206621130 on Build GRCh38
Transcripts
NM_003812.3 XM_005246932.3 XM_011512086.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4932393 c.509+2T>C p.? Chr.2: 206481310-206481310 on Build GRCh38
238681 c.463A>G p.I155V Chr.2: 206481262-206481262 on Build GRCh38
3425992 c.490C>A p.L164I Chr.2: 206481289-206481289 on Build GRCh38
7145934 c.474C>T p.F158F Chr.2: 206481273-206481273 on Build GRCh38
3576796 c.471C>T p.A157A Chr.2: 206481270-206481270 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs994471276 Chr.2: 206481124-206481124 on Build GRCh38
rs35864832 Chr.2: 206481004-206481005 on Build GRCh38
rs770179584 Chr.2: 206481327-206481329 on Build GRCh38
rs748026373 Chr.2: 206481304-206481304 on Build GRCh38
rs373661314 Chr.2: 206481294-206481294 on Build GRCh38
rs776509677 Chr.2: 206481249-206481249 on Build GRCh38
rs202037149 Chr.2: 206481270-206481270 on Build GRCh38
rs994053513 Chr.2: 206480906-206480906 on Build GRCh38
rs369357303 Chr.2: 206481010-206481010 on Build GRCh38
rs753145912 Chr.2: 206481269-206481269 on Build GRCh38
rs963710865 Chr.2: 206480974-206480974 on Build GRCh38
rs938964298 Chr.2: 206481126-206481126 on Build GRCh38
rs977625560 Chr.2: 206481116-206481116 on Build GRCh38
rs372881378 Chr.2: 206481041-206481041 on Build GRCh38
rs755607321 Chr.2: 206481196-206481196 on Build GRCh38
rs1001945194 Chr.2: 206481026-206481026 on Build GRCh38
rs760898127 Chr.2: 206481239-206481239 on Build GRCh38
rs955323127 Chr.2: 206480917-206480917 on Build GRCh38
rs921298430 Chr.2: 206480885-206480885 on Build GRCh38
rs750001691 Chr.2: 206481195-206481195 on Build GRCh38
rs1009867047 Chr.2: 206480947-206480947 on Build GRCh38
rs778688136 Chr.2: 206481302-206481302 on Build GRCh38
rs765613816 Chr.2: 206481261-206481261 on Build GRCh38
rs144108353 Chr.2: 206481299-206481299 on Build GRCh38
rs936652172 Chr.2: 206480892-206480892 on Build GRCh38
rs906417147 Chr.2: 206481334-206481334 on Build GRCh38
rs754988103 Chr.2: 206481301-206481301 on Build GRCh38
rs148171905 Chr.2: 206481233-206481233 on Build GRCh38
rs942031814 Chr.2: 206481168-206481168 on Build GRCh38
rs760194188 Chr.2: 206480954-206480954 on Build GRCh38
rs779643871 Chr.2: 206481297-206481297 on Build GRCh38
rs75293874 Chr.2: 206480946-206480946 on Build GRCh38
rs3213811 Chr.2: 206481102-206481102 on Build GRCh38
rs187764512 Chr.2: 206481107-206481107 on Build GRCh38
rs749249872 Chr.2: 206481208-206481208 on Build GRCh38
rs779371187 Chr.2: 206481206-206481206 on Build GRCh38
rs111649087 Chr.2: 206481290-206481290 on Build GRCh38
rs772764731 Chr.2: 206480949-206480949 on Build GRCh38
rs771059445 Chr.2: 206481244-206481244 on Build GRCh38
rs951680586 Chr.2: 206481175-206481175 on Build GRCh38
rs747925680 Chr.2: 206481228-206481228 on Build GRCh38
rs989841071 Chr.2: 206480977-206480977 on Build GRCh38
rs986851152 Chr.2: 206480945-206480945 on Build GRCh38
rs906347815 Chr.2: 206481004-206481004 on Build GRCh38
rs561936375 Chr.2: 206480849-206480849 on Build GRCh38
rs983420799 Chr.2: 206481176-206481176 on Build GRCh38
rs778788344 Chr.2: 206481224-206481224 on Build GRCh38
rs1053746147 Chr.2: 206480897-206480897 on Build GRCh38
rs577379429 Chr.2: 206480909-206480909 on Build GRCh38
rs533597701 Chr.2: 206481070-206481070 on Build GRCh38
rs914362269 Chr.2: 206480986-206480986 on Build GRCh38
rs146209053 Chr.2: 206481274-206481274 on Build GRCh38
rs780791614 Chr.2: 206481189-206481189 on Build GRCh38
rs545524157 Chr.2: 206480844-206480844 on Build GRCh38
rs1055774311 Chr.2: 206481149-206481149 on Build GRCh38
rs898302476 Chr.2: 206480899-206480899 on Build GRCh38
rs141683172 Chr.2: 206481273-206481273 on Build GRCh38
rs768434765 Chr.2: 206481209-206481209 on Build GRCh38
rs541179329 Chr.2: 206480922-206480922 on Build GRCh38
rs758257170 Chr.2: 206481323-206481323 on Build GRCh38
rs760122536 Chr.2: 206481255-206481255 on Build GRCh38
rs1033388538 Chr.2: 206481027-206481027 on Build GRCh38
rs542908957 Chr.2: 206481150-206481150 on Build GRCh38
rs1046252874 Chr.2: 206480963-206480963 on Build GRCh38
rs141978401 Chr.2: 206481238-206481238 on Build GRCh38

View Full Product Details