Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 170433466-170434046 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN11
Entrez Gene ID
5010
Gene Name
claudin 11
Gene Aliases
OSP, OTM
UniGene
Hs.31595
Gene Chromosome Location
Chr.3: 170418865-170434691 on Build GRCh38
Transcripts
NM_001185056.1 NM_005602.5

SNP Detail

SNP ID Chromosome Location
rs114923460 Chr.3: 170433694-170433694 on Build GRCh38
rs1041177105 Chr.3: 170433702-170433702 on Build GRCh38
rs956598275 Chr.3: 170433841-170433841 on Build GRCh38
rs969244665 Chr.3: 170433940-170433940 on Build GRCh38
rs553149835 Chr.3: 170433984-170433984 on Build GRCh38
rs141295233 Chr.3: 170433886-170433886 on Build GRCh38
rs766133981 Chr.3: 170433764-170433764 on Build GRCh38
rs1026177425 Chr.3: 170433526-170433526 on Build GRCh38
rs931314389 Chr.3: 170433612-170433612 on Build GRCh38
rs534914207 Chr.3: 170433983-170433983 on Build GRCh38
rs1028652732 Chr.3: 170433520-170433520 on Build GRCh38
rs920008785 Chr.3: 170433981-170433981 on Build GRCh38
rs146584959 Chr.3: 170433546-170433546 on Build GRCh38
rs917787743 Chr.3: 170433571-170433571 on Build GRCh38
rs71306669 Chr.3: 170433533-170433533 on Build GRCh38
rs945676783 Chr.3: 170433592-170433592 on Build GRCh38
rs934242701 Chr.3: 170433984-170433984 on Build GRCh38
rs992007914 Chr.3: 170433564-170433564 on Build GRCh38
rs13973 Chr.3: 170433903-170433903 on Build GRCh38
rs926365992 Chr.3: 170433561-170433561 on Build GRCh38
rs979337129 Chr.3: 170433540-170433540 on Build GRCh38
rs116573207 Chr.3: 170433670-170433670 on Build GRCh38
rs143257986 Chr.3: 170433957-170433957 on Build GRCh38
rs76935838 Chr.3: 170433760-170433760 on Build GRCh38
rs761111067 Chr.3: 170433925-170433925 on Build GRCh38
rs944290413 Chr.3: 170433996-170433996 on Build GRCh38
rs1043013711 Chr.3: 170433603-170433603 on Build GRCh38
rs761890708 Chr.3: 170433585-170433585 on Build GRCh38
rs772652873 Chr.3: 170433588-170433588 on Build GRCh38
rs567602707 Chr.3: 170433962-170433962 on Build GRCh38
rs1026247280 Chr.3: 170433788-170433788 on Build GRCh38
rs934170587 Chr.3: 170433640-170433641 on Build GRCh38
rs77614151 Chr.3: 170433562-170433562 on Build GRCh38
rs922860406 Chr.3: 170433954-170433954 on Build GRCh38
rs548502969 Chr.3: 170433870-170433870 on Build GRCh38
rs908563380 Chr.3: 170433995-170433995 on Build GRCh38
rs1045212397 Chr.3: 170433593-170433593 on Build GRCh38
rs967698655 Chr.3: 170433782-170433782 on Build GRCh38
rs1047272975 Chr.3: 170433637-170433637 on Build GRCh38
rs900050578 Chr.3: 170433708-170433708 on Build GRCh38
rs1030891412 Chr.3: 170433836-170433836 on Build GRCh38
rs561756412 Chr.3: 170433527-170433527 on Build GRCh38
rs917756631 Chr.3: 170433904-170433904 on Build GRCh38
rs184479005 Chr.3: 170433720-170433720 on Build GRCh38
rs989621746 Chr.3: 170433855-170433855 on Build GRCh38
rs79247818 Chr.3: 170433987-170433987 on Build GRCh38
rs981942344 Chr.3: 170433797-170433797 on Build GRCh38
rs919931450 Chr.3: 170433595-170433595 on Build GRCh38
rs1041312497 Chr.3: 170434011-170434011 on Build GRCh38
rs914935182 Chr.3: 170433856-170433856 on Build GRCh38
rs945053968 Chr.3: 170433868-170433868 on Build GRCh38
rs767563573 Chr.3: 170433662-170433662 on Build GRCh38
rs761368769 Chr.3: 170433808-170433808 on Build GRCh38

View Full Product Details