Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 73557439-73557718 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRA2
Entrez Gene ID
57763
Gene Name
ankyrin repeat family A member 2
Gene Aliases
ANKRA
UniGene
Hs.239154
Gene Chromosome Location
Chr.5: 73552200-73565705 on Build GRCh38
Transcripts
NM_023039.4 XM_005248560.2 XM_017009678.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7425992 c.449-2A>T p.? Chr.5: 73557642-73557642 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1050560854 Chr.5: 73557646-73557646 on Build GRCh38
rs368575233 Chr.5: 73557623-73557623 on Build GRCh38
rs747312994 Chr.5: 73557587-73557587 on Build GRCh38
rs752090959 Chr.5: 73557669-73557669 on Build GRCh38
rs778296728 Chr.5: 73557586-73557586 on Build GRCh38
rs1640 Chr.5: 73557489-73557489 on Build GRCh38
rs764825624 Chr.5: 73557662-73557662 on Build GRCh38
rs908773426 Chr.5: 73557528-73557528 on Build GRCh38
rs371577319 Chr.5: 73557600-73557600 on Build GRCh38
rs369766687 Chr.5: 73557650-73557650 on Build GRCh38
rs753402607 Chr.5: 73557537-73557537 on Build GRCh38
rs946571327 Chr.5: 73557530-73557530 on Build GRCh38
rs758076210 Chr.5: 73557684-73557684 on Build GRCh38
rs185418308 Chr.5: 73557484-73557484 on Build GRCh38
rs763837067 Chr.5: 73557687-73557687 on Build GRCh38
rs985819858 Chr.5: 73557521-73557521 on Build GRCh38
rs113255787 Chr.5: 73557602-73557602 on Build GRCh38
rs953371895 Chr.5: 73557515-73557515 on Build GRCh38
rs200156523 Chr.5: 73557589-73557589 on Build GRCh38
rs781762042 Chr.5: 73557590-73557590 on Build GRCh38
rs769890640 Chr.5: 73557601-73557601 on Build GRCh38
rs773055704 Chr.5: 73557621-73557621 on Build GRCh38
rs374915261 Chr.5: 73557607-73557607 on Build GRCh38
rs189925198 Chr.5: 73557539-73557539 on Build GRCh38
rs143788121 Chr.5: 73557582-73557582 on Build GRCh38
rs766351457 Chr.5: 73557649-73557649 on Build GRCh38
rs766298034 Chr.5: 73557529-73557529 on Build GRCh38
rs768797428 Chr.5: 73557619-73557619 on Build GRCh38
rs538617300 Chr.5: 73557695-73557695 on Build GRCh38
rs971780229 Chr.5: 73557485-73557485 on Build GRCh38
rs752426421 Chr.5: 73557581-73557581 on Build GRCh38
rs548602540 Chr.5: 73557505-73557509 on Build GRCh38
rs759351273 Chr.5: 73557655-73557655 on Build GRCh38
rs764837281 Chr.5: 73557568-73557568 on Build GRCh38
rs911123488 Chr.5: 73557525-73557525 on Build GRCh38

View Full Product Details