Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 130289314-130289869 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPA2
Entrez Gene ID
1358
Gene Name
carboxypeptidase A2
Gene Aliases
-
UniGene
Hs.490038
Gene Chromosome Location
Chr.7: 130266863-130289798 on Build GRCh38
Transcripts
NM_001869.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6634392 c.1157G>A p.R386H Chr.7: 130289644-130289644 on Build GRCh38
3922697 c.1252C>T p.P418S Chr.7: 130289739-130289739 on Build GRCh38
7362432 c.1150A>T p.T384S Chr.7: 130289637-130289637 on Build GRCh38
3928824 c.1237C>G p.H413D Chr.7: 130289724-130289724 on Build GRCh38
79688 c.1194C>G p.P398P Chr.7: 130289681-130289681 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs977559175 Chr.7: 130289643-130289643 on Build GRCh38
rs777021718 Chr.7: 130289551-130289551 on Build GRCh38
rs991377984 Chr.7: 130289593-130289593 on Build GRCh38
rs781355624 Chr.7: 130289576-130289576 on Build GRCh38
rs73146781 Chr.7: 130289833-130289833 on Build GRCh38
rs6953189 Chr.7: 130289809-130289809 on Build GRCh38
rs377343685 Chr.7: 130289525-130289525 on Build GRCh38
rs750787614 Chr.7: 130289667-130289667 on Build GRCh38
rs927882333 Chr.7: 130289844-130289847 on Build GRCh38
rs373858339 Chr.7: 130289764-130289764 on Build GRCh38
rs146602328 Chr.7: 130289644-130289644 on Build GRCh38
rs143727692 Chr.7: 130289738-130289738 on Build GRCh38
rs895167889 Chr.7: 130289828-130289828 on Build GRCh38
rs574215245 Chr.7: 130289668-130289668 on Build GRCh38
rs1054861098 Chr.7: 130289609-130289609 on Build GRCh38
rs766782512 Chr.7: 130289758-130289758 on Build GRCh38
rs199532190 Chr.7: 130289638-130289638 on Build GRCh38
rs914383202 Chr.7: 130289596-130289596 on Build GRCh38
rs544571490 Chr.7: 130289746-130289746 on Build GRCh38
rs562556003 Chr.7: 130289715-130289715 on Build GRCh38
rs1041309165 Chr.7: 130289357-130289357 on Build GRCh38
rs199917253 Chr.7: 130289552-130289552 on Build GRCh38
rs942017589 Chr.7: 130289506-130289506 on Build GRCh38
rs973551301 Chr.7: 130289808-130289810 on Build GRCh38
rs943745742 Chr.7: 130289345-130289345 on Build GRCh38
rs201758735 Chr.7: 130289553-130289553 on Build GRCh38
rs535576967 Chr.7: 130289507-130289507 on Build GRCh38
rs767992647 Chr.7: 130289666-130289666 on Build GRCh38
rs888642423 Chr.7: 130289454-130289454 on Build GRCh38
rs141058490 Chr.7: 130289648-130289648 on Build GRCh38
rs923437423 Chr.7: 130289435-130289435 on Build GRCh38
rs1004419667 Chr.7: 130289730-130289730 on Build GRCh38
rs774291988 Chr.7: 130289740-130289740 on Build GRCh38
rs778605354 Chr.7: 130289610-130289610 on Build GRCh38
rs372211619 Chr.7: 130289585-130289585 on Build GRCh38
rs770771335 Chr.7: 130289621-130289621 on Build GRCh38
rs752122123 Chr.7: 130289654-130289654 on Build GRCh38
rs763801452 Chr.7: 130289843-130289843 on Build GRCh38
rs752355949 Chr.7: 130289687-130289687 on Build GRCh38
rs772929768 Chr.7: 130289521-130289521 on Build GRCh38
rs200847245 Chr.7: 130289645-130289645 on Build GRCh38
rs148949036 Chr.7: 130289704-130289704 on Build GRCh38
rs772014284 Chr.7: 130289519-130289519 on Build GRCh38
rs757928983 Chr.7: 130289688-130289688 on Build GRCh38
rs751181451 Chr.7: 130289844-130289844 on Build GRCh38
rs368767764 Chr.7: 130289573-130289573 on Build GRCh38
rs768116789 Chr.7: 130289744-130289744 on Build GRCh38
rs906332379 Chr.7: 130289588-130289588 on Build GRCh38
rs747023544 Chr.7: 130289703-130289703 on Build GRCh38
rs143283211 Chr.7: 130289458-130289458 on Build GRCh38
rs766824299 Chr.7: 130289476-130289476 on Build GRCh38
rs754673870 Chr.7: 130289607-130289607 on Build GRCh38
rs997110938 Chr.7: 130289399-130289399 on Build GRCh38
rs1048566848 Chr.7: 130289438-130289438 on Build GRCh38
rs954877191 Chr.7: 130289436-130289436 on Build GRCh38
rs781002064 Chr.7: 130289711-130289711 on Build GRCh38
rs200603173 Chr.7: 130289616-130289616 on Build GRCh38
rs745689460 Chr.7: 130289712-130289712 on Build GRCh38
rs746852490 Chr.7: 130289617-130289617 on Build GRCh38
rs565287924 Chr.7: 130289762-130289762 on Build GRCh38
rs373156748 Chr.7: 130289842-130289842 on Build GRCh38
rs888602408 Chr.7: 130289693-130289693 on Build GRCh38
rs764655652 Chr.7: 130289707-130289707 on Build GRCh38
rs761192573 Chr.7: 130289382-130289382 on Build GRCh38
rs377236326 Chr.7: 130289731-130289731 on Build GRCh38
rs773806722 Chr.7: 130289745-130289745 on Build GRCh38
rs370431439 Chr.7: 130289743-130289743 on Build GRCh38
rs192814266 Chr.7: 130289546-130289546 on Build GRCh38
rs550369907 Chr.7: 130289649-130289649 on Build GRCh38
rs1033610861 Chr.7: 130289814-130289814 on Build GRCh38
rs375275902 Chr.7: 130289613-130289613 on Build GRCh38
rs769541759 Chr.7: 130289714-130289714 on Build GRCh38
rs762216172 Chr.7: 130289656-130289656 on Build GRCh38
rs751972467 Chr.7: 130289571-130289571 on Build GRCh38
rs757971457 Chr.7: 130289769-130289769 on Build GRCh38
rs374830405 Chr.7: 130289547-130289547 on Build GRCh38
rs548752707 Chr.7: 130289364-130289364 on Build GRCh38
rs750998057 Chr.7: 130289772-130289772 on Build GRCh38
rs937842728 Chr.7: 130289594-130289594 on Build GRCh38
rs199962603 Chr.7: 130289701-130289701 on Build GRCh38
rs146796996 Chr.7: 130289628-130289628 on Build GRCh38
rs373702907 Chr.7: 130289526-130289526 on Build GRCh38
rs778729879 Chr.7: 130289681-130289681 on Build GRCh38
rs756746465 Chr.7: 130289782-130289782 on Build GRCh38
rs763723161 Chr.7: 130289771-130289771 on Build GRCh38

View Full Product Details