Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 109769383-109769907 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM222A
Entrez Gene ID
84915
Gene Name
family with sequence similarity 222 member A
Gene Aliases
C12orf34
UniGene
Hs.661785
Gene Chromosome Location
Chr.12: 109714382-109770509 on Build GRCh38
Transcripts
NM_032829.2 XM_006719654.2 XM_017020055.1 NR_026661.2 NR_026662.2

Gene Symbol
FAM222A-AS1
Entrez Gene ID
84983
Gene Name
FAM222A antisense RNA 1
Gene Aliases
-
UniGene
Hs.675961
Gene Chromosome Location
Chr.12: 109734209-109773487 on Build GRCh38
Transcripts
NM_032829.2 XM_006719654.2 XM_017020055.1 NR_026661.2 NR_026662.2

SNP Detail

SNP ID Chromosome Location
rs548519526 Chr.12: 109769549-109769549 on Build GRCh38
rs1056091620 Chr.12: 109769574-109769575 on Build GRCh38
rs780493373 Chr.12: 109769618-109769618 on Build GRCh38
rs988336757 Chr.12: 109769493-109769493 on Build GRCh38
rs1014951809 Chr.12: 109769412-109769412 on Build GRCh38
rs1036194523 Chr.12: 109769595-109769595 on Build GRCh38
rs1012905534 Chr.12: 109769415-109769415 on Build GRCh38
rs1049664797 Chr.12: 109769807-109769807 on Build GRCh38
rs918312313 Chr.12: 109769778-109769778 on Build GRCh38
rs887818611 Chr.12: 109769408-109769408 on Build GRCh38
rs958310324 Chr.12: 109769716-109769716 on Build GRCh38
rs773097066 Chr.12: 109769626-109769627 on Build GRCh38
rs751004558 Chr.12: 109769494-109769494 on Build GRCh38
rs868817586 Chr.12: 109769450-109769450 on Build GRCh38
rs887731021 Chr.12: 109769580-109769580 on Build GRCh38
rs1028314358 Chr.12: 109769404-109769404 on Build GRCh38
rs1045509096 Chr.12: 109769539-109769539 on Build GRCh38
rs991206614 Chr.12: 109769802-109769802 on Build GRCh38
rs753711417 Chr.12: 109769788-109769788 on Build GRCh38
rs981702619 Chr.12: 109769434-109769434 on Build GRCh38
rs868844528 Chr.12: 109769767-109769767 on Build GRCh38
rs1011808410 Chr.12: 109769713-109769713 on Build GRCh38
rs375939363 Chr.12: 109769443-109769443 on Build GRCh38
rs185302215 Chr.12: 109769782-109769782 on Build GRCh38
rs943766532 Chr.12: 109769481-109769481 on Build GRCh38
rs1023085981 Chr.12: 109769428-109769428 on Build GRCh38
rs749687676 Chr.12: 109769664-109769664 on Build GRCh38
rs985078030 Chr.12: 109769469-109769469 on Build GRCh38
rs929462713 Chr.12: 109769537-109769537 on Build GRCh38
rs912977170 Chr.12: 109769495-109769495 on Build GRCh38
rs889092669 Chr.12: 109769665-109769665 on Build GRCh38
rs888015435 Chr.12: 109769625-109769625 on Build GRCh38
rs912191100 Chr.12: 109769756-109769756 on Build GRCh38
rs970891473 Chr.12: 109769764-109769764 on Build GRCh38
rs907287436 Chr.12: 109769677-109769677 on Build GRCh38
rs953936258 Chr.12: 109769726-109769726 on Build GRCh38
rs550022525 Chr.12: 109769768-109769768 on Build GRCh38
rs990115381 Chr.12: 109769456-109769456 on Build GRCh38
rs71458305 Chr.12: 109769583-109769583 on Build GRCh38
rs953736037 Chr.12: 109769459-109769459 on Build GRCh38
rs1021526809 Chr.12: 109769718-109769718 on Build GRCh38
rs997522926 Chr.12: 109769637-109769637 on Build GRCh38
rs369362505 Chr.12: 109769458-109769458 on Build GRCh38
rs1032167613 Chr.12: 109769694-109769694 on Build GRCh38
rs532685645 Chr.12: 109769423-109769423 on Build GRCh38
rs144369209 Chr.12: 109769536-109769536 on Build GRCh38
rs965510316 Chr.12: 109769765-109769765 on Build GRCh38
rs565211115 Chr.12: 109769752-109769753 on Build GRCh38
rs559978000 Chr.12: 109769438-109769438 on Build GRCh38
rs570064789 Chr.12: 109769585-109769585 on Build GRCh38
rs955940850 Chr.12: 109769698-109769698 on Build GRCh38
rs912324087 Chr.12: 109769473-109769473 on Build GRCh38
rs779312617 Chr.12: 109769740-109769740 on Build GRCh38
rs547709868 Chr.12: 109769429-109769429 on Build GRCh38
rs986759559 Chr.12: 109769442-109769442 on Build GRCh38
rs1007581428 Chr.12: 109769693-109769694 on Build GRCh38
rs1012933617 Chr.12: 109769627-109769627 on Build GRCh38
rs941916218 Chr.12: 109769868-109769868 on Build GRCh38
rs1049343141 Chr.12: 109769547-109769547 on Build GRCh38
rs1002923608 Chr.12: 109769687-109769687 on Build GRCh38
rs897667263 Chr.12: 109769622-109769622 on Build GRCh38
rs115059906 Chr.12: 109769727-109769727 on Build GRCh38

View Full Product Details