Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 197523214-197523812 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPE1-MOB4
Entrez Gene ID
100529241
Gene Name
HSPE1-MOB4 readthrough
Gene Aliases
HSPE1-PHOCN
UniGene
Hs.633165
Gene Chromosome Location
Chr.2: 197499997-197553699 on Build GRCh38
Transcripts
NM_001202485.1 NM_001100819.2 NM_001204094.1 NM_015387.4 NM_199482.3

Gene Symbol
MOB4
Entrez Gene ID
25843
Gene Name
MOB family member 4, phocein
Gene Aliases
2C4D, CGI-95, MOB1, MOB3, MOBKL3, PHOCN, PREI3
UniGene
Hs.633165
Gene Chromosome Location
Chr.2: 197515571-197553699 on Build GRCh38
Transcripts
NM_001202485.1 NM_001100819.2 NM_001204094.1 NM_015387.4 NM_199482.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7290789 c.61-6T>A p.? Chr.2: 197523618-197523618 on Build GRCh38
1404301 c.77C>A p.P26H Chr.2: 197523640-197523640 on Build GRCh38
1614100 c.76C>T p.P26S Chr.2: 197523639-197523639 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1043303956 Chr.2: 197523538-197523538 on Build GRCh38
rs750822367 Chr.2: 197523763-197523763 on Build GRCh38
rs555276132 Chr.2: 197523267-197523267 on Build GRCh38
rs768922790 Chr.2: 197523653-197523653 on Build GRCh38
rs372104828 Chr.2: 197523732-197523732 on Build GRCh38
rs907241043 Chr.2: 197523452-197523452 on Build GRCh38
rs1026563480 Chr.2: 197523349-197523349 on Build GRCh38
rs980164105 Chr.2: 197523451-197523451 on Build GRCh38
rs746529407 Chr.2: 197523317-197523317 on Build GRCh38
rs1055943592 Chr.2: 197523484-197523484 on Build GRCh38
rs773682236 Chr.2: 197523612-197523612 on Build GRCh38
rs767505239 Chr.2: 197523609-197523609 on Build GRCh38
rs185320958 Chr.2: 197523247-197523247 on Build GRCh38
rs762880404 Chr.2: 197523644-197523644 on Build GRCh38
rs750951017 Chr.2: 197523573-197523573 on Build GRCh38
rs575073295 Chr.2: 197523426-197523426 on Build GRCh38
rs950669859 Chr.2: 197523350-197523350 on Build GRCh38
rs1001710646 Chr.2: 197523453-197523453 on Build GRCh38
rs1053327807 Chr.2: 197523509-197523509 on Build GRCh38
rs548615502 Chr.2: 197523296-197523296 on Build GRCh38
rs774417627 Chr.2: 197523495-197523495 on Build GRCh38
rs1020941575 Chr.2: 197523541-197523541 on Build GRCh38
rs957574567 Chr.2: 197523246-197523246 on Build GRCh38
rs908902136 Chr.2: 197523381-197523381 on Build GRCh38
rs1019665660 Chr.2: 197523281-197523281 on Build GRCh38
rs1045853338 Chr.2: 197523436-197523436 on Build GRCh38
rs938430315 Chr.2: 197523499-197523499 on Build GRCh38
rs190181503 Chr.2: 197523437-197523437 on Build GRCh38
rs746874716 Chr.2: 197523598-197523598 on Build GRCh38
rs35917320 Chr.2: 197523602-197523603 on Build GRCh38
rs540371188 Chr.2: 197523591-197523591 on Build GRCh38
rs774428873 Chr.2: 197523677-197523677 on Build GRCh38
rs373819349 Chr.2: 197523605-197523605 on Build GRCh38
rs375428652 Chr.2: 197523631-197523631 on Build GRCh38
rs984633614 Chr.2: 197523352-197523352 on Build GRCh38
rs202101608 Chr.2: 197523578-197523578 on Build GRCh38
rs368401341 Chr.2: 197523729-197523729 on Build GRCh38
rs769683553 Chr.2: 197523614-197523614 on Build GRCh38
rs757844625 Chr.2: 197523731-197523731 on Build GRCh38
rs147955934 Chr.2: 197523536-197523536 on Build GRCh38
rs777224040 Chr.2: 197523597-197523597 on Build GRCh38
rs923012850 Chr.2: 197523288-197523288 on Build GRCh38
rs370842176 Chr.2: 197523311-197523311 on Build GRCh38
rs975963676 Chr.2: 197523342-197523342 on Build GRCh38
rs1019935711 Chr.2: 197523257-197523257 on Build GRCh38
rs941929389 Chr.2: 197523351-197523351 on Build GRCh38
rs985947502 Chr.2: 197523748-197523748 on Build GRCh38
rs1036219252 Chr.2: 197523377-197523377 on Build GRCh38
rs374385112 Chr.2: 197523303-197523307 on Build GRCh38
rs767969805 Chr.2: 197523711-197523711 on Build GRCh38
rs765766386 Chr.2: 197523546-197523546 on Build GRCh38
rs751005457 Chr.2: 197523712-197523712 on Build GRCh38
rs879795151 Chr.2: 197523343-197523343 on Build GRCh38
rs368168359 Chr.2: 197523730-197523730 on Build GRCh38
rs754582109 Chr.2: 197523575-197523575 on Build GRCh38
rs762323082 Chr.2: 197523705-197523705 on Build GRCh38
rs971990431 Chr.2: 197523779-197523779 on Build GRCh38
rs771688254 Chr.2: 197523254-197523254 on Build GRCh38
rs778369826 Chr.2: 197523735-197523736 on Build GRCh38
rs891938263 Chr.2: 197523512-197523512 on Build GRCh38
rs967104068 Chr.2: 197523547-197523547 on Build GRCh38
rs761295048 Chr.2: 197523717-197523717 on Build GRCh38
rs953243281 Chr.2: 197523309-197523309 on Build GRCh38
rs963168987 Chr.2: 197523773-197523773 on Build GRCh38
rs909073884 Chr.2: 197523754-197523754 on Build GRCh38
rs77102528 Chr.2: 197523606-197523606 on Build GRCh38
rs928429604 Chr.2: 197523481-197523481 on Build GRCh38
rs76963731 Chr.2: 197523262-197523262 on Build GRCh38
rs745777224 Chr.2: 197523607-197523607 on Build GRCh38
rs370712205 Chr.2: 197523586-197523586 on Build GRCh38

View Full Product Details