Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 35280835-35281361 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF76
Entrez Gene ID
7629
Gene Name
zinc finger protein 76
Gene Aliases
D6S229E, ZNF523, Zfp523
UniGene
Hs.388024
Gene Chromosome Location
Chr.6: 35259473-35295987 on Build GRCh38
Transcripts
NM_001292032.1 NM_003427.4 XM_011514846.2 XM_011514847.2 XM_011514849.2 XM_011514850.2 XM_011514854.2 XM_011514855.2 XM_017011248.1 XM_017011249.1 XM_017011250.1 XM_017011251.1 XM_017011252.1 XM_017011253.1 XM_017011254.1 XM_017011255.1 XM_017011256.1 XM_017011257.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4586688 c.1_2insT p.? Chr.6: 35281152-35281153 on Build GRCh38
4797132 c.42G>A p.G14G Chr.6: 35281193-35281193 on Build GRCh38
6222759 c.24G>A p.T8T Chr.6: 35281175-35281175 on Build GRCh38
7310716 c.63A>G p.Q21Q Chr.6: 35281214-35281214 on Build GRCh38
3697808 c.55G>A p.V19I Chr.6: 35281206-35281206 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs775483063 Chr.6: 35281202-35281202 on Build GRCh38
rs1017834638 Chr.6: 35281100-35281100 on Build GRCh38
rs576747479 Chr.6: 35281292-35281292 on Build GRCh38
rs750061659 Chr.6: 35281150-35281150 on Build GRCh38
rs766192182 Chr.6: 35281096-35281096 on Build GRCh38
rs969403881 Chr.6: 35281110-35281110 on Build GRCh38
rs1044987267 Chr.6: 35280946-35280946 on Build GRCh38
rs955664628 Chr.6: 35281026-35281026 on Build GRCh38
rs755003951 Chr.6: 35281170-35281170 on Build GRCh38
rs765879229 Chr.6: 35281255-35281255 on Build GRCh38
rs375121325 Chr.6: 35281118-35281118 on Build GRCh38
rs143421103 Chr.6: 35281193-35281193 on Build GRCh38
rs144522603 Chr.6: 35281206-35281206 on Build GRCh38
rs753463181 Chr.6: 35281259-35281259 on Build GRCh38
rs944764327 Chr.6: 35281181-35281181 on Build GRCh38
rs934758581 Chr.6: 35280865-35280865 on Build GRCh38
rs1013073436 Chr.6: 35280891-35280891 on Build GRCh38
rs897557750 Chr.6: 35281008-35281008 on Build GRCh38
rs1023580600 Chr.6: 35280978-35280978 on Build GRCh38
rs949029285 Chr.6: 35281328-35281328 on Build GRCh38
rs552938519 Chr.6: 35281330-35281330 on Build GRCh38
rs990239351 Chr.6: 35281033-35281033 on Build GRCh38
rs772868996 Chr.6: 35280987-35280987 on Build GRCh38
rs751962571 Chr.6: 35281116-35281116 on Build GRCh38
rs538630278 Chr.6: 35281013-35281013 on Build GRCh38
rs572000205 Chr.6: 35281006-35281006 on Build GRCh38
rs565997420 Chr.6: 35281152-35281152 on Build GRCh38
rs76952654 Chr.6: 35281214-35281214 on Build GRCh38
rs1037332434 Chr.6: 35281321-35281321 on Build GRCh38
rs772006422 Chr.6: 35281185-35281185 on Build GRCh38
rs746106907 Chr.6: 35281198-35281198 on Build GRCh38
rs113888808 Chr.6: 35281211-35281211 on Build GRCh38
rs550482430 Chr.6: 35280979-35280979 on Build GRCh38
rs755747457 Chr.6: 35281248-35281248 on Build GRCh38
rs747959025 Chr.6: 35280889-35280889 on Build GRCh38
rs779465419 Chr.6: 35281153-35281153 on Build GRCh38
rs540630437 Chr.6: 35281320-35281320 on Build GRCh38
rs554687025 Chr.6: 35281180-35281180 on Build GRCh38
rs954126141 Chr.6: 35281136-35281136 on Build GRCh38
rs949256205 Chr.6: 35280914-35280914 on Build GRCh38
rs750056338 Chr.6: 35281225-35281225 on Build GRCh38
rs375995502 Chr.6: 35281174-35281174 on Build GRCh38
rs368577134 Chr.6: 35281208-35281208 on Build GRCh38
rs769697243 Chr.6: 35280954-35280954 on Build GRCh38
rs1045332000 Chr.6: 35281283-35281283 on Build GRCh38
rs547783389 Chr.6: 35281030-35281030 on Build GRCh38
rs762992645 Chr.6: 35281005-35281005 on Build GRCh38
rs954848769 Chr.6: 35281041-35281041 on Build GRCh38
rs763419976 Chr.6: 35281205-35281205 on Build GRCh38
rs780881460 Chr.6: 35281129-35281129 on Build GRCh38
rs201061431 Chr.6: 35281137-35281137 on Build GRCh38
rs1021511428 Chr.6: 35281134-35281134 on Build GRCh38
rs767781945 Chr.6: 35281215-35281215 on Build GRCh38
rs139970455 Chr.6: 35281163-35281163 on Build GRCh38

View Full Product Details