Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 142367481-142368030 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRG6
Entrez Gene ID
57211
Gene Name
adhesion G protein-coupled receptor G6
Gene Aliases
APG1, DREG, GPR126, LCCS9, PS1TP2, VIGR
UniGene
Hs.743302
Gene Chromosome Location
Chr.6: 142301919-142446266 on Build GRCh38
Transcripts
NM_001032394.2 NM_001032395.2 NM_020455.5 NM_198569.2 XM_005267061.3 XM_006715516.2 XM_006715517.2 XM_006715518.2 XM_011535964.1 XM_017011085.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5518699 c.125G>A p.R42Q Chr.6: 142367590-142367590 on Build GRCh38
5662012 c.221C>T p.P74L Chr.6: 142367686-142367686 on Build GRCh38
6470805 c.224C>A p.T75N Chr.6: 142367689-142367689 on Build GRCh38
1208626 c.285T>G p.I95M Chr.6: 142367750-142367750 on Build GRCh38
258653 c.287A>C p.Y96S Chr.6: 142367752-142367752 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747867805 Chr.6: 142367642-142367642 on Build GRCh38
rs200175575 Chr.6: 142367562-142367562 on Build GRCh38
rs548140209 Chr.6: 142367603-142367603 on Build GRCh38
rs375452464 Chr.6: 142367718-142367718 on Build GRCh38
rs1052324066 Chr.6: 142367937-142367937 on Build GRCh38
rs529155878 Chr.6: 142367599-142367599 on Build GRCh38
rs769227436 Chr.6: 142367788-142367788 on Build GRCh38
rs1046301794 Chr.6: 142367524-142367524 on Build GRCh38
rs552622032 Chr.6: 142367651-142367651 on Build GRCh38
rs984281460 Chr.6: 142367786-142367786 on Build GRCh38
rs529096589 Chr.6: 142367946-142367946 on Build GRCh38
rs754147976 Chr.6: 142367594-142367594 on Build GRCh38
rs766540867 Chr.6: 142367704-142367704 on Build GRCh38
rs751587735 Chr.6: 142367716-142367716 on Build GRCh38
rs756981737 Chr.6: 142367902-142367902 on Build GRCh38
rs561557732 Chr.6: 142367934-142367934 on Build GRCh38
rs745415054 Chr.6: 142367911-142367911 on Build GRCh38
rs894895318 Chr.6: 142367500-142367500 on Build GRCh38
rs377682546 Chr.6: 142367836-142367836 on Build GRCh38
rs370942737 Chr.6: 142367854-142367854 on Build GRCh38
rs749548167 Chr.6: 142367550-142367551 on Build GRCh38
rs192254609 Chr.6: 142367680-142367680 on Build GRCh38
rs774136698 Chr.6: 142367987-142367988 on Build GRCh38
rs771941015 Chr.6: 142367563-142367565 on Build GRCh38
rs777776056 Chr.6: 142367887-142367887 on Build GRCh38
rs949281059 Chr.6: 142367517-142367517 on Build GRCh38
rs753176909 Chr.6: 142367578-142367578 on Build GRCh38
rs562474541 Chr.6: 142367963-142367963 on Build GRCh38
rs768166001 Chr.6: 142367566-142367566 on Build GRCh38
rs530897758 Chr.6: 142367985-142367985 on Build GRCh38
rs746811790 Chr.6: 142367533-142367533 on Build GRCh38
rs202040079 Chr.6: 142367667-142367667 on Build GRCh38
rs116955726 Chr.6: 142367674-142367674 on Build GRCh38
rs376097107 Chr.6: 142367539-142367539 on Build GRCh38
rs544100145 Chr.6: 142367960-142367960 on Build GRCh38
rs754986214 Chr.6: 142367717-142367717 on Build GRCh38
rs748002231 Chr.6: 142367727-142367727 on Build GRCh38
rs373562749 Chr.6: 142367692-142367692 on Build GRCh38
rs751711883 Chr.6: 142367833-142367833 on Build GRCh38
rs140723393 Chr.6: 142367819-142367819 on Build GRCh38
rs770504097 Chr.6: 142367657-142367657 on Build GRCh38
rs777208798 Chr.6: 142367920-142367920 on Build GRCh38
rs35699755 Chr.6: 142367690-142367690 on Build GRCh38
rs374848824 Chr.6: 142367617-142367617 on Build GRCh38
rs762170852 Chr.6: 142367808-142367808 on Build GRCh38
rs1052713376 Chr.6: 142367629-142367629 on Build GRCh38
rs762533791 Chr.6: 142367563-142367563 on Build GRCh38
rs749058357 Chr.6: 142367893-142367893 on Build GRCh38
rs749003590 Chr.6: 142367758-142367758 on Build GRCh38
rs756263268 Chr.6: 142367538-142367539 on Build GRCh38
rs758433134 Chr.6: 142367608-142367608 on Build GRCh38
rs750259708 Chr.6: 142367579-142367579 on Build GRCh38
rs201862423 Chr.6: 142367605-142367605 on Build GRCh38
rs988988423 Chr.6: 142367899-142367899 on Build GRCh38
rs756103287 Chr.6: 142367859-142367859 on Build GRCh38
rs367816376 Chr.6: 142367584-142367584 on Build GRCh38
rs561621715 Chr.6: 142367555-142367555 on Build GRCh38
rs12173993 Chr.6: 142367687-142367687 on Build GRCh38
rs553605945 Chr.6: 142367675-142367675 on Build GRCh38
rs202204069 Chr.6: 142367931-142367931 on Build GRCh38
rs375234396 Chr.6: 142367689-142367689 on Build GRCh38
rs773950881 Chr.6: 142367772-142367772 on Build GRCh38
rs745914625 Chr.6: 142367520-142367520 on Build GRCh38
rs371346154 Chr.6: 142367518-142367518 on Build GRCh38
rs771416765 Chr.6: 142367914-142367914 on Build GRCh38
rs781112000 Chr.6: 142367633-142367633 on Build GRCh38
rs755936718 Chr.6: 142367737-142367737 on Build GRCh38
rs779916470 Chr.6: 142367529-142367529 on Build GRCh38
rs376684988 Chr.6: 142367837-142367837 on Build GRCh38
rs746908868 Chr.6: 142367624-142367624 on Build GRCh38
rs969949780 Chr.6: 142367933-142367934 on Build GRCh38
rs138992283 Chr.6: 142367643-142367643 on Build GRCh38
rs17280293 Chr.6: 142367832-142367832 on Build GRCh38
rs1018382501 Chr.6: 142367707-142367707 on Build GRCh38
rs75679810 Chr.6: 142367905-142367905 on Build GRCh38
rs368529000 Chr.6: 142367743-142367743 on Build GRCh38
rs770283884 Chr.6: 142367932-142367932 on Build GRCh38
rs776163478 Chr.6: 142367551-142367551 on Build GRCh38
rs370872563 Chr.6: 142367830-142367830 on Build GRCh38
rs780389547 Chr.6: 142367546-142367549 on Build GRCh38
rs772891975 Chr.6: 142367559-142367559 on Build GRCh38
rs776962394 Chr.6: 142367792-142367792 on Build GRCh38
rs980360126 Chr.6: 142367913-142367913 on Build GRCh38
rs927292104 Chr.6: 142367622-142367622 on Build GRCh38
rs369762357 Chr.6: 142367783-142367783 on Build GRCh38
rs762553859 Chr.6: 142367656-142367656 on Build GRCh38
rs770700301 Chr.6: 142367764-142367764 on Build GRCh38
rs917394949 Chr.6: 142367901-142367901 on Build GRCh38
rs768465606 Chr.6: 142367630-142367630 on Build GRCh38
rs780706631 Chr.6: 142367508-142367508 on Build GRCh38
rs376059023 Chr.6: 142367828-142367828 on Build GRCh38
rs761586203 Chr.6: 142367553-142367553 on Build GRCh38
rs758486246 Chr.6: 142367703-142367703 on Build GRCh38

View Full Product Details