Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 58146517-58147049 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM110B
Entrez Gene ID
90362
Gene Name
family with sequence similarity 110 member B
Gene Aliases
C8orf72
UniGene
Hs.154652
Gene Chromosome Location
Chr.8: 57994444-58149718 on Build GRCh38
Transcripts
NM_147189.2 XM_005251324.2 XM_005251325.3 XM_005251326.2 XM_017013948.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6658080 c.814G>A p.A272T Chr.8: 58147044-58147044 on Build GRCh38
275043 c.723C>T p.A241A Chr.8: 58146953-58146953 on Build GRCh38
4498516 c.521C>T p.P174L Chr.8: 58146751-58146751 on Build GRCh38
2719671 c.602C>T p.S201L Chr.8: 58146832-58146832 on Build GRCh38
5409730 c.647C>T p.P216L Chr.8: 58146877-58146877 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1051973816 Chr.8: 58146849-58146849 on Build GRCh38
rs757100650 Chr.8: 58146591-58146591 on Build GRCh38
rs868676674 Chr.8: 58146554-58146554 on Build GRCh38
rs368070087 Chr.8: 58146674-58146674 on Build GRCh38
rs766614164 Chr.8: 58146561-58146561 on Build GRCh38
rs372969421 Chr.8: 58146929-58146929 on Build GRCh38
rs772874905 Chr.8: 58146880-58146880 on Build GRCh38
rs999027649 Chr.8: 58146847-58146847 on Build GRCh38
rs183366212 Chr.8: 58146752-58146752 on Build GRCh38
rs200606254 Chr.8: 58146801-58146801 on Build GRCh38
rs372946396 Chr.8: 58146715-58146715 on Build GRCh38
rs758690171 Chr.8: 58146921-58146921 on Build GRCh38
rs755553412 Chr.8: 58146930-58146930 on Build GRCh38
rs765086827 Chr.8: 58146673-58146673 on Build GRCh38
rs903232922 Chr.8: 58146841-58146841 on Build GRCh38
rs761588397 Chr.8: 58146740-58146740 on Build GRCh38
rs755776342 Chr.8: 58146578-58146578 on Build GRCh38
rs995106560 Chr.8: 58146978-58146978 on Build GRCh38
rs764848330 Chr.8: 58146741-58146741 on Build GRCh38
rs578133975 Chr.8: 58146697-58146697 on Build GRCh38
rs766479168 Chr.8: 58147027-58147027 on Build GRCh38
rs150740446 Chr.8: 58146870-58146870 on Build GRCh38
rs767139353 Chr.8: 58146575-58146575 on Build GRCh38
rs765357865 Chr.8: 58146908-58146908 on Build GRCh38
rs142134782 Chr.8: 58146790-58146790 on Build GRCh38
rs191940292 Chr.8: 58146607-58146607 on Build GRCh38
rs748440229 Chr.8: 58146536-58146536 on Build GRCh38
rs750518242 Chr.8: 58147019-58147019 on Build GRCh38
rs760186756 Chr.8: 58146635-58146635 on Build GRCh38
rs752249974 Chr.8: 58146576-58146576 on Build GRCh38
rs34235613 Chr.8: 58146669-58146669 on Build GRCh38
rs767330523 Chr.8: 58146903-58146903 on Build GRCh38
rs1050875030 Chr.8: 58146701-58146701 on Build GRCh38
rs762536865 Chr.8: 58146882-58146882 on Build GRCh38
rs749004407 Chr.8: 58146953-58146953 on Build GRCh38
rs778624249 Chr.8: 58146602-58146602 on Build GRCh38
rs777975066 Chr.8: 58146773-58146773 on Build GRCh38
rs757443076 Chr.8: 58146906-58146906 on Build GRCh38
rs138142337 Chr.8: 58146655-58146655 on Build GRCh38
rs559377618 Chr.8: 58146927-58146927 on Build GRCh38
rs369797232 Chr.8: 58146814-58146814 on Build GRCh38
rs908478846 Chr.8: 58147017-58147017 on Build GRCh38
rs746209150 Chr.8: 58146719-58146719 on Build GRCh38
rs201466528 Chr.8: 58146657-58146657 on Build GRCh38
rs767303162 Chr.8: 58146985-58146985 on Build GRCh38
rs145048107 Chr.8: 58146943-58146943 on Build GRCh38
rs760061732 Chr.8: 58146572-58146572 on Build GRCh38
rs753848783 Chr.8: 58146781-58146781 on Build GRCh38
rs142742538 Chr.8: 58146618-58146618 on Build GRCh38
rs772689608 Chr.8: 58146637-58146637 on Build GRCh38
rs573032382 Chr.8: 58146957-58146957 on Build GRCh38
rs769033676 Chr.8: 58146855-58146855 on Build GRCh38
rs992230368 Chr.8: 58146932-58146932 on Build GRCh38
rs771339526 Chr.8: 58146629-58146629 on Build GRCh38
rs199616146 Chr.8: 58146748-58146748 on Build GRCh38
rs763347134 Chr.8: 58146559-58146559 on Build GRCh38
rs765481261 Chr.8: 58146995-58146995 on Build GRCh38
rs967805762 Chr.8: 58146794-58146794 on Build GRCh38
rs967773157 Chr.8: 58146568-58146568 on Build GRCh38
rs755184938 Chr.8: 58146728-58146729 on Build GRCh38
rs760630107 Chr.8: 58146991-58146991 on Build GRCh38
rs759220388 Chr.8: 58146980-58146980 on Build GRCh38
rs762768864 Chr.8: 58146749-58146749 on Build GRCh38
rs771272946 Chr.8: 58146708-58146708 on Build GRCh38
rs1018390985 Chr.8: 58146634-58146634 on Build GRCh38
rs747622051 Chr.8: 58146854-58146854 on Build GRCh38
rs759729835 Chr.8: 58146818-58146818 on Build GRCh38
rs776004086 Chr.8: 58146733-58146733 on Build GRCh38
rs199778704 Chr.8: 58146731-58146731 on Build GRCh38
rs753990670 Chr.8: 58146904-58146904 on Build GRCh38
rs147552704 Chr.8: 58146977-58146977 on Build GRCh38
rs781584797 Chr.8: 58146935-58146935 on Build GRCh38
rs750681364 Chr.8: 58146914-58146914 on Build GRCh38
rs753614030 Chr.8: 58146586-58146586 on Build GRCh38
rs763776582 Chr.8: 58146584-58146584 on Build GRCh38
rs748608099 Chr.8: 58146936-58146936 on Build GRCh38
rs773498874 Chr.8: 58146558-58146558 on Build GRCh38
rs373901437 Chr.8: 58146672-58146672 on Build GRCh38
rs774009638 Chr.8: 58146899-58146899 on Build GRCh38
rs1019228834 Chr.8: 58146734-58146734 on Build GRCh38
rs759571587 Chr.8: 58146902-58146902 on Build GRCh38
rs758379762 Chr.8: 58146677-58146677 on Build GRCh38
rs146051624 Chr.8: 58146628-58146628 on Build GRCh38
rs772639877 Chr.8: 58146946-58146946 on Build GRCh38
rs775018870 Chr.8: 58146571-58146571 on Build GRCh38
rs770472744 Chr.8: 58146956-58146956 on Build GRCh38
rs775311156 Chr.8: 58146990-58146990 on Build GRCh38
rs369509423 Chr.8: 58146703-58146703 on Build GRCh38
rs543657981 Chr.8: 58146729-58146729 on Build GRCh38
rs773517395 Chr.8: 58146622-58146622 on Build GRCh38
rs375271536 Chr.8: 58146619-58146619 on Build GRCh38
rs766164756 Chr.8: 58146895-58146895 on Build GRCh38
rs776257329 Chr.8: 58146866-58146866 on Build GRCh38
rs758743192 Chr.8: 58147026-58147026 on Build GRCh38
rs148663255 Chr.8: 58146692-58146692 on Build GRCh38
rs138169488 Chr.8: 58146614-58146614 on Build GRCh38
rs369051680 Chr.8: 58146922-58146922 on Build GRCh38
rs756242593 Chr.8: 58146696-58146696 on Build GRCh38
rs779936754 Chr.8: 58146685-58146685 on Build GRCh38
rs779242800 Chr.8: 58146789-58146789 on Build GRCh38
rs770044019 Chr.8: 58146557-58146557 on Build GRCh38
rs757598715 Chr.8: 58146787-58146787 on Build GRCh38
rs142623322 Chr.8: 58146665-58146665 on Build GRCh38
rs750259825 Chr.8: 58146742-58146742 on Build GRCh38
rs753974775 Chr.8: 58146716-58146727 on Build GRCh38

View Full Product Details